Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96649533-96650030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
UniGene
Hs.534025
Gene Chromosome Location
Chr.2: 96642719-96704887 on Build GRCh38
Transcripts
NM_001293083.1 XM_011512110.2 XM_011512111.2 XM_011512113.2 XM_011512114.2 XM_011512115.2 XM_011512116.2 XM_011512117.2 XM_011512118.2 XM_011512119.2 XM_011512120.2 XM_011512122.2 XM_011512125.2 XM_011512126.2 XM_011512127.2 XM_011512128.2 XM_017005222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1725174 c.137T>A p.V46D Chr.2: 96649630-96649630 on Build GRCh38
4930786 c.135T>C p.T45T Chr.2: 96649628-96649628 on Build GRCh38
4141309 c.180G>T p.K60N Chr.2: 96649673-96649673 on Build GRCh38
4141308 c.390G>T p.K130N Chr.2: 96649673-96649673 on Build GRCh38
4930785 c.345T>C p.T115T Chr.2: 96649628-96649628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767699403 Chr.2: 96649716-96649716 on Build GRCh38
rs962418534 Chr.2: 96649909-96649909 on Build GRCh38
rs529924152 Chr.2: 96649875-96649875 on Build GRCh38
rs991295068 Chr.2: 96649915-96649915 on Build GRCh38
rs17685664 Chr.2: 96649833-96649833 on Build GRCh38
rs902750759 Chr.2: 96649923-96649923 on Build GRCh38
rs764644529 Chr.2: 96649666-96649666 on Build GRCh38
rs1049886206 Chr.2: 96649893-96649893 on Build GRCh38
rs533272823 Chr.2: 96649902-96649902 on Build GRCh38
rs998814851 Chr.2: 96649690-96649690 on Build GRCh38
rs893194874 Chr.2: 96649711-96649711 on Build GRCh38
rs763901808 Chr.2: 96649566-96649566 on Build GRCh38
rs1027452564 Chr.2: 96649670-96649670 on Build GRCh38
rs1003224428 Chr.2: 96649946-96649946 on Build GRCh38
rs377603016 Chr.2: 96649658-96649658 on Build GRCh38
rs774974090 Chr.2: 96649707-96649707 on Build GRCh38
rs775852542 Chr.2: 96649629-96649629 on Build GRCh38
rs950214911 Chr.2: 96649927-96649927 on Build GRCh38
rs377607845 Chr.2: 96649785-96649785 on Build GRCh38
rs367651018 Chr.2: 96649743-96649743 on Build GRCh38
rs976706455 Chr.2: 96649829-96649829 on Build GRCh38
rs770550418 Chr.2: 96649598-96649598 on Build GRCh38
rs951319591 Chr.2: 96649892-96649892 on Build GRCh38
rs751463175 Chr.2: 96649680-96649680 on Build GRCh38
rs915378102 Chr.2: 96649899-96649899 on Build GRCh38
rs750009484 Chr.2: 96649855-96649855 on Build GRCh38
rs1018505699 Chr.2: 96649807-96649807 on Build GRCh38
rs776496251 Chr.2: 96649957-96649957 on Build GRCh38
rs185249959 Chr.2: 96649832-96649832 on Build GRCh38
rs1003191617 Chr.2: 96649577-96649577 on Build GRCh38
rs982021766 Chr.2: 96649928-96649928 on Build GRCh38
rs902217063 Chr.2: 96649571-96649571 on Build GRCh38
rs757226336 Chr.2: 96649820-96649820 on Build GRCh38
rs934911281 Chr.2: 96649983-96649983 on Build GRCh38
rs772006315 Chr.2: 96649601-96649601 on Build GRCh38
rs542834527 Chr.2: 96649649-96649649 on Build GRCh38
rs558247781 Chr.2: 96649997-96649998 on Build GRCh38
rs763254732 Chr.2: 96649632-96649632 on Build GRCh38
rs542504019 Chr.2: 96649922-96649922 on Build GRCh38
rs1034796816 Chr.2: 96649583-96649583 on Build GRCh38
rs1042668639 Chr.2: 96649918-96649918 on Build GRCh38
rs114180580 Chr.2: 96649709-96649709 on Build GRCh38
rs1007670438 Chr.2: 96649769-96649769 on Build GRCh38
rs377188714 Chr.2: 96649938-96649938 on Build GRCh38
rs774885487 Chr.2: 96649821-96649821 on Build GRCh38

View Full Product Details