Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96650096-96650589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
UniGene
Hs.534025
Gene Chromosome Location
Chr.2: 96642719-96704887 on Build GRCh38
Transcripts
NM_001293083.1 XM_011512110.2 XM_011512111.2 XM_011512113.2 XM_011512114.2 XM_011512115.2 XM_011512116.2 XM_011512117.2 XM_011512118.2 XM_011512119.2 XM_011512120.2 XM_011512122.2 XM_011512125.2 XM_011512126.2 XM_011512127.2 XM_011512128.2 XM_017005222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4437480 c.419C>T p.T140M Chr.2: 96650204-96650204 on Build GRCh38
2825685 c.484T>A p.S162T Chr.2: 96650269-96650269 on Build GRCh38
5670830 c.264G>A p.R88R Chr.2: 96650259-96650259 on Build GRCh38
2825686 c.274T>A p.S92T Chr.2: 96650269-96650269 on Build GRCh38
5914930 c.185-5C>T p.? Chr.2: 96650175-96650175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016775107 Chr.2: 96650533-96650533 on Build GRCh38
rs997989824 Chr.2: 96650139-96650139 on Build GRCh38
rs558143302 Chr.2: 96650359-96650359 on Build GRCh38
rs981602534 Chr.2: 96650344-96650344 on Build GRCh38
rs758400250 Chr.2: 96650266-96650266 on Build GRCh38
rs36034205 Chr.2: 96650177-96650178 on Build GRCh38
rs1022676232 Chr.2: 96650289-96650289 on Build GRCh38
rs777327299 Chr.2: 96650325-96650325 on Build GRCh38
rs1004079553 Chr.2: 96650532-96650532 on Build GRCh38
rs990439675 Chr.2: 96650433-96650433 on Build GRCh38
rs947701947 Chr.2: 96650529-96650529 on Build GRCh38
rs770573191 Chr.2: 96650335-96650335 on Build GRCh38
rs150933838 Chr.2: 96650455-96650455 on Build GRCh38
rs866082435 Chr.2: 96650149-96650149 on Build GRCh38
rs933429802 Chr.2: 96650497-96650497 on Build GRCh38
rs1029464583 Chr.2: 96650182-96650182 on Build GRCh38
rs754791525 Chr.2: 96650222-96650222 on Build GRCh38
rs1050867853 Chr.2: 96650502-96650502 on Build GRCh38
rs1005102763 Chr.2: 96650127-96650127 on Build GRCh38
rs1048825612 Chr.2: 96650557-96650557 on Build GRCh38
rs377426781 Chr.2: 96650252-96650252 on Build GRCh38
rs760894128 Chr.2: 96650204-96650204 on Build GRCh38
rs901898324 Chr.2: 96650486-96650486 on Build GRCh38
rs368306646 Chr.2: 96650383-96650383 on Build GRCh38
rs534764279 Chr.2: 96650126-96650126 on Build GRCh38
rs886982165 Chr.2: 96650531-96650531 on Build GRCh38
rs868140710 Chr.2: 96650538-96650538 on Build GRCh38
rs946934125 Chr.2: 96650337-96650337 on Build GRCh38
rs922566028 Chr.2: 96650262-96650262 on Build GRCh38
rs373410981 Chr.2: 96650208-96650208 on Build GRCh38
rs962441369 Chr.2: 96650248-96650248 on Build GRCh38
rs557262821 Chr.2: 96650524-96650524 on Build GRCh38
rs954136761 Chr.2: 96650296-96650296 on Build GRCh38
rs766274317 Chr.2: 96650284-96650285 on Build GRCh38
rs774812636 Chr.2: 96650449-96650449 on Build GRCh38
rs537044745 Chr.2: 96650474-96650474 on Build GRCh38
rs766851338 Chr.2: 96650333-96650333 on Build GRCh38
rs978637992 Chr.2: 96650355-96650355 on Build GRCh38
rs934878939 Chr.2: 96650401-96650401 on Build GRCh38
rs752602865 Chr.2: 96650253-96650253 on Build GRCh38
rs201179043 Chr.2: 96650312-96650312 on Build GRCh38
rs924209169 Chr.2: 96650369-96650369 on Build GRCh38
rs1004110571 Chr.2: 96650205-96650205 on Build GRCh38
rs1039447839 Chr.2: 96650475-96650475 on Build GRCh38
rs887508089 Chr.2: 96650562-96650562 on Build GRCh38
rs750526466 Chr.2: 96650151-96650151 on Build GRCh38
rs768113507 Chr.2: 96650558-96650558 on Build GRCh38

View Full Product Details