Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96816967-96817473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM3
Entrez Gene ID
26505
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Gene Aliases
ACDP3
UniGene
Hs.711617
Gene Chromosome Location
Chr.2: 96815894-96837109 on Build GRCh38
Transcripts
NM_017623.4 NM_199078.2 XM_011510957.2 XM_017003800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
419058 c.1155C>G p.F385L Chr.2: 96817432-96817432 on Build GRCh38
5947662 c.1166_1187del22 p.L390fs*23 Chr.2: 96817443-96817464 on Build GRCh38
5957091 c.1125C>A p.D375E Chr.2: 96817402-96817402 on Build GRCh38
7111230 c.1029G>A p.Q343Q Chr.2: 96817306-96817306 on Build GRCh38
133443 c.916_917AA>CT p.K306L Chr.2: 96817193-96817194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566949738 Chr.2: 96817237-96817237 on Build GRCh38
rs1043885396 Chr.2: 96816995-96816995 on Build GRCh38
rs761871186 Chr.2: 96817401-96817401 on Build GRCh38
rs763500068 Chr.2: 96817028-96817028 on Build GRCh38
rs758383435 Chr.2: 96817236-96817236 on Build GRCh38
rs750648325 Chr.2: 96817126-96817126 on Build GRCh38
rs756950333 Chr.2: 96817420-96817420 on Build GRCh38
rs867129720 Chr.2: 96817253-96817253 on Build GRCh38
rs552657529 Chr.2: 96817344-96817344 on Build GRCh38
rs970127104 Chr.2: 96817107-96817107 on Build GRCh38
rs761858710 Chr.2: 96817072-96817072 on Build GRCh38
rs12995235 Chr.2: 96817191-96817191 on Build GRCh38
rs763919255 Chr.2: 96817393-96817393 on Build GRCh38
rs747679335 Chr.2: 96817159-96817159 on Build GRCh38
rs532654967 Chr.2: 96817135-96817135 on Build GRCh38
rs767453014 Chr.2: 96817200-96817200 on Build GRCh38
rs891405826 Chr.2: 96817033-96817033 on Build GRCh38
rs144557284 Chr.2: 96817437-96817437 on Build GRCh38
rs748934807 Chr.2: 96817191-96817196 on Build GRCh38
rs760811058 Chr.2: 96817277-96817277 on Build GRCh38
rs370077039 Chr.2: 96817262-96817262 on Build GRCh38
rs892256757 Chr.2: 96817249-96817249 on Build GRCh38
rs766572645 Chr.2: 96817279-96817279 on Build GRCh38
rs939302507 Chr.2: 96817121-96817121 on Build GRCh38
rs770244604 Chr.2: 96817010-96817010 on Build GRCh38
rs767084300 Chr.2: 96817402-96817402 on Build GRCh38
rs769179461 Chr.2: 96817261-96817261 on Build GRCh38
rs765129663 Chr.2: 96817232-96817232 on Build GRCh38
rs149921109 Chr.2: 96817343-96817343 on Build GRCh38
rs759786951 Chr.2: 96817292-96817292 on Build GRCh38
rs770627299 Chr.2: 96817343-96817343 on Build GRCh38
rs926205265 Chr.2: 96817011-96817023 on Build GRCh38
rs1008481900 Chr.2: 96817140-96817140 on Build GRCh38
rs775834165 Chr.2: 96817450-96817450 on Build GRCh38
rs11687092 Chr.2: 96817171-96817171 on Build GRCh38
rs764162708 Chr.2: 96817327-96817327 on Build GRCh38
rs764705972 Chr.2: 96817032-96817032 on Build GRCh38
rs760825307 Chr.2: 96817213-96817213 on Build GRCh38
rs538556488 Chr.2: 96817368-96817368 on Build GRCh38
rs138281711 Chr.2: 96817225-96817225 on Build GRCh38
rs754842613 Chr.2: 96817157-96817157 on Build GRCh38
rs755651729 Chr.2: 96817409-96817409 on Build GRCh38
rs376328896 Chr.2: 96817357-96817357 on Build GRCh38
rs202012890 Chr.2: 96817230-96817230 on Build GRCh38
rs1031961443 Chr.2: 96817224-96817224 on Build GRCh38
rs770720977 Chr.2: 96817182-96817182 on Build GRCh38
rs374358338 Chr.2: 96817222-96817222 on Build GRCh38
rs748550967 Chr.2: 96817267-96817267 on Build GRCh38
rs762867969 Chr.2: 96817389-96817389 on Build GRCh38
rs769897406 Chr.2: 96817447-96817447 on Build GRCh38
rs752990296 Chr.2: 96817417-96817417 on Build GRCh38
rs373041892 Chr.2: 96817271-96817271 on Build GRCh38
rs573097940 Chr.2: 96817346-96817346 on Build GRCh38
rs773360282 Chr.2: 96817211-96817211 on Build GRCh38
rs756385956 Chr.2: 96817127-96817127 on Build GRCh38
rs370817712 Chr.2: 96817382-96817382 on Build GRCh38
rs763574789 Chr.2: 96817284-96817284 on Build GRCh38
rs749741199 Chr.2: 96817187-96817187 on Build GRCh38
rs753328913 Chr.2: 96817388-96817388 on Build GRCh38
rs1028492919 Chr.2: 96817056-96817056 on Build GRCh38
rs753647148 Chr.2: 96817139-96817139 on Build GRCh38
rs997431775 Chr.2: 96817055-96817055 on Build GRCh38
rs749962291 Chr.2: 96817404-96817404 on Build GRCh38
rs1024648988 Chr.2: 96817276-96817276 on Build GRCh38
rs891300122 Chr.2: 96817163-96817163 on Build GRCh38
rs767754950 Chr.2: 96817125-96817125 on Build GRCh38
rs778221126 Chr.2: 96817423-96817423 on Build GRCh38
rs151151816 Chr.2: 96817425-96817425 on Build GRCh38
rs746594885 Chr.2: 96817180-96817180 on Build GRCh38
rs148629978 Chr.2: 96817369-96817369 on Build GRCh38
rs755850728 Chr.2: 96817361-96817361 on Build GRCh38
rs752549461 Chr.2: 96817317-96817317 on Build GRCh38
rs981054026 Chr.2: 96817111-96817111 on Build GRCh38
rs147430912 Chr.2: 96817240-96817240 on Build GRCh38
rs747068163 Chr.2: 96817373-96817373 on Build GRCh38
rs775935042 Chr.2: 96817185-96817185 on Build GRCh38
rs1015625758 Chr.2: 96817076-96817076 on Build GRCh38
rs926452477 Chr.2: 96817112-96817112 on Build GRCh38
rs368247965 Chr.2: 96817365-96817365 on Build GRCh38
rs1026985678 Chr.2: 96817351-96817351 on Build GRCh38
rs778887659 Chr.2: 96817158-96817158 on Build GRCh38
rs868585007 Chr.2: 96817394-96817394 on Build GRCh38
rs200171507 Chr.2: 96817189-96817189 on Build GRCh38
rs779823178 Chr.2: 96817413-96817413 on Build GRCh38
rs759697136 Chr.2: 96817384-96817384 on Build GRCh38
rs200517700 Chr.2: 96817169-96817169 on Build GRCh38
rs770960372 Chr.2: 96817378-96817378 on Build GRCh38
rs751590296 Chr.2: 96817396-96817396 on Build GRCh38
rs945050546 Chr.2: 96817155-96817155 on Build GRCh38
rs772643290 Chr.2: 96817270-96817270 on Build GRCh38
rs920011811 Chr.2: 96817103-96817103 on Build GRCh38
rs955641564 Chr.2: 96817063-96817063 on Build GRCh38
rs1008470648 Chr.2: 96817071-96817071 on Build GRCh38
rs761919472 Chr.2: 96817196-96817196 on Build GRCh38
rs752538461 Chr.2: 96817234-96817234 on Build GRCh38
rs749651383 Chr.2: 96817252-96817252 on Build GRCh38
rs746536994 Chr.2: 96817238-96817238 on Build GRCh38
rs1056294896 Chr.2: 96817122-96817122 on Build GRCh38
rs373639442 Chr.2: 96817177-96817177 on Build GRCh38
rs776019140 Chr.2: 96817027-96817027 on Build GRCh38
rs764894369 Chr.2: 96817315-96817315 on Build GRCh38
rs781182520 Chr.2: 96817442-96817442 on Build GRCh38
rs751658323 Chr.2: 96817042-96817042 on Build GRCh38
rs866589246 Chr.2: 96817296-96817296 on Build GRCh38
rs183035620 Chr.2: 96817320-96817320 on Build GRCh38
rs781051911 Chr.2: 96817250-96817250 on Build GRCh38
rs746015520 Chr.2: 96817443-96817443 on Build GRCh38
rs776886146 Chr.2: 96817286-96817286 on Build GRCh38
rs974070090 Chr.2: 96817099-96817099 on Build GRCh38
rs891280907 Chr.2: 96817181-96817181 on Build GRCh38
rs960546793 Chr.2: 96817433-96817433 on Build GRCh38

View Full Product Details