Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96827611-96828129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM3
Entrez Gene ID
26505
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Gene Aliases
ACDP3
UniGene
Hs.711617
Gene Chromosome Location
Chr.2: 96815894-96837109 on Build GRCh38
Transcripts
NM_017623.4 NM_199078.2 XM_011510957.2 XM_017003800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138312 c.1552G>A p.E518K Chr.2: 96827763-96827763 on Build GRCh38
290305 c.1711G>A p.G571R Chr.2: 96828120-96828120 on Build GRCh38
6565523 c.1680C>T p.L560L Chr.2: 96827891-96827891 on Build GRCh38
6439428 c.1660C>T p.P554S Chr.2: 96827871-96827871 on Build GRCh38
6631329 c.1569C>T p.H523H Chr.2: 96827780-96827780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898168947 Chr.2: 96827998-96827998 on Build GRCh38
rs368815174 Chr.2: 96828049-96828049 on Build GRCh38
rs563778358 Chr.2: 96827739-96827739 on Build GRCh38
rs940428248 Chr.2: 96827830-96827830 on Build GRCh38
rs140550487 Chr.2: 96827754-96827754 on Build GRCh38
rs773988981 Chr.2: 96827854-96827854 on Build GRCh38
rs777762408 Chr.2: 96827827-96827827 on Build GRCh38
rs778311053 Chr.2: 96827894-96827894 on Build GRCh38
rs925953854 Chr.2: 96827850-96827850 on Build GRCh38
rs763642553 Chr.2: 96827755-96827755 on Build GRCh38
rs753679398 Chr.2: 96827686-96827686 on Build GRCh38
rs1045355546 Chr.2: 96828002-96828002 on Build GRCh38
rs762920032 Chr.2: 96827846-96827846 on Build GRCh38
rs752608693 Chr.2: 96827711-96827712 on Build GRCh38
rs777610222 Chr.2: 96827791-96827792 on Build GRCh38
rs779520248 Chr.2: 96827721-96827721 on Build GRCh38
rs769584782 Chr.2: 96827845-96827845 on Build GRCh38
rs781154754 Chr.2: 96827943-96827943 on Build GRCh38
rs995381773 Chr.2: 96827918-96827918 on Build GRCh38
rs753062919 Chr.2: 96827882-96827882 on Build GRCh38
rs937118901 Chr.2: 96827985-96827985 on Build GRCh38
rs752226422 Chr.2: 96827984-96827984 on Build GRCh38
rs563932286 Chr.2: 96827828-96827828 on Build GRCh38
rs899439925 Chr.2: 96827632-96827632 on Build GRCh38
rs147683550 Chr.2: 96827768-96827768 on Build GRCh38
rs765285754 Chr.2: 96827688-96827688 on Build GRCh38
rs201760396 Chr.2: 96827701-96827701 on Build GRCh38
rs746109031 Chr.2: 96827840-96827840 on Build GRCh38
rs777459316 Chr.2: 96827707-96827707 on Build GRCh38
rs757287932 Chr.2: 96827911-96827911 on Build GRCh38
rs375599243 Chr.2: 96827832-96827832 on Build GRCh38
rs995110953 Chr.2: 96827661-96827661 on Build GRCh38
rs758918218 Chr.2: 96827823-96827823 on Build GRCh38
rs570023249 Chr.2: 96828035-96828035 on Build GRCh38
rs956662672 Chr.2: 96828050-96828050 on Build GRCh38
rs974859500 Chr.2: 96828079-96828079 on Build GRCh38
rs372666354 Chr.2: 96827872-96827872 on Build GRCh38
rs149545001 Chr.2: 96827895-96827895 on Build GRCh38
rs775353845 Chr.2: 96828054-96828054 on Build GRCh38
rs751460213 Chr.2: 96827741-96827741 on Build GRCh38
rs559375951 Chr.2: 96827822-96827822 on Build GRCh38
rs755741624 Chr.2: 96827874-96827874 on Build GRCh38
rs982104142 Chr.2: 96827697-96827697 on Build GRCh38
rs371647288 Chr.2: 96827724-96827724 on Build GRCh38
rs755407214 Chr.2: 96827798-96827798 on Build GRCh38
rs185649363 Chr.2: 96827815-96827815 on Build GRCh38
rs372553939 Chr.2: 96828069-96828069 on Build GRCh38
rs575970892 Chr.2: 96827736-96827736 on Build GRCh38
rs780960640 Chr.2: 96827776-96827776 on Build GRCh38
rs201846760 Chr.2: 96827750-96827750 on Build GRCh38
rs890225259 Chr.2: 96827818-96827818 on Build GRCh38
rs913521415 Chr.2: 96827873-96827873 on Build GRCh38
rs755801523 Chr.2: 96828013-96828013 on Build GRCh38
rs751211274 Chr.2: 96827712-96827712 on Build GRCh38
rs768696883 Chr.2: 96828067-96828067 on Build GRCh38
rs766378650 Chr.2: 96827699-96827706 on Build GRCh38
rs758901872 Chr.2: 96827885-96827885 on Build GRCh38
rs1038408995 Chr.2: 96827990-96827990 on Build GRCh38
rs988672208 Chr.2: 96827717-96827717 on Build GRCh38
rs376394445 Chr.2: 96827862-96827862 on Build GRCh38
rs549843750 Chr.2: 96828030-96828030 on Build GRCh38
rs1024269124 Chr.2: 96827668-96827668 on Build GRCh38
rs766223488 Chr.2: 96827704-96827704 on Build GRCh38
rs752127621 Chr.2: 96827693-96827693 on Build GRCh38
rs757552747 Chr.2: 96828023-96828023 on Build GRCh38
rs1045883645 Chr.2: 96827635-96827635 on Build GRCh38
rs921809362 Chr.2: 96827764-96827764 on Build GRCh38
rs748020518 Chr.2: 96827979-96827979 on Build GRCh38
rs756986436 Chr.2: 96827716-96827716 on Build GRCh38
rs545895724 Chr.2: 96827921-96827922 on Build GRCh38
rs754125772 Chr.2: 96827780-96827780 on Build GRCh38
rs200102903 Chr.2: 96827833-96827833 on Build GRCh38
rs776286499 Chr.2: 96827744-96827744 on Build GRCh38
rs765578171 Chr.2: 96827876-96827876 on Build GRCh38
rs545903554 Chr.2: 96827719-96827719 on Build GRCh38
rs78693080 Chr.2: 96828036-96828036 on Build GRCh38
rs374635848 Chr.2: 96827702-96827702 on Build GRCh38
rs756073103 Chr.2: 96827729-96827729 on Build GRCh38
rs748142488 Chr.2: 96827722-96827722 on Build GRCh38
rs141442711 Chr.2: 96828048-96828048 on Build GRCh38
rs368520967 Chr.2: 96827749-96827749 on Build GRCh38
rs371837473 Chr.2: 96827849-96827849 on Build GRCh38
rs533286911 Chr.2: 96827955-96827955 on Build GRCh38
rs749157649 Chr.2: 96828066-96828066 on Build GRCh38
rs1051375019 Chr.2: 96827791-96827791 on Build GRCh38
rs780503858 Chr.2: 96827718-96827718 on Build GRCh38
rs375803257 Chr.2: 96828077-96828077 on Build GRCh38
rs899348026 Chr.2: 96827903-96827903 on Build GRCh38
rs772609359 Chr.2: 96828094-96828094 on Build GRCh38
rs767042346 Chr.2: 96827863-96827863 on Build GRCh38
rs760306737 Chr.2: 96828104-96828104 on Build GRCh38
rs751121625 Chr.2: 96827756-96827756 on Build GRCh38
rs773300851 Chr.2: 96827729-96827729 on Build GRCh38
rs766111921 Chr.2: 96827680-96827680 on Build GRCh38

View Full Product Details