Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96828724-96829214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM3
Entrez Gene ID
26505
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Gene Aliases
ACDP3
UniGene
Hs.711617
Gene Chromosome Location
Chr.2: 96815894-96837109 on Build GRCh38
Transcripts
NM_017623.4 NM_199078.2 XM_011510957.2 XM_017003800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1023993 c.1980G>T p.Q660H Chr.2: 96829055-96829055 on Build GRCh38
6631331 c.1996G>A p.D666N Chr.2: 96829071-96829071 on Build GRCh38
4673151 c.2054C>T p.A685V Chr.2: 96829129-96829129 on Build GRCh38
2825845 c.1925C>T p.T642M Chr.2: 96829000-96829000 on Build GRCh38
419057 c.1961G>A p.R654Q Chr.2: 96829036-96829036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369816313 Chr.2: 96828973-96828973 on Build GRCh38
rs756457541 Chr.2: 96829156-96829156 on Build GRCh38
rs200572566 Chr.2: 96829001-96829001 on Build GRCh38
rs945605719 Chr.2: 96828867-96828867 on Build GRCh38
rs774514356 Chr.2: 96829014-96829014 on Build GRCh38
rs907261063 Chr.2: 96829050-96829050 on Build GRCh38
rs376441799 Chr.2: 96828922-96828922 on Build GRCh38
rs781707117 Chr.2: 96828989-96828989 on Build GRCh38
rs758109971 Chr.2: 96829143-96829143 on Build GRCh38
rs982941895 Chr.2: 96828955-96828955 on Build GRCh38
rs372006609 Chr.2: 96828743-96828743 on Build GRCh38
rs753492511 Chr.2: 96828966-96828966 on Build GRCh38
rs1046683347 Chr.2: 96829038-96829038 on Build GRCh38
rs376507747 Chr.2: 96829070-96829070 on Build GRCh38
rs770397651 Chr.2: 96829000-96829000 on Build GRCh38
rs375303580 Chr.2: 96828745-96828745 on Build GRCh38
rs202016851 Chr.2: 96829126-96829126 on Build GRCh38
rs763532366 Chr.2: 96828845-96828845 on Build GRCh38
rs138088588 Chr.2: 96828965-96828965 on Build GRCh38
rs780302104 Chr.2: 96829165-96829165 on Build GRCh38
rs369097574 Chr.2: 96829018-96829018 on Build GRCh38
rs535195074 Chr.2: 96828916-96828916 on Build GRCh38
rs764783462 Chr.2: 96829129-96829129 on Build GRCh38
rs762041604 Chr.2: 96829016-96829016 on Build GRCh38
rs753458682 Chr.2: 96828962-96828962 on Build GRCh38
rs749740500 Chr.2: 96829181-96829181 on Build GRCh38
rs776592449 Chr.2: 96829123-96829123 on Build GRCh38
rs995719072 Chr.2: 96828917-96828917 on Build GRCh38
rs758128177 Chr.2: 96829045-96829045 on Build GRCh38
rs752321314 Chr.2: 96829142-96829142 on Build GRCh38
rs752348135 Chr.2: 96829037-96829037 on Build GRCh38
rs1043952852 Chr.2: 96828902-96828902 on Build GRCh38
rs552078704 Chr.2: 96828795-96828795 on Build GRCh38
rs771383235 Chr.2: 96828984-96828984 on Build GRCh38
rs894479147 Chr.2: 96829108-96829108 on Build GRCh38
rs745544210 Chr.2: 96829130-96829130 on Build GRCh38
rs746309611 Chr.2: 96828999-96828999 on Build GRCh38
rs1041794599 Chr.2: 96828800-96828800 on Build GRCh38
rs550854662 Chr.2: 96828808-96828808 on Build GRCh38
rs555377174 Chr.2: 96828923-96828923 on Build GRCh38
rs571685984 Chr.2: 96828925-96828925 on Build GRCh38
rs147409622 Chr.2: 96828803-96828803 on Build GRCh38
rs376792071 Chr.2: 96828784-96828784 on Build GRCh38
rs760570294 Chr.2: 96829117-96829117 on Build GRCh38
rs563336716 Chr.2: 96829107-96829107 on Build GRCh38
rs866306085 Chr.2: 96829146-96829146 on Build GRCh38
rs1002858030 Chr.2: 96828835-96828835 on Build GRCh38
rs142725857 Chr.2: 96828797-96828797 on Build GRCh38
rs763886263 Chr.2: 96829153-96829153 on Build GRCh38
rs140443890 Chr.2: 96829035-96829035 on Build GRCh38
rs750227113 Chr.2: 96829023-96829023 on Build GRCh38
rs761632895 Chr.2: 96828949-96828949 on Build GRCh38
rs148612997 Chr.2: 96829121-96829121 on Build GRCh38
rs747347943 Chr.2: 96828981-96828981 on Build GRCh38
rs202173651 Chr.2: 96828980-96828980 on Build GRCh38
rs774423218 Chr.2: 96829106-96829106 on Build GRCh38
rs1053583140 Chr.2: 96828753-96828753 on Build GRCh38
rs373268251 Chr.2: 96829057-96829057 on Build GRCh38
rs751286696 Chr.2: 96828751-96828751 on Build GRCh38
rs374701098 Chr.2: 96829002-96829002 on Build GRCh38
rs750307064 Chr.2: 96828957-96828957 on Build GRCh38
rs201129132 Chr.2: 96829155-96829155 on Build GRCh38
rs142499606 Chr.2: 96829071-96829071 on Build GRCh38
rs772350650 Chr.2: 96829109-96829109 on Build GRCh38
rs768728722 Chr.2: 96829077-96829077 on Build GRCh38
rs149301704 Chr.2: 96829036-96829036 on Build GRCh38
rs767327041 Chr.2: 96828956-96828956 on Build GRCh38
rs960425645 Chr.2: 96829079-96829079 on Build GRCh38
rs914180155 Chr.2: 96828860-96828860 on Build GRCh38
rs951541163 Chr.2: 96828941-96828941 on Build GRCh38
rs760604647 Chr.2: 96829034-96829034 on Build GRCh38
rs755441003 Chr.2: 96829182-96829182 on Build GRCh38
rs188826295 Chr.2: 96828871-96828871 on Build GRCh38
rs756063823 Chr.2: 96828958-96828958 on Build GRCh38
rs1055763971 Chr.2: 96829104-96829104 on Build GRCh38
rs201082018 Chr.2: 96829111-96829111 on Build GRCh38
rs143763352 Chr.2: 96829052-96829052 on Build GRCh38
rs200109873 Chr.2: 96829003-96829003 on Build GRCh38

View Full Product Details