Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97083856-97084359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD2B
Entrez Gene ID
151313
Gene Name
fumarylacetoacetate hydrolase domain containing 2B
Gene Aliases
-
UniGene
Hs.567723
Gene Chromosome Location
Chr.2: 97080738-97094882 on Build GRCh38
Transcripts
NM_001320848.1 NM_199336.2 XM_011510745.2 XM_011510746.2 XM_011510747.2 XM_011510748.2 XM_011510750.2 XM_017003470.1 XM_017003471.1 XM_017003472.1 XM_017003473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1410121 c.709_711delTGC p.C237delC Chr.2: 97084252-97084254 on Build GRCh38
6092735 c.724G>T p.E242* Chr.2: 97084239-97084239 on Build GRCh38
722811 c.757G>A p.V253I Chr.2: 97084206-97084206 on Build GRCh38
7260994 c.843A>G p.P281P Chr.2: 97083987-97083987 on Build GRCh38
1326911 c.751C>A p.Q251K Chr.2: 97084212-97084212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047764343 Chr.2: 97084082-97084082 on Build GRCh38
rs769493043 Chr.2: 97084250-97084250 on Build GRCh38
rs768628477 Chr.2: 97083996-97083996 on Build GRCh38
rs748293779 Chr.2: 97084017-97084017 on Build GRCh38
rs768371180 Chr.2: 97084326-97084326 on Build GRCh38
rs757040095 Chr.2: 97084261-97084261 on Build GRCh38
rs775549903 Chr.2: 97084251-97084251 on Build GRCh38
rs778003216 Chr.2: 97084121-97084122 on Build GRCh38
rs752426119 Chr.2: 97084295-97084295 on Build GRCh38
rs781452898 Chr.2: 97083983-97083983 on Build GRCh38
rs1031295197 Chr.2: 97084284-97084284 on Build GRCh38
rs754773697 Chr.2: 97084134-97084134 on Build GRCh38
rs771490149 Chr.2: 97084053-97084062 on Build GRCh38
rs755120149 Chr.2: 97083988-97083988 on Build GRCh38
rs759874895 Chr.2: 97083938-97083938 on Build GRCh38
rs762162971 Chr.2: 97084152-97084152 on Build GRCh38
rs370319389 Chr.2: 97083923-97083923 on Build GRCh38
rs750119093 Chr.2: 97083932-97083932 on Build GRCh38
rs745398719 Chr.2: 97084327-97084327 on Build GRCh38
rs867487547 Chr.2: 97084090-97084090 on Build GRCh38
rs755254004 Chr.2: 97084180-97084180 on Build GRCh38
rs557448239 Chr.2: 97084234-97084234 on Build GRCh38
rs140352445 Chr.2: 97084013-97084013 on Build GRCh38
rs757216055 Chr.2: 97084326-97084326 on Build GRCh38
rs374707075 Chr.2: 97084002-97084002 on Build GRCh38
rs955704740 Chr.2: 97084309-97084309 on Build GRCh38
rs375434642 Chr.2: 97084239-97084239 on Build GRCh38
rs772737653 Chr.2: 97084011-97084011 on Build GRCh38
rs1012725609 Chr.2: 97084156-97084156 on Build GRCh38
rs778502141 Chr.2: 97084199-97084199 on Build GRCh38
rs773854720 Chr.2: 97083913-97083913 on Build GRCh38
rs758251307 Chr.2: 97084215-97084215 on Build GRCh38
rs777664545 Chr.2: 97084307-97084307 on Build GRCh38
rs774224775 Chr.2: 97084000-97084000 on Build GRCh38
rs772048794 Chr.2: 97084138-97084138 on Build GRCh38
rs781434604 Chr.2: 97083996-97083998 on Build GRCh38
rs377270443 Chr.2: 97084139-97084139 on Build GRCh38
rs535387721 Chr.2: 97084108-97084108 on Build GRCh38
rs764630873 Chr.2: 97084047-97084047 on Build GRCh38
rs997920413 Chr.2: 97084068-97084068 on Build GRCh38
rs762944762 Chr.2: 97083908-97083908 on Build GRCh38
rs898308074 Chr.2: 97084142-97084142 on Build GRCh38
rs780481770 Chr.2: 97084126-97084126 on Build GRCh38
rs779929013 Chr.2: 97083994-97083994 on Build GRCh38
rs375760322 Chr.2: 97083981-97083981 on Build GRCh38
rs759616753 Chr.2: 97084280-97084280 on Build GRCh38
rs747985950 Chr.2: 97084209-97084209 on Build GRCh38
rs757804155 Chr.2: 97084057-97084057 on Build GRCh38
rs765543173 Chr.2: 97083948-97083948 on Build GRCh38
rs576107978 Chr.2: 97084271-97084271 on Build GRCh38
rs751654514 Chr.2: 97083980-97083980 on Build GRCh38
rs768151716 Chr.2: 97084158-97084158 on Build GRCh38
rs754060056 Chr.2: 97084174-97084174 on Build GRCh38
rs769317792 Chr.2: 97084328-97084328 on Build GRCh38
rs767198921 Chr.2: 97083925-97083925 on Build GRCh38
rs756332304 Chr.2: 97084167-97084167 on Build GRCh38
rs367665505 Chr.2: 97084058-97084058 on Build GRCh38
rs534709393 Chr.2: 97084216-97084216 on Build GRCh38
rs746861856 Chr.2: 97084301-97084301 on Build GRCh38
rs879120167 Chr.2: 97083907-97083907 on Build GRCh38
rs745470134 Chr.2: 97084247-97084247 on Build GRCh38
rs766682338 Chr.2: 97084170-97084170 on Build GRCh38
rs558162125 Chr.2: 97084133-97084133 on Build GRCh38
rs745797387 Chr.2: 97084133-97084138 on Build GRCh38
rs778890641 Chr.2: 97084136-97084136 on Build GRCh38
rs369223989 Chr.2: 97084056-97084056 on Build GRCh38
rs752884099 Chr.2: 97084021-97084022 on Build GRCh38
rs372972282 Chr.2: 97084140-97084140 on Build GRCh38
rs145484440 Chr.2: 97084270-97084270 on Build GRCh38
rs548829930 Chr.2: 97084233-97084233 on Build GRCh38
rs758226009 Chr.2: 97084297-97084297 on Build GRCh38
rs556055479 Chr.2: 97083949-97083949 on Build GRCh38
rs951366701 Chr.2: 97083880-97083880 on Build GRCh38
rs756346594 Chr.2: 97084087-97084087 on Build GRCh38
rs774811524 Chr.2: 97084150-97084150 on Build GRCh38
rs773899048 Chr.2: 97084264-97084264 on Build GRCh38
rs751447150 Chr.2: 97084311-97084311 on Build GRCh38
rs113222096 Chr.2: 97083993-97083993 on Build GRCh38
rs778903553 Chr.2: 97084312-97084312 on Build GRCh38
rs778201348 Chr.2: 97083979-97083979 on Build GRCh38
rs768483720 Chr.2: 97083911-97083911 on Build GRCh38
rs185017037 Chr.2: 97084096-97084096 on Build GRCh38
rs766029852 Chr.2: 97084014-97084014 on Build GRCh38
rs779973879 Chr.2: 97084230-97084231 on Build GRCh38
rs764316293 Chr.2: 97084141-97084141 on Build GRCh38
rs1054721818 Chr.2: 97084026-97084026 on Build GRCh38
rs765551519 Chr.2: 97084291-97084291 on Build GRCh38
rs763293469 Chr.2: 97084042-97084042 on Build GRCh38
rs772703870 Chr.2: 97084257-97084257 on Build GRCh38
rs758842463 Chr.2: 97083953-97083953 on Build GRCh38
rs766713691 Chr.2: 97083908-97083917 on Build GRCh38
rs762296956 Chr.2: 97084252-97084252 on Build GRCh38
rs1031642627 Chr.2: 97084066-97084066 on Build GRCh38
rs777750406 Chr.2: 97084232-97084232 on Build GRCh38
rs775815149 Chr.2: 97084038-97084038 on Build GRCh38
rs772196498 Chr.2: 97084136-97084138 on Build GRCh38
rs751966944 Chr.2: 97083987-97083988 on Build GRCh38
rs753985714 Chr.2: 97084275-97084275 on Build GRCh38
rs58875686 Chr.2: 97084137-97084137 on Build GRCh38
rs750536561 Chr.2: 97084063-97084063 on Build GRCh38

View Full Product Details