Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97091529-97092038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD2B
Entrez Gene ID
151313
Gene Name
fumarylacetoacetate hydrolase domain containing 2B
Gene Aliases
-
UniGene
Hs.567723
Gene Chromosome Location
Chr.2: 97080738-97094882 on Build GRCh38
Transcripts
NM_001320848.1 NM_001320849.1 NM_199336.2 XM_011510745.2 XM_011510746.2 XM_011510747.2 XM_011510748.2 XM_011510750.2 XM_017003470.1 XM_017003471.1 XM_017003472.1 XM_017003473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5978413 c.93G>C p.Q31H Chr.2: 97091614-97091614 on Build GRCh38
7157086 c.139G>A p.G47R Chr.2: 97091568-97091568 on Build GRCh38
1483391 c.49C>T p.Q17* Chr.2: 97091658-97091658 on Build GRCh38
4096457 c.176A>G p.D59G Chr.2: 97091531-97091531 on Build GRCh38
6159280 c.3G>T p.M1I Chr.2: 97091704-97091704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754846245 Chr.2: 97091586-97091586 on Build GRCh38
rs759704748 Chr.2: 97091669-97091669 on Build GRCh38
rs557053515 Chr.2: 97091601-97091601 on Build GRCh38
rs748584192 Chr.2: 97091728-97091728 on Build GRCh38
rs572162639 Chr.2: 97092017-97092017 on Build GRCh38
rs892003336 Chr.2: 97091910-97091910 on Build GRCh38
rs755833967 Chr.2: 97091703-97091703 on Build GRCh38
rs746944772 Chr.2: 97091641-97091641 on Build GRCh38
rs115068103 Chr.2: 97091757-97091757 on Build GRCh38
rs974778705 Chr.2: 97091953-97091953 on Build GRCh38
rs147575393 Chr.2: 97091677-97091677 on Build GRCh38
rs780858052 Chr.2: 97091728-97091728 on Build GRCh38
rs865918283 Chr.2: 97091680-97091680 on Build GRCh38
rs570524093 Chr.2: 97091802-97091802 on Build GRCh38
rs745494248 Chr.2: 97091684-97091684 on Build GRCh38
rs776965336 Chr.2: 97091665-97091665 on Build GRCh38
rs774888124 Chr.2: 97091623-97091623 on Build GRCh38
rs923249411 Chr.2: 97091752-97091752 on Build GRCh38
rs756793643 Chr.2: 97091817-97091817 on Build GRCh38
rs758334032 Chr.2: 97091609-97091609 on Build GRCh38
rs536375395 Chr.2: 97091991-97091991 on Build GRCh38
rs954703918 Chr.2: 97091985-97091985 on Build GRCh38
rs1038573560 Chr.2: 97091827-97091827 on Build GRCh38
rs555192064 Chr.2: 97091621-97091621 on Build GRCh38
rs541005193 Chr.2: 97091672-97091672 on Build GRCh38
rs779365261 Chr.2: 97091726-97091726 on Build GRCh38
rs939870401 Chr.2: 97091796-97091796 on Build GRCh38
rs550618430 Chr.2: 97091894-97091894 on Build GRCh38
rs542121159 Chr.2: 97091978-97091978 on Build GRCh38
rs983457501 Chr.2: 97091998-97091998 on Build GRCh38
rs770998419 Chr.2: 97091751-97091751 on Build GRCh38
rs145518288 Chr.2: 97091842-97091842 on Build GRCh38
rs577589666 Chr.2: 97091727-97091727 on Build GRCh38
rs564773691 Chr.2: 97091683-97091683 on Build GRCh38
rs537050527 Chr.2: 97091891-97091891 on Build GRCh38
rs201241480 Chr.2: 97091553-97091553 on Build GRCh38
rs1022181195 Chr.2: 97091930-97091930 on Build GRCh38
rs766684206 Chr.2: 97091584-97091584 on Build GRCh38
rs528766834 Chr.2: 97091835-97091835 on Build GRCh38
rs780101262 Chr.2: 97091595-97091603 on Build GRCh38
rs764083529 Chr.2: 97091676-97091676 on Build GRCh38
rs752498834 Chr.2: 97091673-97091673 on Build GRCh38
rs147890593 Chr.2: 97091610-97091610 on Build GRCh38
rs529037413 Chr.2: 97091768-97091768 on Build GRCh38
rs572285620 Chr.2: 97091625-97091625 on Build GRCh38
rs747040888 Chr.2: 97091747-97091747 on Build GRCh38
rs760754158 Chr.2: 97091582-97091582 on Build GRCh38
rs574332948 Chr.2: 97091975-97091975 on Build GRCh38
rs868733356 Chr.2: 97091681-97091681 on Build GRCh38
rs753575109 Chr.2: 97091585-97091585 on Build GRCh38
rs906849275 Chr.2: 97091870-97091870 on Build GRCh38
rs1011838850 Chr.2: 97091924-97091924 on Build GRCh38
rs370856399 Chr.2: 97091679-97091679 on Build GRCh38
rs752586731 Chr.2: 97091604-97091604 on Build GRCh38
rs141266955 Chr.2: 97091631-97091631 on Build GRCh38
rs571586259 Chr.2: 97091889-97091889 on Build GRCh38
rs778895012 Chr.2: 97091957-97091957 on Build GRCh38
rs956224847 Chr.2: 97091626-97091626 on Build GRCh38
rs563382692 Chr.2: 97091767-97091767 on Build GRCh38
rs141760069 Chr.2: 97091830-97091830 on Build GRCh38
rs760878256 Chr.2: 97091641-97091641 on Build GRCh38
rs1257084 Chr.2: 97091772-97091772 on Build GRCh38
rs750536625 Chr.2: 97091575-97091575 on Build GRCh38
rs201521399 Chr.2: 97091559-97091559 on Build GRCh38
rs754746214 Chr.2: 97091683-97091685 on Build GRCh38
rs149711241 Chr.2: 97091614-97091614 on Build GRCh38
rs773898977 Chr.2: 97091743-97091743 on Build GRCh38
rs556173542 Chr.2: 97091941-97091941 on Build GRCh38
rs773414590 Chr.2: 97091637-97091637 on Build GRCh38
rs780936470 Chr.2: 97091615-97091615 on Build GRCh38
rs1002569371 Chr.2: 97091878-97091878 on Build GRCh38
rs914748122 Chr.2: 97091652-97091652 on Build GRCh38
rs746578128 Chr.2: 97091611-97091611 on Build GRCh38
rs766483177 Chr.2: 97091656-97091656 on Build GRCh38
rs772704348 Chr.2: 97091742-97091742 on Build GRCh38
rs1031925668 Chr.2: 97091887-97091887 on Build GRCh38
rs948446845 Chr.2: 97091836-97091836 on Build GRCh38
rs137898672 Chr.2: 97092006-97092006 on Build GRCh38
rs567419163 Chr.2: 97091971-97091971 on Build GRCh38

View Full Product Details