Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 98391740-98392143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA3
Entrez Gene ID
1261
Gene Name
cyclic nucleotide gated channel alpha 3
Gene Aliases
ACHM2, CCNC1, CCNCa, CCNCalpha, CNCG3, CNG3
UniGene
Hs.234785
Gene Chromosome Location
Chr.2: 98346155-98399154 on Build GRCh38
Transcripts
NM_001079878.1 NM_001298.2 XM_006712243.2 XM_011510554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2914029 c.576C>T p.F192F Chr.2: 98391873-98391873 on Build GRCh38
1743038 c.627G>A p.S209S Chr.2: 98391924-98391924 on Build GRCh38
6631133 c.668G>A p.R223Q Chr.2: 98391965-98391965 on Build GRCh38
2914032 c.667C>T p.R223W Chr.2: 98391964-98391964 on Build GRCh38
4141324 c.624C>T p.Y208Y Chr.2: 98391921-98391921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781751772 Chr.2: 98391837-98391837 on Build GRCh38
rs770713600 Chr.2: 98391958-98391958 on Build GRCh38
rs889946926 Chr.2: 98392067-98392067 on Build GRCh38
rs748436031 Chr.2: 98391993-98391993 on Build GRCh38
rs1029807208 Chr.2: 98391827-98391827 on Build GRCh38
rs2271040 Chr.2: 98391791-98391791 on Build GRCh38
rs532298545 Chr.2: 98391808-98391808 on Build GRCh38
rs369896847 Chr.2: 98391830-98391830 on Build GRCh38
rs757650055 Chr.2: 98391906-98391906 on Build GRCh38
rs1051791901 Chr.2: 98392066-98392066 on Build GRCh38
rs923842843 Chr.2: 98391903-98391903 on Build GRCh38
rs758601557 Chr.2: 98392008-98392008 on Build GRCh38
rs928168608 Chr.2: 98391773-98391773 on Build GRCh38
rs866553549 Chr.2: 98391873-98391873 on Build GRCh38
rs771894602 Chr.2: 98391951-98391951 on Build GRCh38
rs898350927 Chr.2: 98392115-98392115 on Build GRCh38
rs545951813 Chr.2: 98391980-98391980 on Build GRCh38
rs998354244 Chr.2: 98392053-98392053 on Build GRCh38
rs780239212 Chr.2: 98391924-98391924 on Build GRCh38
rs766732326 Chr.2: 98391975-98391975 on Build GRCh38
rs562081454 Chr.2: 98392117-98392117 on Build GRCh38
rs753973144 Chr.2: 98391977-98391977 on Build GRCh38
rs946860150 Chr.2: 98391992-98391992 on Build GRCh38
rs553995770 Chr.2: 98391930-98391930 on Build GRCh38
rs533596832 Chr.2: 98391923-98391923 on Build GRCh38
rs947788175 Chr.2: 98391772-98391772 on Build GRCh38
rs770929126 Chr.2: 98392013-98392013 on Build GRCh38
rs772982333 Chr.2: 98391955-98391955 on Build GRCh38
rs968039620 Chr.2: 98391896-98391896 on Build GRCh38
rs893580169 Chr.2: 98391799-98391799 on Build GRCh38
rs542293960 Chr.2: 98392096-98392096 on Build GRCh38
rs749592128 Chr.2: 98391847-98391847 on Build GRCh38
rs375673700 Chr.2: 98391874-98391874 on Build GRCh38
rs540328507 Chr.2: 98391840-98391840 on Build GRCh38
rs572401814 Chr.2: 98392118-98392118 on Build GRCh38
rs766621130 Chr.2: 98392012-98392022 on Build GRCh38
rs768061703 Chr.2: 98391970-98391970 on Build GRCh38
rs556360697 Chr.2: 98392012-98392012 on Build GRCh38
rs779966971 Chr.2: 98391844-98391844 on Build GRCh38
rs1022375154 Chr.2: 98392097-98392097 on Build GRCh38
rs750806023 Chr.2: 98391971-98391971 on Build GRCh38
rs186227495 Chr.2: 98391774-98391774 on Build GRCh38
rs769970199 Chr.2: 98391843-98391843 on Build GRCh38
rs138958917 Chr.2: 98391964-98391964 on Build GRCh38
rs1004121380 Chr.2: 98392089-98392089 on Build GRCh38
rs143265049 Chr.2: 98391894-98391894 on Build GRCh38
rs748346243 Chr.2: 98391934-98391934 on Build GRCh38
rs200979604 Chr.2: 98391892-98391892 on Build GRCh38
rs751011572 Chr.2: 98391912-98391912 on Build GRCh38
rs141395850 Chr.2: 98391959-98391959 on Build GRCh38
rs376876249 Chr.2: 98391888-98391888 on Build GRCh38
rs760574115 Chr.2: 98391876-98391876 on Build GRCh38
rs887049504 Chr.2: 98392088-98392088 on Build GRCh38
rs576588154 Chr.2: 98392055-98392055 on Build GRCh38
rs372335705 Chr.2: 98391814-98391814 on Build GRCh38
rs537970293 Chr.2: 98391829-98391829 on Build GRCh38
rs377125146 Chr.2: 98391815-98391815 on Build GRCh38
rs188984295 Chr.2: 98391811-98391811 on Build GRCh38
rs928276131 Chr.2: 98391954-98391954 on Build GRCh38
rs778932631 Chr.2: 98391985-98391985 on Build GRCh38
rs774147269 Chr.2: 98391852-98391852 on Build GRCh38
rs776385883 Chr.2: 98392015-98392015 on Build GRCh38
rs373527569 Chr.2: 98391851-98391851 on Build GRCh38
rs374497578 Chr.2: 98391900-98391900 on Build GRCh38
rs781676123 Chr.2: 98391911-98391911 on Build GRCh38
rs762668060 Chr.2: 98391965-98391965 on Build GRCh38
rs572888940 Chr.2: 98391792-98391792 on Build GRCh38
rs1010598837 Chr.2: 98391804-98391804 on Build GRCh38
rs1051201455 Chr.2: 98392075-98392075 on Build GRCh38
rs761554853 Chr.2: 98391869-98391869 on Build GRCh38
rs754022156 Chr.2: 98392049-98392049 on Build GRCh38
rs377318981 Chr.2: 98391963-98391963 on Build GRCh38
rs901535728 Chr.2: 98392023-98392023 on Build GRCh38
rs1016832855 Chr.2: 98392105-98392105 on Build GRCh38
rs2271041 Chr.2: 98391889-98391889 on Build GRCh38
rs954358826 Chr.2: 98391841-98391841 on Build GRCh38
rs978112233 Chr.2: 98391897-98391897 on Build GRCh38
rs745971598 Chr.2: 98391842-98391842 on Build GRCh38
rs779055526 Chr.2: 98391932-98391932 on Build GRCh38

View Full Product Details