Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99360124-99360624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5B
Entrez Gene ID
9669
Gene Name
eukaryotic translation initiation factor 5B
Gene Aliases
IF2
UniGene
Hs.158688
Gene Chromosome Location
Chr.2: 99337371-99400265 on Build GRCh38
Transcripts
NM_015904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6092678 c.39C>T p.T13T Chr.2: 99360239-99360239 on Build GRCh38
1192769 c.142delA p.K50fs*10 Chr.2: 99360342-99360342 on Build GRCh38
6159243 c.151C>T p.Q51* Chr.2: 99360351-99360351 on Build GRCh38
1024347 c.142A>G p.K48E Chr.2: 99360342-99360342 on Build GRCh38
6653089 c.45T>C p.D15D Chr.2: 99360245-99360245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754621538 Chr.2: 99360507-99360507 on Build GRCh38
rs1001929690 Chr.2: 99360571-99360571 on Build GRCh38
rs370159221 Chr.2: 99360558-99360558 on Build GRCh38
rs201319020 Chr.2: 99360352-99360352 on Build GRCh38
rs761770096 Chr.2: 99360286-99360286 on Build GRCh38
rs750997151 Chr.2: 99360592-99360592 on Build GRCh38
rs770014660 Chr.2: 99360521-99360521 on Build GRCh38
rs761668754 Chr.2: 99360429-99360429 on Build GRCh38
rs758901642 Chr.2: 99360349-99360351 on Build GRCh38
rs755818023 Chr.2: 99360296-99360296 on Build GRCh38
rs757666404 Chr.2: 99360584-99360584 on Build GRCh38
rs202087026 Chr.2: 99360232-99360232 on Build GRCh38
rs373543039 Chr.2: 99360368-99360368 on Build GRCh38
rs910646309 Chr.2: 99360344-99360344 on Build GRCh38
rs879846696 Chr.2: 99360174-99360174 on Build GRCh38
rs753142117 Chr.2: 99360320-99360320 on Build GRCh38
rs780434811 Chr.2: 99360552-99360555 on Build GRCh38
rs765497615 Chr.2: 99360234-99360234 on Build GRCh38
rs775824937 Chr.2: 99360395-99360395 on Build GRCh38
rs759128069 Chr.2: 99360578-99360578 on Build GRCh38
rs374733590 Chr.2: 99360255-99360255 on Build GRCh38
rs775485454 Chr.2: 99360244-99360244 on Build GRCh38
rs766066634 Chr.2: 99360484-99360484 on Build GRCh38
rs370672608 Chr.2: 99360435-99360435 on Build GRCh38
rs747336625 Chr.2: 99360554-99360555 on Build GRCh38
rs376601754 Chr.2: 99360407-99360407 on Build GRCh38
rs767018176 Chr.2: 99360289-99360289 on Build GRCh38
rs140951041 Chr.2: 99360585-99360585 on Build GRCh38
rs904942416 Chr.2: 99360562-99360562 on Build GRCh38
rs760279243 Chr.2: 99360498-99360498 on Build GRCh38
rs768956059 Chr.2: 99360593-99360596 on Build GRCh38
rs753458115 Chr.2: 99360489-99360489 on Build GRCh38
rs780141118 Chr.2: 99360526-99360526 on Build GRCh38
rs41280587 Chr.2: 99360350-99360350 on Build GRCh38
rs1015949579 Chr.2: 99360575-99360575 on Build GRCh38
rs772985096 Chr.2: 99360206-99360206 on Build GRCh38
rs749997963 Chr.2: 99360457-99360457 on Build GRCh38
rs925928656 Chr.2: 99360379-99360379 on Build GRCh38
rs957008860 Chr.2: 99360291-99360291 on Build GRCh38
rs749476006 Chr.2: 99360530-99360530 on Build GRCh38
rs963308619 Chr.2: 99360237-99360237 on Build GRCh38
rs867591947 Chr.2: 99360470-99360470 on Build GRCh38
rs778008681 Chr.2: 99360514-99360514 on Build GRCh38
rs778152940 Chr.2: 99360334-99360334 on Build GRCh38
rs751935737 Chr.2: 99360582-99360582 on Build GRCh38
rs749961066 Chr.2: 99360342-99360342 on Build GRCh38
rs114030806 Chr.2: 99360541-99360541 on Build GRCh38
rs971945909 Chr.2: 99360249-99360249 on Build GRCh38
rs988866552 Chr.2: 99360292-99360292 on Build GRCh38
rs756731616 Chr.2: 99360312-99360313 on Build GRCh38
rs537016230 Chr.2: 99360264-99360264 on Build GRCh38
rs763184183 Chr.2: 99360406-99360406 on Build GRCh38
rs924203481 Chr.2: 99360190-99360190 on Build GRCh38
rs757897545 Chr.2: 99360516-99360516 on Build GRCh38
rs914281615 Chr.2: 99360328-99360328 on Build GRCh38
rs933151229 Chr.2: 99360487-99360487 on Build GRCh38
rs767424906 Chr.2: 99360445-99360445 on Build GRCh38
rs778446197 Chr.2: 99360341-99360342 on Build GRCh38
rs774127825 Chr.2: 99360408-99360408 on Build GRCh38
rs781763830 Chr.2: 99360517-99360517 on Build GRCh38
rs745987409 Chr.2: 99360519-99360519 on Build GRCh38
rs1053399154 Chr.2: 99360496-99360496 on Build GRCh38
rs756620581 Chr.2: 99360595-99360595 on Build GRCh38
rs879176976 Chr.2: 99360386-99360386 on Build GRCh38
rs369962372 Chr.2: 99360257-99360257 on Build GRCh38
rs764929776 Chr.2: 99360580-99360580 on Build GRCh38
rs574008986 Chr.2: 99360342-99360342 on Build GRCh38
rs571284174 Chr.2: 99360214-99360214 on Build GRCh38
rs189331174 Chr.2: 99360515-99360515 on Build GRCh38
rs779674930 Chr.2: 99360310-99360310 on Build GRCh38
rs954606506 Chr.2: 99360285-99360285 on Build GRCh38
rs368721938 Chr.2: 99360577-99360577 on Build GRCh38
rs200711395 Chr.2: 99360597-99360599 on Build GRCh38
rs772033897 Chr.2: 99360547-99360547 on Build GRCh38
rs373467460 Chr.2: 99360295-99360295 on Build GRCh38
rs151032660 Chr.2: 99360559-99360559 on Build GRCh38
rs751472493 Chr.2: 99360276-99360276 on Build GRCh38
rs758670640 Chr.2: 99360333-99360333 on Build GRCh38
rs773108895 Chr.2: 99360555-99360555 on Build GRCh38
rs868154681 Chr.2: 99360309-99360309 on Build GRCh38
rs768865403 Chr.2: 99360535-99360535 on Build GRCh38
rs557337566 Chr.2: 99360337-99360337 on Build GRCh38
rs763707555 Chr.2: 99360566-99360566 on Build GRCh38
rs552085788 Chr.2: 99360171-99360171 on Build GRCh38
rs1001876014 Chr.2: 99360201-99360201 on Build GRCh38

View Full Product Details