Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99368968-99369498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5B
Entrez Gene ID
9669
Gene Name
eukaryotic translation initiation factor 5B
Gene Aliases
IF2
UniGene
Hs.158688
Gene Chromosome Location
Chr.2: 99337371-99400265 on Build GRCh38
Transcripts
NM_015904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466917 c.1460C>T p.P487L Chr.2: 99369464-99369464 on Build GRCh38
7308824 c.1388-8T>G p.? Chr.2: 99369384-99369384 on Build GRCh38
6303255 c.1427T>A p.M476K Chr.2: 99369431-99369431 on Build GRCh38
1024354 c.1422A>C p.E474D Chr.2: 99369426-99369426 on Build GRCh38
2914600 c.1403A>G p.E468G Chr.2: 99369407-99369407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530214657 Chr.2: 99369044-99369044 on Build GRCh38
rs746966926 Chr.2: 99369374-99369374 on Build GRCh38
rs1011074392 Chr.2: 99369010-99369010 on Build GRCh38
rs199806211 Chr.2: 99369412-99369412 on Build GRCh38
rs1020994541 Chr.2: 99369370-99369370 on Build GRCh38
rs981230053 Chr.2: 99369233-99369233 on Build GRCh38
rs1040107287 Chr.2: 99369109-99369109 on Build GRCh38
rs1041175303 Chr.2: 99369465-99369465 on Build GRCh38
rs533629182 Chr.2: 99369308-99369308 on Build GRCh38
rs931326949 Chr.2: 99369362-99369362 on Build GRCh38
rs748958387 Chr.2: 99369421-99369421 on Build GRCh38
rs768658826 Chr.2: 99369389-99369389 on Build GRCh38
rs896432838 Chr.2: 99369329-99369329 on Build GRCh38
rs967199612 Chr.2: 99369021-99369021 on Build GRCh38
rs567704112 Chr.2: 99369028-99369028 on Build GRCh38
rs779636560 Chr.2: 99369413-99369413 on Build GRCh38
rs757290796 Chr.2: 99369404-99369404 on Build GRCh38
rs775465894 Chr.2: 99369363-99369365 on Build GRCh38
rs772485802 Chr.2: 99369352-99369352 on Build GRCh38
rs954406161 Chr.2: 99369017-99369017 on Build GRCh38
rs957049274 Chr.2: 99369157-99369157 on Build GRCh38
rs574125507 Chr.2: 99369282-99369282 on Build GRCh38
rs1028492431 Chr.2: 99369002-99369002 on Build GRCh38
rs991575689 Chr.2: 99369267-99369267 on Build GRCh38
rs967210860 Chr.2: 99369227-99369227 on Build GRCh38
rs752950261 Chr.2: 99369380-99369380 on Build GRCh38
rs375834698 Chr.2: 99369260-99369260 on Build GRCh38
rs200368757 Chr.2: 99369145-99369145 on Build GRCh38
rs1011525319 Chr.2: 99369204-99369204 on Build GRCh38
rs555244105 Chr.2: 99369207-99369207 on Build GRCh38
rs531511952 Chr.2: 99368994-99368994 on Build GRCh38
rs550911759 Chr.2: 99369027-99369027 on Build GRCh38
rs534020163 Chr.2: 99369128-99369128 on Build GRCh38
rs759997724 Chr.2: 99369451-99369451 on Build GRCh38
rs913101352 Chr.2: 99369011-99369011 on Build GRCh38
rs761302026 Chr.2: 99369357-99369357 on Build GRCh38
rs780962619 Chr.2: 99369407-99369407 on Build GRCh38
rs188348934 Chr.2: 99369250-99369250 on Build GRCh38
rs766791794 Chr.2: 99369365-99369365 on Build GRCh38
rs771515690 Chr.2: 99369446-99369446 on Build GRCh38
rs1023265466 Chr.2: 99369074-99369074 on Build GRCh38
rs937358879 Chr.2: 99369040-99369040 on Build GRCh38
rs765254055 Chr.2: 99369372-99369372 on Build GRCh38
rs747302426 Chr.2: 99369444-99369444 on Build GRCh38
rs931584425 Chr.2: 99369059-99369059 on Build GRCh38
rs770981725 Chr.2: 99369354-99369355 on Build GRCh38
rs762651773 Chr.2: 99369351-99369351 on Build GRCh38
rs749373529 Chr.2: 99369355-99369355 on Build GRCh38
rs745423759 Chr.2: 99369411-99369411 on Build GRCh38
rs764220537 Chr.2: 99369474-99369474 on Build GRCh38
rs765305365 Chr.2: 99369460-99369460 on Build GRCh38
rs565763308 Chr.2: 99369048-99369048 on Build GRCh38
rs905040742 Chr.2: 99369239-99369239 on Build GRCh38
rs763963671 Chr.2: 99369397-99369397 on Build GRCh38
rs759588753 Chr.2: 99369369-99369369 on Build GRCh38
rs901608695 Chr.2: 99369470-99369470 on Build GRCh38
rs1056176311 Chr.2: 99369043-99369043 on Build GRCh38
rs552517019 Chr.2: 99369198-99369198 on Build GRCh38
rs776730231 Chr.2: 99369367-99369367 on Build GRCh38
rs969471229 Chr.2: 99369097-99369097 on Build GRCh38
rs567054397 Chr.2: 99369071-99369071 on Build GRCh38
rs763297124 Chr.2: 99369472-99369472 on Build GRCh38
rs1008344157 Chr.2: 99369103-99369103 on Build GRCh38
rs373807627 Chr.2: 99369424-99369424 on Build GRCh38
rs775301435 Chr.2: 99369342-99369342 on Build GRCh38
rs553918850 Chr.2: 99369026-99369026 on Build GRCh38
rs777145889 Chr.2: 99369447-99369447 on Build GRCh38
rs758549416 Chr.2: 99369383-99369383 on Build GRCh38
rs995876702 Chr.2: 99369129-99369129 on Build GRCh38
rs930414873 Chr.2: 99369190-99369190 on Build GRCh38
rs954229780 Chr.2: 99369422-99369422 on Build GRCh38
rs1029050444 Chr.2: 99369137-99369137 on Build GRCh38
rs918992325 Chr.2: 99369089-99369089 on Build GRCh38
rs147438767 Chr.2: 99369403-99369403 on Build GRCh38
rs773388179 Chr.2: 99369353-99369353 on Build GRCh38
rs775491897 Chr.2: 99369464-99369464 on Build GRCh38

View Full Product Details