Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99547507-99548049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF3
Entrez Gene ID
3899
Gene Name
AF4/FMR2 family member 3
Gene Aliases
LAF4, MLLT2-like
UniGene
Hs.444414
Gene Chromosome Location
Chr.2: 99545861-100142583 on Build GRCh38
Transcripts
NM_001025108.1 NM_002285.2 XM_005263943.3 XM_005263945.3 XM_011511169.2 XM_011511170.2 XM_011511171.2 XM_011511172.2 XM_011511173.2 XM_011511174.2 XM_011511175.2 XM_011511176.2 XM_011511177.2 XM_011511178.2 XM_011511179.2 XM_017004085.1 XM_017004086.1 XM_017004087.1

SNP Detail

SNP ID Chromosome Location
rs780012081 Chr.2: 99547616-99547616 on Build GRCh38
rs1036167533 Chr.2: 99547572-99547572 on Build GRCh38
rs931827366 Chr.2: 99547903-99547903 on Build GRCh38
rs536695863 Chr.2: 99547937-99547937 on Build GRCh38
rs563815800 Chr.2: 99547638-99547638 on Build GRCh38
rs944128477 Chr.2: 99547860-99547860 on Build GRCh38
rs755189129 Chr.2: 99547857-99547857 on Build GRCh38
rs145341496 Chr.2: 99547631-99547631 on Build GRCh38
rs981539152 Chr.2: 99547657-99547657 on Build GRCh38
rs996086978 Chr.2: 99547554-99547554 on Build GRCh38
rs186929250 Chr.2: 99547640-99547640 on Build GRCh38
rs1041784032 Chr.2: 99547955-99547955 on Build GRCh38
rs901675353 Chr.2: 99547902-99547902 on Build GRCh38
rs983208689 Chr.2: 99547827-99547827 on Build GRCh38
rs1035425440 Chr.2: 99547996-99547996 on Build GRCh38
rs958902137 Chr.2: 99547720-99547720 on Build GRCh38
rs931303938 Chr.2: 99547800-99547800 on Build GRCh38
rs1035130793 Chr.2: 99547675-99547675 on Build GRCh38
rs113658725 Chr.2: 99547921-99547921 on Build GRCh38
rs1035461303 Chr.2: 99547984-99547984 on Build GRCh38
rs979687759 Chr.2: 99547755-99547755 on Build GRCh38
rs988231133 Chr.2: 99547715-99547715 on Build GRCh38
rs968185499 Chr.2: 99547735-99547735 on Build GRCh38
rs566237388 Chr.2: 99547823-99547823 on Build GRCh38
rs1036093536 Chr.2: 99547933-99547933 on Build GRCh38
rs893060302 Chr.2: 99547929-99547929 on Build GRCh38
rs991682203 Chr.2: 99547721-99547721 on Build GRCh38
rs1002224757 Chr.2: 99547959-99547959 on Build GRCh38
rs994776144 Chr.2: 99547573-99547573 on Build GRCh38
rs181091855 Chr.2: 99547906-99547906 on Build GRCh38
rs944573118 Chr.2: 99547544-99547545 on Build GRCh38
rs191359200 Chr.2: 99547816-99547816 on Build GRCh38
rs541143472 Chr.2: 99547958-99547958 on Build GRCh38
rs1035810540 Chr.2: 99547688-99547688 on Build GRCh38
rs1002575103 Chr.2: 99547965-99547965 on Build GRCh38
rs921481620 Chr.2: 99547787-99547787 on Build GRCh38
rs1028955588 Chr.2: 99547562-99547562 on Build GRCh38
rs528798649 Chr.2: 99547873-99547873 on Build GRCh38
rs892789340 Chr.2: 99547580-99547580 on Build GRCh38
rs1002577312 Chr.2: 99547674-99547674 on Build GRCh38
rs778223050 Chr.2: 99547712-99547712 on Build GRCh38
rs920208944 Chr.2: 99547789-99547789 on Build GRCh38
rs1011670940 Chr.2: 99547581-99547581 on Build GRCh38
rs748493585 Chr.2: 99547963-99547963 on Build GRCh38

View Full Product Details