Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101349298-101349843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREG2
Entrez Gene ID
200407
Gene Name
cellular repressor of E1A stimulated genes 2
Gene Aliases
-
UniGene
Hs.732289
Gene Chromosome Location
Chr.2: 101348354-101387970 on Build GRCh38
Transcripts
NM_153836.3

SNP Detail

SNP ID Chromosome Location
rs951198194 Chr.2: 101349626-101349626 on Build GRCh38
rs924098353 Chr.2: 101349818-101349818 on Build GRCh38
rs945001315 Chr.2: 101349618-101349618 on Build GRCh38
rs866035785 Chr.2: 101349400-101349400 on Build GRCh38
rs983503416 Chr.2: 101349658-101349658 on Build GRCh38
rs894617334 Chr.2: 101349779-101349779 on Build GRCh38
rs1047126456 Chr.2: 101349808-101349808 on Build GRCh38
rs938867322 Chr.2: 101349683-101349683 on Build GRCh38
rs181649950 Chr.2: 101349420-101349420 on Build GRCh38
rs200453959 Chr.2: 101349753-101349754 on Build GRCh38
rs4638816 Chr.2: 101349399-101349399 on Build GRCh38
rs1042125377 Chr.2: 101349629-101349629 on Build GRCh38
rs999230261 Chr.2: 101349504-101349504 on Build GRCh38
rs1009356469 Chr.2: 101349369-101349369 on Build GRCh38
rs1032059122 Chr.2: 101349522-101349522 on Build GRCh38
rs759539237 Chr.2: 101349674-101349674 on Build GRCh38
rs1042111046 Chr.2: 101349382-101349382 on Build GRCh38
rs796991728 Chr.2: 101349478-101349478 on Build GRCh38
rs989799395 Chr.2: 101349609-101349609 on Build GRCh38
rs77234017 Chr.2: 101349755-101349755 on Build GRCh38
rs903367704 Chr.2: 101349387-101349387 on Build GRCh38
rs932606831 Chr.2: 101349577-101349577 on Build GRCh38
rs115133028 Chr.2: 101349679-101349679 on Build GRCh38
rs1006529871 Chr.2: 101349561-101349561 on Build GRCh38
rs977105976 Chr.2: 101349607-101349608 on Build GRCh38
rs538384189 Chr.2: 101349800-101349800 on Build GRCh38
rs976551832 Chr.2: 101349566-101349566 on Build GRCh38
rs749504249 Chr.2: 101349741-101349741 on Build GRCh38
rs1013208374 Chr.2: 101349814-101349814 on Build GRCh38
rs146532595 Chr.2: 101349596-101349596 on Build GRCh38
rs149200684 Chr.2: 101349738-101349738 on Build GRCh38
rs949692112 Chr.2: 101349443-101349443 on Build GRCh38
rs949654043 Chr.2: 101349791-101349793 on Build GRCh38
rs372863822 Chr.2: 101349528-101349528 on Build GRCh38
rs186845924 Chr.2: 101349615-101349615 on Build GRCh38
rs6543031 Chr.2: 101349402-101349402 on Build GRCh38
rs965225402 Chr.2: 101349553-101349553 on Build GRCh38
rs1053484011 Chr.2: 101349331-101349331 on Build GRCh38
rs79593073 Chr.2: 101349753-101349753 on Build GRCh38
rs1057261129 Chr.2: 101349701-101349701 on Build GRCh38
rs73943455 Chr.2: 101349401-101349401 on Build GRCh38
rs757910923 Chr.2: 101349398-101349400 on Build GRCh38
rs976592168 Chr.2: 101349330-101349330 on Build GRCh38
rs902716110 Chr.2: 101349484-101349484 on Build GRCh38
rs11677690 Chr.2: 101349576-101349576 on Build GRCh38
rs753012739 Chr.2: 101349336-101349336 on Build GRCh38
rs550610535 Chr.2: 101349533-101349533 on Build GRCh38
rs867958754 Chr.2: 101349740-101349740 on Build GRCh38

View Full Product Details