Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 124221363-124221870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
UniGene
Hs.660653
Gene Chromosome Location
Chr.2: 124025287-124921201 on Build GRCh38
Transcripts
NM_130773.3 XM_006712258.1 XM_017003316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5955469 c.83-2A>G p.? Chr.2: 124221703-124221703 on Build GRCh38
1318189 c.143A>T p.D48V Chr.2: 124221765-124221765 on Build GRCh38
5672725 c.187+1G>T p.? Chr.2: 124221810-124221810 on Build GRCh38
7199984 c.128T>C p.F43S Chr.2: 124221750-124221750 on Build GRCh38
5860040 c.104C>T p.A35V Chr.2: 124221726-124221726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193289884 Chr.2: 124221452-124221452 on Build GRCh38
rs781042103 Chr.2: 124221753-124221753 on Build GRCh38
rs771461179 Chr.2: 124221601-124221601 on Build GRCh38
rs1003198124 Chr.2: 124221574-124221574 on Build GRCh38
rs751646975 Chr.2: 124221678-124221678 on Build GRCh38
rs573553026 Chr.2: 124221743-124221743 on Build GRCh38
rs955644402 Chr.2: 124221536-124221536 on Build GRCh38
rs753178503 Chr.2: 124221805-124221805 on Build GRCh38
rs796820951 Chr.2: 124221533-124221533 on Build GRCh38
rs183853031 Chr.2: 124221415-124221415 on Build GRCh38
rs763961372 Chr.2: 124221681-124221681 on Build GRCh38
rs115075882 Chr.2: 124221492-124221492 on Build GRCh38
rs1008924971 Chr.2: 124221552-124221552 on Build GRCh38
rs893982045 Chr.2: 124221510-124221510 on Build GRCh38
rs372174479 Chr.2: 124221768-124221768 on Build GRCh38
rs961337192 Chr.2: 124221583-124221583 on Build GRCh38
rs764460083 Chr.2: 124221828-124221828 on Build GRCh38
rs749730305 Chr.2: 124221572-124221572 on Build GRCh38
rs902152931 Chr.2: 124221573-124221573 on Build GRCh38
rs990620991 Chr.2: 124221619-124221619 on Build GRCh38
rs577268745 Chr.2: 124221587-124221587 on Build GRCh38
rs866407624 Chr.2: 124221747-124221747 on Build GRCh38
rs371812691 Chr.2: 124221487-124221487 on Build GRCh38
rs949311242 Chr.2: 124221714-124221714 on Build GRCh38
rs543391642 Chr.2: 124221420-124221420 on Build GRCh38
rs758397323 Chr.2: 124221688-124221690 on Build GRCh38
rs185431634 Chr.2: 124221687-124221687 on Build GRCh38
rs553346590 Chr.2: 124221440-124221440 on Build GRCh38
rs768609837 Chr.2: 124221746-124221746 on Build GRCh38
rs778499567 Chr.2: 124221717-124221717 on Build GRCh38
rs536642205 Chr.2: 124221671-124221671 on Build GRCh38
rs900262369 Chr.2: 124221491-124221491 on Build GRCh38
rs756489300 Chr.2: 124221706-124221706 on Build GRCh38
rs114129158 Chr.2: 124221441-124221441 on Build GRCh38
rs753080754 Chr.2: 124221695-124221695 on Build GRCh38
rs759581983 Chr.2: 124221793-124221793 on Build GRCh38
rs890516072 Chr.2: 124221481-124221481 on Build GRCh38
rs752654564 Chr.2: 124221784-124221784 on Build GRCh38
rs76918335 Chr.2: 124221582-124221582 on Build GRCh38
rs750084392 Chr.2: 124221731-124221731 on Build GRCh38
rs374252837 Chr.2: 124221785-124221785 on Build GRCh38
rs188297422 Chr.2: 124221416-124221416 on Build GRCh38
rs766580095 Chr.2: 124221670-124221670 on Build GRCh38
rs375152177 Chr.2: 124221736-124221736 on Build GRCh38
rs367926323 Chr.2: 124221739-124221739 on Build GRCh38
rs370359251 Chr.2: 124221689-124221689 on Build GRCh38
rs542472391 Chr.2: 124221776-124221776 on Build GRCh38
rs974058123 Chr.2: 124221602-124221602 on Build GRCh38
rs781243324 Chr.2: 124221697-124221697 on Build GRCh38
rs553167461 Chr.2: 124221825-124221825 on Build GRCh38
rs139603853 Chr.2: 124221835-124221835 on Build GRCh38
rs941276635 Chr.2: 124221396-124221396 on Build GRCh38
rs573397470 Chr.2: 124221567-124221567 on Build GRCh38
rs565307534 Chr.2: 124221393-124221393 on Build GRCh38
rs557548096 Chr.2: 124221538-124221538 on Build GRCh38
rs959381939 Chr.2: 124221611-124221611 on Build GRCh38
rs919971856 Chr.2: 124221620-124221620 on Build GRCh38
rs879152497 Chr.2: 124221500-124221500 on Build GRCh38
rs774454344 Chr.2: 124221787-124221787 on Build GRCh38
rs747934668 Chr.2: 124221771-124221771 on Build GRCh38
rs768017057 Chr.2: 124221797-124221797 on Build GRCh38

View Full Product Details