Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130755371-130755860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER3
Entrez Gene ID
205147
Gene Name
APC membrane recruitment protein 3
Gene Aliases
FAM123C
UniGene
Hs.369289
Gene Chromosome Location
Chr.2: 130754475-130768134 on Build GRCh38
Transcripts
NM_001105193.1 NM_152698.2 XM_011510793.2

SNP Detail

SNP ID Chromosome Location
rs535576846 Chr.2: 130755472-130755472 on Build GRCh38
rs967787781 Chr.2: 130755469-130755469 on Build GRCh38
rs947210880 Chr.2: 130755414-130755414 on Build GRCh38
rs765652327 Chr.2: 130755708-130755708 on Build GRCh38
rs932716974 Chr.2: 130755576-130755576 on Build GRCh38
rs1015835520 Chr.2: 130755727-130755727 on Build GRCh38
rs944071178 Chr.2: 130755830-130755830 on Build GRCh38
rs978864264 Chr.2: 130755820-130755820 on Build GRCh38
rs114232877 Chr.2: 130755770-130755770 on Build GRCh38
rs912473713 Chr.2: 130755751-130755764 on Build GRCh38
rs924448490 Chr.2: 130755625-130755625 on Build GRCh38
rs986550543 Chr.2: 130755776-130755776 on Build GRCh38
rs934602494 Chr.2: 130755628-130755628 on Build GRCh38
rs565470837 Chr.2: 130755575-130755575 on Build GRCh38
rs960615630 Chr.2: 130755722-130755722 on Build GRCh38
rs940225166 Chr.2: 130755559-130755559 on Build GRCh38
rs539413757 Chr.2: 130755537-130755537 on Build GRCh38
rs564384223 Chr.2: 130755825-130755825 on Build GRCh38
rs777198448 Chr.2: 130755600-130755600 on Build GRCh38
rs992778927 Chr.2: 130755515-130755515 on Build GRCh38
rs1033353140 Chr.2: 130755720-130755720 on Build GRCh38
rs1005283663 Chr.2: 130755712-130755712 on Build GRCh38
rs895698546 Chr.2: 130755694-130755694 on Build GRCh38
rs974206705 Chr.2: 130755563-130755563 on Build GRCh38
rs747503162 Chr.2: 130755391-130755391 on Build GRCh38
rs974244027 Chr.2: 130755769-130755769 on Build GRCh38
rs568582654 Chr.2: 130755440-130755440 on Build GRCh38
rs947465070 Chr.2: 130755612-130755612 on Build GRCh38
rs566626938 Chr.2: 130755667-130755667 on Build GRCh38
rs191788340 Chr.2: 130755828-130755828 on Build GRCh38
rs889069720 Chr.2: 130755407-130755407 on Build GRCh38
rs541542568 Chr.2: 130755688-130755688 on Build GRCh38
rs555678545 Chr.2: 130755587-130755587 on Build GRCh38
rs1006816535 Chr.2: 130755517-130755517 on Build GRCh38
rs1002336879 Chr.2: 130755718-130755718 on Build GRCh38
rs944645069 Chr.2: 130755605-130755605 on Build GRCh38
rs924472022 Chr.2: 130755821-130755821 on Build GRCh38
rs903618021 Chr.2: 130755699-130755699 on Build GRCh38
rs1009329233 Chr.2: 130755436-130755436 on Build GRCh38
rs1009203097 Chr.2: 130755413-130755413 on Build GRCh38
rs528301021 Chr.2: 130755396-130755396 on Build GRCh38
rs954174645 Chr.2: 130755778-130755778 on Build GRCh38
rs961005352 Chr.2: 130755514-130755514 on Build GRCh38
rs142557551 Chr.2: 130755476-130755476 on Build GRCh38
rs1023140663 Chr.2: 130755488-130755488 on Build GRCh38
rs1040661761 Chr.2: 130755620-130755620 on Build GRCh38
rs187426959 Chr.2: 130755551-130755551 on Build GRCh38
rs903111766 Chr.2: 130755432-130755432 on Build GRCh38
rs1017591255 Chr.2: 130755790-130755790 on Build GRCh38
rs1018799363 Chr.2: 130755421-130755421 on Build GRCh38
rs546822015 Chr.2: 130755839-130755839 on Build GRCh38
rs1022088238 Chr.2: 130755454-130755454 on Build GRCh38
rs575331677 Chr.2: 130755729-130755729 on Build GRCh38
rs1013242616 Chr.2: 130755726-130755726 on Build GRCh38
rs546440264 Chr.2: 130755424-130755424 on Build GRCh38
rs115394632 Chr.2: 130755698-130755698 on Build GRCh38
rs985911250 Chr.2: 130755813-130755813 on Build GRCh38
rs954716809 Chr.2: 130755584-130755584 on Build GRCh38
rs573160101 Chr.2: 130755555-130755555 on Build GRCh38

View Full Product Details