Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134870929-134871458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACMSD
Entrez Gene ID
130013
Gene Name
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases
-
UniGene
Hs.655728
Gene Chromosome Location
Chr.2: 134838319-134902906 on Build GRCh38
Transcripts
NM_001307983.1 NM_138326.2 XM_005263586.4 XM_005263588.4 XM_005263589.4 XM_005263590.4 XM_011510592.2 XM_017003325.1 XM_017003326.1 NR_036549.1

Gene Symbol
CCNT2-AS1
Entrez Gene ID
100129961
Gene Name
CCNT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.729028
Gene Chromosome Location
Chr.2: 134866633-134918606 on Build GRCh38
Transcripts
NM_001307983.1 NM_138326.2 XM_005263586.4 XM_005263588.4 XM_005263589.4 XM_005263590.4 XM_011510592.2 XM_017003325.1 XM_017003326.1 NR_036549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3115930 c.666C>T p.F222F Chr.2: 134871050-134871050 on Build GRCh38
5914846 c.674G>A p.G225D Chr.2: 134871058-134871058 on Build GRCh38
4402195 c.581G>A p.G194E Chr.2: 134870965-134870965 on Build GRCh38
5678982 c.672T>C p.H224H Chr.2: 134871056-134871056 on Build GRCh38
6567013 c.676+7C>A p.? Chr.2: 134871067-134871067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757198931 Chr.2: 134871032-134871032 on Build GRCh38
rs965696652 Chr.2: 134871327-134871327 on Build GRCh38
rs1026737503 Chr.2: 134871273-134871273 on Build GRCh38
rs747265677 Chr.2: 134870980-134870980 on Build GRCh38
rs755257201 Chr.2: 134871058-134871058 on Build GRCh38
rs1043133237 Chr.2: 134871128-134871128 on Build GRCh38
rs924277020 Chr.2: 134871044-134871044 on Build GRCh38
rs116864404 Chr.2: 134871110-134871110 on Build GRCh38
rs540578998 Chr.2: 134870984-134870984 on Build GRCh38
rs566210096 Chr.2: 134871358-134871358 on Build GRCh38
rs746694024 Chr.2: 134871052-134871052 on Build GRCh38
rs142405012 Chr.2: 134871434-134871434 on Build GRCh38
rs914404207 Chr.2: 134871116-134871116 on Build GRCh38
rs778883483 Chr.2: 134871041-134871041 on Build GRCh38
rs895091088 Chr.2: 134871199-134871199 on Build GRCh38
rs561527847 Chr.2: 134871073-134871073 on Build GRCh38
rs977403610 Chr.2: 134871331-134871331 on Build GRCh38
rs762413529 Chr.2: 134870988-134870988 on Build GRCh38
rs961023973 Chr.2: 134871243-134871243 on Build GRCh38
rs749324987 Chr.2: 134871074-134871074 on Build GRCh38
rs1374290 Chr.2: 134871202-134871202 on Build GRCh38
rs1013900677 Chr.2: 134871222-134871222 on Build GRCh38
rs777035035 Chr.2: 134871360-134871360 on Build GRCh38
rs748508384 Chr.2: 134871070-134871070 on Build GRCh38
rs913454856 Chr.2: 134871425-134871425 on Build GRCh38
rs548133042 Chr.2: 134871426-134871426 on Build GRCh38
rs267598886 Chr.2: 134870965-134870965 on Build GRCh38
rs889940007 Chr.2: 134871338-134871338 on Build GRCh38
rs532570219 Chr.2: 134871281-134871281 on Build GRCh38
rs961139585 Chr.2: 134870993-134870993 on Build GRCh38
rs904579098 Chr.2: 134871170-134871170 on Build GRCh38
rs1048650470 Chr.2: 134871336-134871336 on Build GRCh38
rs34647266 Chr.2: 134871026-134871026 on Build GRCh38
rs202239310 Chr.2: 134871081-134871081 on Build GRCh38
rs1017337774 Chr.2: 134871014-134871014 on Build GRCh38
rs138679619 Chr.2: 134871051-134871051 on Build GRCh38
rs773402188 Chr.2: 134870996-134870996 on Build GRCh38
rs1037000842 Chr.2: 134871262-134871264 on Build GRCh38
rs952415426 Chr.2: 134871294-134871294 on Build GRCh38
rs549455494 Chr.2: 134871111-134871111 on Build GRCh38
rs1000725972 Chr.2: 134871177-134871177 on Build GRCh38
rs766246394 Chr.2: 134871375-134871375 on Build GRCh38
rs370052319 Chr.2: 134871060-134871060 on Build GRCh38
rs913233458 Chr.2: 134871119-134871119 on Build GRCh38
rs530193645 Chr.2: 134871411-134871411 on Build GRCh38
rs1024683671 Chr.2: 134871238-134871238 on Build GRCh38
rs542848101 Chr.2: 134871050-134871050 on Build GRCh38
rs138684700 Chr.2: 134870978-134870978 on Build GRCh38
rs763984016 Chr.2: 134871013-134871013 on Build GRCh38
rs34715486 Chr.2: 134871388-134871388 on Build GRCh38
rs565392166 Chr.2: 134871263-134871263 on Build GRCh38
rs770461483 Chr.2: 134870995-134870995 on Build GRCh38
rs559044226 Chr.2: 134870986-134870986 on Build GRCh38
rs766643624 Chr.2: 134871002-134871002 on Build GRCh38
rs774397899 Chr.2: 134871101-134871101 on Build GRCh38
rs1055337231 Chr.2: 134871194-134871194 on Build GRCh38
rs985676855 Chr.2: 134871305-134871305 on Build GRCh38
rs147672307 Chr.2: 134871124-134871124 on Build GRCh38
rs1006985943 Chr.2: 134871403-134871403 on Build GRCh38
rs760338573 Chr.2: 134871007-134871007 on Build GRCh38
rs770410185 Chr.2: 134871072-134871072 on Build GRCh38
rs1044485840 Chr.2: 134871217-134871217 on Build GRCh38
rs201537963 Chr.2: 134871015-134871015 on Build GRCh38
rs183097083 Chr.2: 134871312-134871312 on Build GRCh38
rs771158185 Chr.2: 134871094-134871094 on Build GRCh38
rs993799623 Chr.2: 134871248-134871248 on Build GRCh38
rs991850875 Chr.2: 134870970-134870970 on Build GRCh38

View Full Product Details