Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135907205-135907753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DARS
Entrez Gene ID
1615
Gene Name
aspartyl-tRNA synthetase
Gene Aliases
HBSL, aspRS
UniGene
Hs.503787
Gene Chromosome Location
Chr.2: 135905881-135986046 on Build GRCh38
Transcripts
NM_001293312.1 NM_001349.3 XM_017003489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4601008 c.1454A>T p.N485I Chr.2: 135907368-135907368 on Build GRCh38
1193515 c.1451A>G p.H484R Chr.2: 135907371-135907371 on Build GRCh38
5894252 c.1499C>T p.T500I Chr.2: 135907323-135907323 on Build GRCh38
6640103 c.1459C>T p.R487C Chr.2: 135907363-135907363 on Build GRCh38
5691998 c.1437G>T p.L479L Chr.2: 135907385-135907385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762273554 Chr.2: 135907314-135907314 on Build GRCh38
rs901313756 Chr.2: 135907246-135907246 on Build GRCh38
rs574202016 Chr.2: 135907685-135907685 on Build GRCh38
rs537444284 Chr.2: 135907603-135907603 on Build GRCh38
rs991391530 Chr.2: 135907469-135907469 on Build GRCh38
rs778444199 Chr.2: 135907275-135907275 on Build GRCh38
rs748680951 Chr.2: 135907437-135907437 on Build GRCh38
rs892393256 Chr.2: 135907506-135907506 on Build GRCh38
rs1040369870 Chr.2: 135907238-135907238 on Build GRCh38
rs755922416 Chr.2: 135907665-135907665 on Build GRCh38
rs567792053 Chr.2: 135907391-135907391 on Build GRCh38
rs745507665 Chr.2: 135907280-135907280 on Build GRCh38
rs77052188 Chr.2: 135907258-135907258 on Build GRCh38
rs16832162 Chr.2: 135907592-135907592 on Build GRCh38
rs974819729 Chr.2: 135907717-135907717 on Build GRCh38
rs533666967 Chr.2: 135907659-135907659 on Build GRCh38
rs985443346 Chr.2: 135907675-135907675 on Build GRCh38
rs570992131 Chr.2: 135907632-135907632 on Build GRCh38
rs78275586 Chr.2: 135907262-135907262 on Build GRCh38
rs909850382 Chr.2: 135907591-135907591 on Build GRCh38
rs367624698 Chr.2: 135907301-135907301 on Build GRCh38
rs960349920 Chr.2: 135907240-135907240 on Build GRCh38
rs547216088 Chr.2: 135907686-135907687 on Build GRCh38
rs971313249 Chr.2: 135907490-135907490 on Build GRCh38
rs77743403 Chr.2: 135907260-135907260 on Build GRCh38
rs770444681 Chr.2: 135907274-135907274 on Build GRCh38
rs992435404 Chr.2: 135907243-135907251 on Build GRCh38
rs76296777 Chr.2: 135907405-135907405 on Build GRCh38
rs918884505 Chr.2: 135907654-135907654 on Build GRCh38
rs777720339 Chr.2: 135907270-135907270 on Build GRCh38
rs1020492946 Chr.2: 135907336-135907336 on Build GRCh38
rs926311986 Chr.2: 135907239-135907245 on Build GRCh38
rs775136184 Chr.2: 135907285-135907285 on Build GRCh38
rs1009863395 Chr.2: 135907511-135907511 on Build GRCh38
rs528754955 Chr.2: 135907532-135907533 on Build GRCh38
rs55815289 Chr.2: 135907701-135907702 on Build GRCh38
rs960420992 Chr.2: 135907602-135907602 on Build GRCh38
rs771645282 Chr.2: 135907282-135907282 on Build GRCh38
rs370805809 Chr.2: 135907390-135907390 on Build GRCh38
rs901411785 Chr.2: 135907671-135907671 on Build GRCh38
rs765342897 Chr.2: 135907322-135907322 on Build GRCh38
rs967111310 Chr.2: 135907341-135907341 on Build GRCh38
rs757199434 Chr.2: 135907377-135907377 on Build GRCh38
rs182814004 Chr.2: 135907605-135907605 on Build GRCh38
rs779639974 Chr.2: 135907402-135907402 on Build GRCh38
rs930076270 Chr.2: 135907568-135907568 on Build GRCh38
rs753793246 Chr.2: 135907362-135907362 on Build GRCh38
rs147077598 Chr.2: 135907342-135907342 on Build GRCh38
rs776915292 Chr.2: 135907434-135907434 on Build GRCh38
rs766655730 Chr.2: 135907340-135907340 on Build GRCh38
rs768931563 Chr.2: 135907293-135907293 on Build GRCh38
rs553902358 Chr.2: 135907679-135907679 on Build GRCh38
rs998335495 Chr.2: 135907257-135907257 on Build GRCh38
rs755901436 Chr.2: 135907351-135907351 on Build GRCh38
rs757925492 Chr.2: 135907399-135907399 on Build GRCh38
rs747323714 Chr.2: 135907288-135907288 on Build GRCh38
rs1039085577 Chr.2: 135907441-135907441 on Build GRCh38
rs773488177 Chr.2: 135907329-135907329 on Build GRCh38
rs769039312 Chr.2: 135907432-135907432 on Build GRCh38
rs763133905 Chr.2: 135907330-135907330 on Build GRCh38
rs376886390 Chr.2: 135907676-135907676 on Build GRCh38
rs998471403 Chr.2: 135907673-135907673 on Build GRCh38
rs919127866 Chr.2: 135907565-135907565 on Build GRCh38
rs968281690 Chr.2: 135907563-135907563 on Build GRCh38
rs763837517 Chr.2: 135907355-135907355 on Build GRCh38
rs373863494 Chr.2: 135907450-135907450 on Build GRCh38
rs919529324 Chr.2: 135907248-135907248 on Build GRCh38
rs748828277 Chr.2: 135907661-135907661 on Build GRCh38
rs1007103895 Chr.2: 135907232-135907232 on Build GRCh38
rs574058566 Chr.2: 135907242-135907242 on Build GRCh38
rs367625607 Chr.2: 135907242-135907242 on Build GRCh38
rs917181294 Chr.2: 135907489-135907489 on Build GRCh38
rs550214818 Chr.2: 135907572-135907572 on Build GRCh38
rs767750753 Chr.2: 135907470-135907470 on Build GRCh38
rs587776984 Chr.2: 135907363-135907363 on Build GRCh38
rs773874094 Chr.2: 135907453-135907453 on Build GRCh38
rs756024844 Chr.2: 135907496-135907496 on Build GRCh38
rs991838929 Chr.2: 135907650-135907650 on Build GRCh38
rs960213085 Chr.2: 135907709-135907709 on Build GRCh38

View Full Product Details