Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151801925-151802445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5A
Entrez Gene ID
26225
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
ARFLP5, ARL5
UniGene
Hs.470233
Gene Chromosome Location
Chr.2: 151800966-151828495 on Build GRCh38
Transcripts
NM_001037174.1 NM_012097.3 NM_177985.2

SNP Detail

SNP ID Chromosome Location
rs749835322 Chr.2: 151802296-151802300 on Build GRCh38
rs189627810 Chr.2: 151802228-151802228 on Build GRCh38
rs993910074 Chr.2: 151802060-151802060 on Build GRCh38
rs746189506 Chr.2: 151801953-151801953 on Build GRCh38
rs372896293 Chr.2: 151802367-151802367 on Build GRCh38
rs901895997 Chr.2: 151802048-151802048 on Build GRCh38
rs186394807 Chr.2: 151802184-151802184 on Build GRCh38
rs916998143 Chr.2: 151802156-151802156 on Build GRCh38
rs530367243 Chr.2: 151802025-151802025 on Build GRCh38
rs1005457294 Chr.2: 151802073-151802073 on Build GRCh38
rs776153113 Chr.2: 151802406-151802406 on Build GRCh38
rs369149829 Chr.2: 151802285-151802285 on Build GRCh38
rs769386915 Chr.2: 151802012-151802012 on Build GRCh38
rs553275572 Chr.2: 151802302-151802302 on Build GRCh38
rs1040475494 Chr.2: 151802041-151802041 on Build GRCh38
rs936237401 Chr.2: 151801955-151801955 on Build GRCh38
rs200949920 Chr.2: 151802108-151802109 on Build GRCh38
rs991247080 Chr.2: 151802136-151802136 on Build GRCh38
rs1017217245 Chr.2: 151802076-151802076 on Build GRCh38
rs540485185 Chr.2: 151802037-151802037 on Build GRCh38
rs878934296 Chr.2: 151802269-151802269 on Build GRCh38
rs772737104 Chr.2: 151802200-151802200 on Build GRCh38
rs993569550 Chr.2: 151802386-151802386 on Build GRCh38
rs375081080 Chr.2: 151802368-151802368 on Build GRCh38
rs13428623 Chr.2: 151802063-151802063 on Build GRCh38
rs140772338 Chr.2: 151802187-151802187 on Build GRCh38
rs186969549 Chr.2: 151802304-151802304 on Build GRCh38
rs543819015 Chr.2: 151802013-151802013 on Build GRCh38
rs770558871 Chr.2: 151802298-151802298 on Build GRCh38
rs531826472 Chr.2: 151802226-151802226 on Build GRCh38
rs951391174 Chr.2: 151802071-151802071 on Build GRCh38
rs1040527203 Chr.2: 151802314-151802314 on Build GRCh38
rs889270153 Chr.2: 151802034-151802034 on Build GRCh38
rs901948508 Chr.2: 151802327-151802327 on Build GRCh38
rs182351649 Chr.2: 151802235-151802235 on Build GRCh38
rs532347374 Chr.2: 151802169-151802169 on Build GRCh38
rs532475926 Chr.2: 151802122-151802122 on Build GRCh38
rs1048222981 Chr.2: 151802416-151802416 on Build GRCh38

View Full Product Details