Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151815059-151815569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5A
Entrez Gene ID
26225
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
ARFLP5, ARL5
UniGene
Hs.470233
Gene Chromosome Location
Chr.2: 151800966-151828495 on Build GRCh38
Transcripts
NM_001037174.1 NM_012097.3 NM_177985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159192 c.73G>C p.D25H Chr.2: 151815173-151815173 on Build GRCh38
6028415 c.55G>C p.V19L Chr.2: 151815191-151815191 on Build GRCh38
6466337 c.97C>T p.L33F Chr.2: 151815149-151815149 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs528770819 Chr.2: 151815297-151815297 on Build GRCh38
rs115224977 Chr.2: 151815311-151815311 on Build GRCh38
rs1000853691 Chr.2: 151815236-151815236 on Build GRCh38
rs757088388 Chr.2: 151815191-151815191 on Build GRCh38
rs778362660 Chr.2: 151815206-151815206 on Build GRCh38
rs1007352443 Chr.2: 151815171-151815171 on Build GRCh38
rs181632373 Chr.2: 151815497-151815497 on Build GRCh38
rs530929420 Chr.2: 151815417-151815417 on Build GRCh38
rs750102786 Chr.2: 151815207-151815207 on Build GRCh38
rs776470413 Chr.2: 151815224-151815227 on Build GRCh38
rs769852401 Chr.2: 151815125-151815125 on Build GRCh38
rs922494082 Chr.2: 151815187-151815187 on Build GRCh38
rs139336208 Chr.2: 151815475-151815475 on Build GRCh38
rs202017084 Chr.2: 151815240-151815241 on Build GRCh38
rs773259282 Chr.2: 151815247-151815247 on Build GRCh38
rs766588581 Chr.2: 151815139-151815139 on Build GRCh38
rs537504272 Chr.2: 151815213-151815215 on Build GRCh38
rs928978960 Chr.2: 151815433-151815433 on Build GRCh38
rs371769147 Chr.2: 151815268-151815268 on Build GRCh38
rs976030994 Chr.2: 151815378-151815378 on Build GRCh38
rs918901892 Chr.2: 151815426-151815426 on Build GRCh38
rs890249966 Chr.2: 151815135-151815135 on Build GRCh38
rs922533852 Chr.2: 151815382-151815382 on Build GRCh38
rs1037970991 Chr.2: 151815338-151815338 on Build GRCh38
rs1038087218 Chr.2: 151815224-151815224 on Build GRCh38
rs948389356 Chr.2: 151815537-151815537 on Build GRCh38
rs764905991 Chr.2: 151815241-151815241 on Build GRCh38
rs776896976 Chr.2: 151815110-151815110 on Build GRCh38
rs771194130 Chr.2: 151815316-151815316 on Build GRCh38
rs763944827 Chr.2: 151815174-151815174 on Build GRCh38
rs753632465 Chr.2: 151815176-151815176 on Build GRCh38
rs747330393 Chr.2: 151815314-151815314 on Build GRCh38
rs1051424533 Chr.2: 151815460-151815460 on Build GRCh38
rs770229746 Chr.2: 151815241-151815241 on Build GRCh38
rs773491163 Chr.2: 151815132-151815132 on Build GRCh38
rs781730136 Chr.2: 151815224-151815224 on Build GRCh38
rs1017607322 Chr.2: 151815354-151815355 on Build GRCh38
rs747479786 Chr.2: 151815215-151815215 on Build GRCh38
rs374907847 Chr.2: 151815251-151815254 on Build GRCh38
rs755498339 Chr.2: 151815221-151815221 on Build GRCh38
rs559826564 Chr.2: 151815293-151815293 on Build GRCh38
rs2272126 Chr.2: 151815285-151815285 on Build GRCh38
rs1012559822 Chr.2: 151815276-151815277 on Build GRCh38
rs1039301990 Chr.2: 151815481-151815481 on Build GRCh38
rs768444143 Chr.2: 151815223-151815224 on Build GRCh38
rs768983662 Chr.2: 151815108-151815108 on Build GRCh38
rs748398115 Chr.2: 151815239-151815239 on Build GRCh38
rs1038137543 Chr.2: 151815480-151815480 on Build GRCh38
rs376329773 Chr.2: 151815450-151815450 on Build GRCh38
rs111646901 Chr.2: 151815214-151815216 on Build GRCh38
rs779586517 Chr.2: 151815209-151815209 on Build GRCh38
rs779578989 Chr.2: 151815376-151815376 on Build GRCh38
rs577500341 Chr.2: 151815271-151815271 on Build GRCh38
rs908374178 Chr.2: 151815440-151815440 on Build GRCh38
rs763087246 Chr.2: 151815138-151815138 on Build GRCh38
rs752660825 Chr.2: 151815164-151815164 on Build GRCh38
rs890235287 Chr.2: 151815472-151815472 on Build GRCh38
rs757850000 Chr.2: 151815208-151815208 on Build GRCh38
rs72621645 Chr.2: 151815326-151815326 on Build GRCh38
rs762085000 Chr.2: 151815114-151815114 on Build GRCh38
rs543220434 Chr.2: 151815170-151815170 on Build GRCh38
rs988199403 Chr.2: 151815283-151815283 on Build GRCh38
rs955304829 Chr.2: 151815397-151815397 on Build GRCh38
rs933836843 Chr.2: 151815194-151815194 on Build GRCh38

View Full Product Details