Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157327333-157327875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMN
Entrez Gene ID
57471
Gene Name
ermin
Gene Aliases
JN, KIAA1189
UniGene
Hs.443894
Gene Chromosome Location
Chr.2: 157318613-157327713 on Build GRCh38
Transcripts
NM_001009959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5423409 c.7+9A>G p.? Chr.2: 157327464-157327464 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032505682 Chr.2: 157327793-157327793 on Build GRCh38
rs553009965 Chr.2: 157327429-157327429 on Build GRCh38
rs747663540 Chr.2: 157327610-157327610 on Build GRCh38
rs191828311 Chr.2: 157327366-157327366 on Build GRCh38
rs990791272 Chr.2: 157327648-157327648 on Build GRCh38
rs771370548 Chr.2: 157327826-157327826 on Build GRCh38
rs143310506 Chr.2: 157327609-157327609 on Build GRCh38
rs765485475 Chr.2: 157327467-157327468 on Build GRCh38
rs767362194 Chr.2: 157327515-157327515 on Build GRCh38
rs144866227 Chr.2: 157327801-157327801 on Build GRCh38
rs990878710 Chr.2: 157327803-157327803 on Build GRCh38
rs775411911 Chr.2: 157327521-157327521 on Build GRCh38
rs917721466 Chr.2: 157327427-157327427 on Build GRCh38
rs920009526 Chr.2: 157327497-157327497 on Build GRCh38
rs561489174 Chr.2: 157327734-157327750 on Build GRCh38
rs768854852 Chr.2: 157327470-157327470 on Build GRCh38
rs747334915 Chr.2: 157327822-157327822 on Build GRCh38
rs1031060479 Chr.2: 157327743-157327743 on Build GRCh38
rs757155485 Chr.2: 157327374-157327377 on Build GRCh38
rs565468852 Chr.2: 157327791-157327791 on Build GRCh38
rs944265463 Chr.2: 157327625-157327625 on Build GRCh38
rs902278858 Chr.2: 157327787-157327787 on Build GRCh38
rs917809358 Chr.2: 157327650-157327650 on Build GRCh38
rs907978141 Chr.2: 157327703-157327703 on Build GRCh38
rs1020369872 Chr.2: 157327786-157327786 on Build GRCh38
rs949197503 Chr.2: 157327367-157327367 on Build GRCh38
rs566611260 Chr.2: 157327373-157327373 on Build GRCh38
rs969234157 Chr.2: 157327800-157327800 on Build GRCh38
rs189057044 Chr.2: 157327806-157327806 on Build GRCh38
rs1006240846 Chr.2: 157327417-157327417 on Build GRCh38
rs79051607 Chr.2: 157327691-157327691 on Build GRCh38
rs746241395 Chr.2: 157327452-157327452 on Build GRCh38
rs368866948 Chr.2: 157327460-157327460 on Build GRCh38
rs375401835 Chr.2: 157327479-157327479 on Build GRCh38
rs533881972 Chr.2: 157327377-157327377 on Build GRCh38
rs543980079 Chr.2: 157327720-157327720 on Build GRCh38
rs930790430 Chr.2: 157327464-157327464 on Build GRCh38
rs183560738 Chr.2: 157327407-157327407 on Build GRCh38
rs774300905 Chr.2: 157327501-157327501 on Build GRCh38
rs577844805 Chr.2: 157327529-157327529 on Build GRCh38
rs753393665 Chr.2: 157327424-157327424 on Build GRCh38
rs992003785 Chr.2: 157327769-157327769 on Build GRCh38
rs979498884 Chr.2: 157327372-157327372 on Build GRCh38
rs369016426 Chr.2: 157327534-157327534 on Build GRCh38
rs890679255 Chr.2: 157327747-157327747 on Build GRCh38
rs949231278 Chr.2: 157327660-157327660 on Build GRCh38
rs372348998 Chr.2: 157327468-157327468 on Build GRCh38
rs1000606265 Chr.2: 157327788-157327788 on Build GRCh38
rs991971096 Chr.2: 157327419-157327419 on Build GRCh38
rs114049002 Chr.2: 157327547-157327547 on Build GRCh38
rs776761142 Chr.2: 157327478-157327478 on Build GRCh38
rs758749882 Chr.2: 157327457-157327457 on Build GRCh38
rs1030996427 Chr.2: 157327490-157327490 on Build GRCh38
rs908512897 Chr.2: 157327839-157327839 on Build GRCh38
rs980784723 Chr.2: 157327487-157327487 on Build GRCh38
rs931955524 Chr.2: 157327735-157327735 on Build GRCh38
rs139282209 Chr.2: 157327406-157327406 on Build GRCh38
rs556215501 Chr.2: 157327434-157327434 on Build GRCh38
rs1010378236 Chr.2: 157327780-157327780 on Build GRCh38
rs16841621 Chr.2: 157327590-157327590 on Build GRCh38
rs899959730 Chr.2: 157327697-157327697 on Build GRCh38
rs542717323 Chr.2: 157327645-157327645 on Build GRCh38
rs979068950 Chr.2: 157327596-157327596 on Build GRCh38
rs758848647 Chr.2: 157327710-157327710 on Build GRCh38
rs759250937 Chr.2: 157327503-157327503 on Build GRCh38
rs554617130 Chr.2: 157327671-157327671 on Build GRCh38
rs566715773 Chr.2: 157327832-157327832 on Build GRCh38
rs753576492 Chr.2: 157327408-157327408 on Build GRCh38
rs1052050175 Chr.2: 157327744-157327744 on Build GRCh38
rs1038183004 Chr.2: 157327716-157327716 on Build GRCh38
rs750623382 Chr.2: 157327470-157327471 on Build GRCh38
rs1010347084 Chr.2: 157327542-157327542 on Build GRCh38

View Full Product Details