Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162017016-162017516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP4
Entrez Gene ID
1803
Gene Name
dipeptidyl peptidase 4
Gene Aliases
ADABP, ADCP2, CD26, DPPIV, TP103
UniGene
Hs.368912
Gene Chromosome Location
Chr.2: 161992241-162074542 on Build GRCh38
Transcripts
NM_001935.3 XM_005246371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3311207 c.1462G>A p.D488N Chr.2: 162017114-162017114 on Build GRCh38
4085995 c.1459A>C p.N487H Chr.2: 162017117-162017117 on Build GRCh38
160572 c.1456G>A p.V486M Chr.2: 162017120-162017120 on Build GRCh38
5559645 c.1446A>G p.L482L Chr.2: 162017130-162017130 on Build GRCh38
3391040 c.1421G>A p.G474D Chr.2: 162017155-162017155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576990878 Chr.2: 162017492-162017492 on Build GRCh38
rs891695511 Chr.2: 162017218-162017218 on Build GRCh38
rs79932012 Chr.2: 162017258-162017258 on Build GRCh38
rs916062783 Chr.2: 162017484-162017484 on Build GRCh38
rs748624483 Chr.2: 162017079-162017079 on Build GRCh38
rs201175648 Chr.2: 162017142-162017142 on Build GRCh38
rs773782419 Chr.2: 162017214-162017214 on Build GRCh38
rs755235894 Chr.2: 162017175-162017175 on Build GRCh38
rs1019394815 Chr.2: 162017351-162017351 on Build GRCh38
rs201273765 Chr.2: 162017097-162017097 on Build GRCh38
rs957432359 Chr.2: 162017437-162017437 on Build GRCh38
rs537057257 Chr.2: 162017375-162017375 on Build GRCh38
rs1052754964 Chr.2: 162017209-162017209 on Build GRCh38
rs201457658 Chr.2: 162017188-162017188 on Build GRCh38
rs1045447461 Chr.2: 162017047-162017047 on Build GRCh38
rs558654885 Chr.2: 162017465-162017465 on Build GRCh38
rs538420868 Chr.2: 162017230-162017230 on Build GRCh38
rs977930024 Chr.2: 162017421-162017421 on Build GRCh38
rs756507168 Chr.2: 162017185-162017185 on Build GRCh38
rs1015627357 Chr.2: 162017361-162017361 on Build GRCh38
rs896604503 Chr.2: 162017318-162017318 on Build GRCh38
rs771616427 Chr.2: 162017425-162017425 on Build GRCh38
rs760175632 Chr.2: 162017173-162017174 on Build GRCh38
rs1042867639 Chr.2: 162017276-162017276 on Build GRCh38
rs202231326 Chr.2: 162017121-162017121 on Build GRCh38
rs770446040 Chr.2: 162017113-162017113 on Build GRCh38
rs376620801 Chr.2: 162017380-162017380 on Build GRCh38
rs756700985 Chr.2: 162017080-162017080 on Build GRCh38
rs760679238 Chr.2: 162017124-162017124 on Build GRCh38
rs756768949 Chr.2: 162017201-162017201 on Build GRCh38
rs778642213 Chr.2: 162017350-162017350 on Build GRCh38
rs954002424 Chr.2: 162017295-162017295 on Build GRCh38
rs200433211 Chr.2: 162017136-162017136 on Build GRCh38
rs186992080 Chr.2: 162017371-162017371 on Build GRCh38
rs575950626 Chr.2: 162017374-162017374 on Build GRCh38
rs775495576 Chr.2: 162017122-162017122 on Build GRCh38
rs1008083454 Chr.2: 162017308-162017308 on Build GRCh38
rs761253936 Chr.2: 162017388-162017388 on Build GRCh38
rs764110404 Chr.2: 162017128-162017128 on Build GRCh38
rs367770477 Chr.2: 162017184-162017184 on Build GRCh38
rs199653013 Chr.2: 162017199-162017199 on Build GRCh38
rs772501868 Chr.2: 162017108-162017108 on Build GRCh38
rs17848906 Chr.2: 162017042-162017042 on Build GRCh38
rs905700492 Chr.2: 162017288-162017288 on Build GRCh38
rs565773377 Chr.2: 162017292-162017292 on Build GRCh38
rs201980305 Chr.2: 162017171-162017171 on Build GRCh38
rs536540689 Chr.2: 162017337-162017337 on Build GRCh38
rs866806811 Chr.2: 162017429-162017429 on Build GRCh38
rs906855202 Chr.2: 162017145-162017145 on Build GRCh38
rs777088875 Chr.2: 162017099-162017099 on Build GRCh38
rs1027123042 Chr.2: 162017389-162017389 on Build GRCh38
rs774072321 Chr.2: 162017115-162017115 on Build GRCh38
rs748698131 Chr.2: 162017196-162017196 on Build GRCh38
rs912191138 Chr.2: 162017190-162017190 on Build GRCh38
rs752205081 Chr.2: 162017174-162017174 on Build GRCh38
rs985623178 Chr.2: 162017163-162017163 on Build GRCh38
rs974251608 Chr.2: 162017100-162017100 on Build GRCh38
rs375295208 Chr.2: 162017063-162017063 on Build GRCh38
rs200437045 Chr.2: 162017179-162017179 on Build GRCh38
rs199952981 Chr.2: 162017058-162017058 on Build GRCh38
rs373591144 Chr.2: 162017158-162017158 on Build GRCh38
rs1013577075 Chr.2: 162017356-162017356 on Build GRCh38

View Full Product Details