Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 164684956-164685500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COBLL1
Entrez Gene ID
22837
Gene Name
cordon-bleu WH2 repeat protein like 1
Gene Aliases
COBLR1
UniGene
Hs.470457
Gene Chromosome Location
Chr.2: 164680185-164842168 on Build GRCh38
Transcripts
NM_001278458.1 NM_001278460.1 NM_001278461.1 NM_014900.4

SNP Detail

SNP ID Chromosome Location
rs1050536924 Chr.2: 164685189-164685189 on Build GRCh38
rs11558902 Chr.2: 164685273-164685273 on Build GRCh38
rs539698777 Chr.2: 164685112-164685112 on Build GRCh38
rs992891594 Chr.2: 164685287-164685287 on Build GRCh38
rs547317465 Chr.2: 164685119-164685119 on Build GRCh38
rs772766433 Chr.2: 164685023-164685023 on Build GRCh38
rs1052883442 Chr.2: 164685241-164685241 on Build GRCh38
rs926209231 Chr.2: 164685365-164685365 on Build GRCh38
rs576817557 Chr.2: 164685344-164685344 on Build GRCh38
rs890533289 Chr.2: 164685190-164685190 on Build GRCh38
rs901064460 Chr.2: 164685188-164685188 on Build GRCh38
rs968560506 Chr.2: 164685314-164685314 on Build GRCh38
rs907544208 Chr.2: 164685464-164685464 on Build GRCh38
rs1037925233 Chr.2: 164685171-164685171 on Build GRCh38
rs970141102 Chr.2: 164685309-164685309 on Build GRCh38
rs944694746 Chr.2: 164685186-164685186 on Build GRCh38
rs992565088 Chr.2: 164685441-164685441 on Build GRCh38
rs778981609 Chr.2: 164685178-164685178 on Build GRCh38
rs917483417 Chr.2: 164684988-164684988 on Build GRCh38
rs1021503414 Chr.2: 164685312-164685312 on Build GRCh38
rs574550403 Chr.2: 164685231-164685231 on Build GRCh38
rs926093934 Chr.2: 164685146-164685146 on Build GRCh38
rs879223041 Chr.2: 164685469-164685469 on Build GRCh38
rs1028259171 Chr.2: 164685235-164685235 on Build GRCh38
rs959763133 Chr.2: 164685436-164685436 on Build GRCh38
rs556634731 Chr.2: 164685299-164685299 on Build GRCh38
rs908141048 Chr.2: 164685109-164685109 on Build GRCh38
rs888249523 Chr.2: 164685243-164685243 on Build GRCh38
rs961698717 Chr.2: 164685465-164685465 on Build GRCh38
rs980222469 Chr.2: 164685140-164685140 on Build GRCh38
rs1001315863 Chr.2: 164685319-164685319 on Build GRCh38
rs1015350680 Chr.2: 164685267-164685267 on Build GRCh38
rs936179858 Chr.2: 164685170-164685170 on Build GRCh38
rs936168480 Chr.2: 164685398-164685398 on Build GRCh38
rs1023998149 Chr.2: 164685289-164685289 on Build GRCh38
rs377168665 Chr.2: 164685212-164685212 on Build GRCh38
rs988970766 Chr.2: 164685421-164685421 on Build GRCh38
rs772077773 Chr.2: 164685390-164685390 on Build GRCh38
rs949027961 Chr.2: 164685056-164685056 on Build GRCh38
rs114240686 Chr.2: 164685179-164685179 on Build GRCh38
rs73013644 Chr.2: 164685187-164685187 on Build GRCh38
rs1005393578 Chr.2: 164685255-164685255 on Build GRCh38

View Full Product Details