Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 164695314-164695815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COBLL1
Entrez Gene ID
22837
Gene Name
cordon-bleu WH2 repeat protein like 1
Gene Aliases
COBLR1
UniGene
Hs.470457
Gene Chromosome Location
Chr.2: 164680185-164842168 on Build GRCh38
Transcripts
NM_001278458.1 NM_001278460.1 NM_001278461.1 NM_014900.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6204196 c.1834A>G p.I612V Chr.2: 164695672-164695672 on Build GRCh38
6054452 c.2024C>G p.T675S Chr.2: 164695683-164695683 on Build GRCh38
3568664 c.1739C>A p.T580K Chr.2: 164695767-164695767 on Build GRCh38
7422073 c.1906A>T p.T636S Chr.2: 164695801-164695801 on Build GRCh38
127676 c.2025C>T p.N675N Chr.2: 164695481-164695481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554340557 Chr.2: 164695765-164695765 on Build GRCh38
rs778890399 Chr.2: 164695433-164695433 on Build GRCh38
rs754907910 Chr.2: 164695420-164695420 on Build GRCh38
rs769303043 Chr.2: 164695480-164695480 on Build GRCh38
rs776363347 Chr.2: 164695767-164695767 on Build GRCh38
rs201560157 Chr.2: 164695665-164695665 on Build GRCh38
rs761837679 Chr.2: 164695776-164695776 on Build GRCh38
rs372845868 Chr.2: 164695620-164695620 on Build GRCh38
rs200707412 Chr.2: 164695571-164695571 on Build GRCh38
rs201823188 Chr.2: 164695455-164695455 on Build GRCh38
rs569020159 Chr.2: 164695678-164695678 on Build GRCh38
rs750021681 Chr.2: 164695410-164695410 on Build GRCh38
rs372326092 Chr.2: 164695427-164695427 on Build GRCh38
rs138701146 Chr.2: 164695376-164695376 on Build GRCh38
rs762638639 Chr.2: 164695499-164695499 on Build GRCh38
rs565148370 Chr.2: 164695628-164695628 on Build GRCh38
rs368813678 Chr.2: 164695672-164695672 on Build GRCh38
rs140678147 Chr.2: 164695363-164695363 on Build GRCh38
rs369747052 Chr.2: 164695378-164695378 on Build GRCh38
rs747443638 Chr.2: 164695447-164695447 on Build GRCh38
rs755269804 Chr.2: 164695694-164695694 on Build GRCh38
rs749929749 Chr.2: 164695781-164695782 on Build GRCh38
rs761539089 Chr.2: 164695441-164695441 on Build GRCh38
rs141221520 Chr.2: 164695788-164695788 on Build GRCh38
rs116305649 Chr.2: 164695712-164695712 on Build GRCh38
rs753493898 Chr.2: 164695340-164695342 on Build GRCh38
rs998564366 Chr.2: 164695727-164695727 on Build GRCh38
rs775260718 Chr.2: 164695756-164695756 on Build GRCh38
rs147783923 Chr.2: 164695745-164695745 on Build GRCh38
rs752592570 Chr.2: 164695527-164695527 on Build GRCh38
rs766481813 Chr.2: 164695684-164695684 on Build GRCh38
rs780433619 Chr.2: 164695445-164695445 on Build GRCh38
rs781668809 Chr.2: 164695702-164695702 on Build GRCh38
rs910827222 Chr.2: 164695454-164695454 on Build GRCh38
rs779139359 Chr.2: 164695368-164695368 on Build GRCh38
rs951502870 Chr.2: 164695679-164695679 on Build GRCh38
rs200953353 Chr.2: 164695637-164695637 on Build GRCh38
rs545164827 Chr.2: 164695545-164695545 on Build GRCh38
rs777218156 Chr.2: 164695466-164695466 on Build GRCh38
rs754961983 Chr.2: 164695351-164695351 on Build GRCh38
rs770489606 Chr.2: 164695654-164695654 on Build GRCh38
rs143836010 Chr.2: 164695353-164695353 on Build GRCh38
rs751353990 Chr.2: 164695416-164695416 on Build GRCh38
rs963564080 Chr.2: 164695507-164695507 on Build GRCh38
rs778086963 Chr.2: 164695713-164695713 on Build GRCh38
rs145390469 Chr.2: 164695723-164695723 on Build GRCh38
rs146983191 Chr.2: 164695407-164695407 on Build GRCh38
rs766016487 Chr.2: 164695504-164695504 on Build GRCh38
rs746083849 Chr.2: 164695373-164695373 on Build GRCh38
rs760442791 Chr.2: 164695757-164695757 on Build GRCh38
rs758596807 Chr.2: 164695443-164695443 on Build GRCh38
rs748647602 Chr.2: 164695630-164695630 on Build GRCh38
rs149530470 Chr.2: 164695412-164695412 on Build GRCh38
rs764881808 Chr.2: 164695569-164695569 on Build GRCh38
rs143738592 Chr.2: 164695536-164695536 on Build GRCh38
rs117456878 Chr.2: 164695653-164695653 on Build GRCh38
rs369864612 Chr.2: 164695505-164695505 on Build GRCh38
rs761440844 Chr.2: 164695434-164695435 on Build GRCh38
rs148694678 Chr.2: 164695429-164695429 on Build GRCh38
rs970906462 Chr.2: 164695604-164695604 on Build GRCh38
rs1029966535 Chr.2: 164695345-164695345 on Build GRCh38
rs780195921 Chr.2: 164695586-164695586 on Build GRCh38
rs751788114 Chr.2: 164695691-164695691 on Build GRCh38
rs548981667 Chr.2: 164695676-164695676 on Build GRCh38
rs752645823 Chr.2: 164695426-164695426 on Build GRCh38
rs111939559 Chr.2: 164695382-164695382 on Build GRCh38
rs201427085 Chr.2: 164695383-164695383 on Build GRCh38
rs938051215 Chr.2: 164695644-164695644 on Build GRCh38
rs766514749 Chr.2: 164695781-164695781 on Build GRCh38
rs539988251 Chr.2: 164695645-164695645 on Build GRCh38
rs764849088 Chr.2: 164695409-164695409 on Build GRCh38
rs764003186 Chr.2: 164695675-164695675 on Build GRCh38
rs774048874 Chr.2: 164695421-164695421 on Build GRCh38
rs181143305 Chr.2: 164695375-164695375 on Build GRCh38
rs748666097 Chr.2: 164695469-164695469 on Build GRCh38
rs759271742 Chr.2: 164695518-164695518 on Build GRCh38
rs1023877689 Chr.2: 164695585-164695585 on Build GRCh38
rs751469877 Chr.2: 164695341-164695341 on Build GRCh38
rs534277685 Chr.2: 164695749-164695749 on Build GRCh38
rs749807720 Chr.2: 164695722-164695722 on Build GRCh38
rs750508324 Chr.2: 164695563-164695563 on Build GRCh38
rs150990193 Chr.2: 164695657-164695657 on Build GRCh38
rs189600717 Chr.2: 164695354-164695354 on Build GRCh38
rs772650612 Chr.2: 164695492-164695492 on Build GRCh38
rs576695380 Chr.2: 164695526-164695526 on Build GRCh38
rs146060190 Chr.2: 164695533-164695533 on Build GRCh38
rs747245315 Chr.2: 164695614-164695614 on Build GRCh38
rs775312261 Chr.2: 164695667-164695667 on Build GRCh38
rs781710185 Chr.2: 164695629-164695629 on Build GRCh38

View Full Product Details