Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165756964-165757469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT3
Entrez Gene ID
2591
Gene Name
polypeptide N-acetylgalactosaminyltransferase 3
Gene Aliases
GalNAc-T3, HFTC, HHS
UniGene
Hs.170986
Gene Chromosome Location
Chr.2: 165747803-165796352 on Build GRCh38
Transcripts
NM_004482.3 XM_005246449.1 XM_011510929.1 XM_017003770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4686293 c.1252C>T p.R418C Chr.2: 165757187-165757187 on Build GRCh38
7327755 c.1311delT p.R438fs*39 Chr.2: 165757128-165757128 on Build GRCh38
6006798 c.1376C>A p.A459E Chr.2: 165757063-165757063 on Build GRCh38
4808859 c.1296T>C p.I432I Chr.2: 165757143-165757143 on Build GRCh38
3312716 c.1313G>A p.R438H Chr.2: 165757126-165757126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530681634 Chr.2: 165757187-165757187 on Build GRCh38
rs369192067 Chr.2: 165757001-165757001 on Build GRCh38
rs763773431 Chr.2: 165757031-165757035 on Build GRCh38
rs769087118 Chr.2: 165757283-165757283 on Build GRCh38
rs1041076405 Chr.2: 165757097-165757097 on Build GRCh38
rs753149443 Chr.2: 165757017-165757017 on Build GRCh38
rs778454917 Chr.2: 165757035-165757035 on Build GRCh38
rs761245509 Chr.2: 165757149-165757149 on Build GRCh38
rs373934847 Chr.2: 165757039-165757039 on Build GRCh38
rs776071404 Chr.2: 165757047-165757049 on Build GRCh38
rs990047792 Chr.2: 165757405-165757405 on Build GRCh38
rs1014315018 Chr.2: 165757424-165757424 on Build GRCh38
rs764741963 Chr.2: 165757175-165757175 on Build GRCh38
rs772171197 Chr.2: 165757105-165757105 on Build GRCh38
rs372866842 Chr.2: 165757279-165757279 on Build GRCh38
rs945095657 Chr.2: 165757052-165757052 on Build GRCh38
rs149017052 Chr.2: 165757212-165757212 on Build GRCh38
rs373760885 Chr.2: 165757055-165757055 on Build GRCh38
rs761323471 Chr.2: 165757018-165757018 on Build GRCh38
rs753440832 Chr.2: 165757031-165757031 on Build GRCh38
rs781073900 Chr.2: 165757068-165757068 on Build GRCh38
rs143396063 Chr.2: 165757186-165757186 on Build GRCh38
rs41268663 Chr.2: 165757278-165757278 on Build GRCh38
rs546559490 Chr.2: 165757360-165757360 on Build GRCh38
rs150937941 Chr.2: 165757245-165757245 on Build GRCh38
rs953115493 Chr.2: 165757172-165757172 on Build GRCh38
rs377375741 Chr.2: 165757029-165757029 on Build GRCh38
rs746790593 Chr.2: 165757047-165757047 on Build GRCh38
rs913426773 Chr.2: 165757276-165757276 on Build GRCh38
rs779970449 Chr.2: 165757040-165757040 on Build GRCh38
rs751626865 Chr.2: 165757264-165757264 on Build GRCh38
rs751393704 Chr.2: 165757195-165757195 on Build GRCh38
rs199930810 Chr.2: 165757120-165757120 on Build GRCh38
rs756789966 Chr.2: 165757033-165757033 on Build GRCh38
rs970980017 Chr.2: 165757313-165757313 on Build GRCh38
rs181587563 Chr.2: 165757010-165757010 on Build GRCh38
rs777464821 Chr.2: 165757116-165757116 on Build GRCh38
rs772378720 Chr.2: 165757277-165757277 on Build GRCh38
rs750849793 Chr.2: 165757003-165757003 on Build GRCh38
rs764730691 Chr.2: 165757127-165757127 on Build GRCh38
rs761090844 Chr.2: 165757121-165757121 on Build GRCh38
rs146096050 Chr.2: 165757091-165757091 on Build GRCh38
rs777232394 Chr.2: 165757138-165757138 on Build GRCh38
rs71426780 Chr.2: 165757357-165757357 on Build GRCh38
rs962402343 Chr.2: 165757262-165757262 on Build GRCh38
rs761371894 Chr.2: 165757250-165757250 on Build GRCh38
rs757287453 Chr.2: 165757232-165757232 on Build GRCh38
rs745875008 Chr.2: 165757330-165757330 on Build GRCh38
rs574830911 Chr.2: 165757363-165757363 on Build GRCh38
rs748019351 Chr.2: 165757227-165757227 on Build GRCh38
rs749399439 Chr.2: 165757072-165757072 on Build GRCh38
rs181865244 Chr.2: 165757331-165757331 on Build GRCh38
rs892207276 Chr.2: 165757048-165757048 on Build GRCh38
rs759839512 Chr.2: 165757006-165757006 on Build GRCh38
rs746035517 Chr.2: 165757100-165757100 on Build GRCh38
rs528982346 Chr.2: 165757425-165757425 on Build GRCh38
rs897693932 Chr.2: 165757439-165757439 on Build GRCh38
rs771096034 Chr.2: 165756997-165756997 on Build GRCh38
rs143799357 Chr.2: 165757202-165757202 on Build GRCh38
rs1027728109 Chr.2: 165757137-165757137 on Build GRCh38
rs1053556617 Chr.2: 165757042-165757042 on Build GRCh38
rs377389049 Chr.2: 165757070-165757070 on Build GRCh38
rs757320645 Chr.2: 165757218-165757218 on Build GRCh38
rs1025370818 Chr.2: 165757299-165757299 on Build GRCh38
rs749900386 Chr.2: 165757179-165757179 on Build GRCh38
rs367803423 Chr.2: 165757338-165757338 on Build GRCh38
rs770992382 Chr.2: 165757075-165757075 on Build GRCh38

View Full Product Details