Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168560980-168561482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS6
Entrez Gene ID
253782
Gene Name
ceramide synthase 6
Gene Aliases
CERS5, LASS6
UniGene
Hs.506829
Gene Chromosome Location
Chr.2: 168456249-168775134 on Build GRCh38
Transcripts
NM_001256126.1 NM_203463.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6326312 c.343C>T p.R115C Chr.2: 168561258-168561258 on Build GRCh38
7296327 c.406A>G p.M136V Chr.2: 168561321-168561321 on Build GRCh38
1009068 c.390G>A p.T130T Chr.2: 168561305-168561305 on Build GRCh38
3314525 c.297G>T p.L99F Chr.2: 168561212-168561212 on Build GRCh38
4086528 c.394T>C p.F132L Chr.2: 168561309-168561309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761730340 Chr.2: 168561212-168561212 on Build GRCh38
rs747876626 Chr.2: 168561360-168561360 on Build GRCh38
rs779475869 Chr.2: 168561277-168561277 on Build GRCh38
rs775949401 Chr.2: 168561044-168561044 on Build GRCh38
rs896440892 Chr.2: 168561117-168561117 on Build GRCh38
rs912026539 Chr.2: 168561250-168561250 on Build GRCh38
rs956301267 Chr.2: 168561230-168561230 on Build GRCh38
rs771791504 Chr.2: 168561011-168561011 on Build GRCh38
rs527813174 Chr.2: 168561178-168561178 on Build GRCh38
rs1009214309 Chr.2: 168561377-168561377 on Build GRCh38
rs775056743 Chr.2: 168561143-168561143 on Build GRCh38
rs138982215 Chr.2: 168561270-168561270 on Build GRCh38
rs768009413 Chr.2: 168561281-168561281 on Build GRCh38
rs750399418 Chr.2: 168561224-168561224 on Build GRCh38
rs773137836 Chr.2: 168561302-168561302 on Build GRCh38
rs750759964 Chr.2: 168561326-168561326 on Build GRCh38
rs765626641 Chr.2: 168561324-168561324 on Build GRCh38
rs766320076 Chr.2: 168561304-168561304 on Build GRCh38
rs372372722 Chr.2: 168561305-168561305 on Build GRCh38
rs570390667 Chr.2: 168561100-168561100 on Build GRCh38
rs944952016 Chr.2: 168561023-168561023 on Build GRCh38
rs769541022 Chr.2: 168561369-168561369 on Build GRCh38
rs758637978 Chr.2: 168561334-168561334 on Build GRCh38
rs1023529232 Chr.2: 168561431-168561431 on Build GRCh38
rs756341540 Chr.2: 168561258-168561258 on Build GRCh38
rs757461077 Chr.2: 168561322-168561322 on Build GRCh38
rs192769684 Chr.2: 168561433-168561433 on Build GRCh38
rs762970617 Chr.2: 168561231-168561231 on Build GRCh38
rs746548982 Chr.2: 168561280-168561280 on Build GRCh38
rs35110784 Chr.2: 168561240-168561241 on Build GRCh38
rs369699507 Chr.2: 168561298-168561298 on Build GRCh38
rs1040321978 Chr.2: 168561402-168561402 on Build GRCh38
rs1012139001 Chr.2: 168561141-168561141 on Build GRCh38
rs763042459 Chr.2: 168561303-168561303 on Build GRCh38
rs576393642 Chr.2: 168561417-168561417 on Build GRCh38
rs369637753 Chr.2: 168561371-168561371 on Build GRCh38
rs747406507 Chr.2: 168561347-168561347 on Build GRCh38
rs756112491 Chr.2: 168561126-168561126 on Build GRCh38
rs754529294 Chr.2: 168561354-168561354 on Build GRCh38
rs769717620 Chr.2: 168561176-168561176 on Build GRCh38
rs183059178 Chr.2: 168561188-168561188 on Build GRCh38
rs777345252 Chr.2: 168561162-168561162 on Build GRCh38
rs749131814 Chr.2: 168561373-168561373 on Build GRCh38
rs765057450 Chr.2: 168561217-168561217 on Build GRCh38
rs766605540 Chr.2: 168561444-168561444 on Build GRCh38
rs769668537 Chr.2: 168561332-168561332 on Build GRCh38
rs1004741486 Chr.2: 168561114-168561114 on Build GRCh38
rs775742017 Chr.2: 168561385-168561385 on Build GRCh38
rs531100865 Chr.2: 168561187-168561187 on Build GRCh38
rs187342645 Chr.2: 168561276-168561276 on Build GRCh38
rs141128121 Chr.2: 168561247-168561247 on Build GRCh38
rs757626907 Chr.2: 168561268-168561268 on Build GRCh38
rs1038683739 Chr.2: 168561024-168561024 on Build GRCh38
rs556558587 Chr.2: 168561299-168561299 on Build GRCh38
rs374976492 Chr.2: 168561294-168561294 on Build GRCh38
rs930406251 Chr.2: 168561045-168561045 on Build GRCh38
rs552802360 Chr.2: 168561107-168561107 on Build GRCh38
rs896282749 Chr.2: 168561352-168561352 on Build GRCh38
rs368464104 Chr.2: 168561099-168561099 on Build GRCh38
rs1028384966 Chr.2: 168561180-168561180 on Build GRCh38
rs752792437 Chr.2: 168561251-168561251 on Build GRCh38
rs1022976574 Chr.2: 168561144-168561144 on Build GRCh38
rs747585832 Chr.2: 168561283-168561285 on Build GRCh38
rs767376329 Chr.2: 168561272-168561272 on Build GRCh38
rs961924691 Chr.2: 168561271-168561271 on Build GRCh38
rs532879010 Chr.2: 168561101-168561101 on Build GRCh38
rs778081351 Chr.2: 168561259-168561259 on Build GRCh38
rs566153680 Chr.2: 168561116-168561116 on Build GRCh38
rs940429464 Chr.2: 168561317-168561317 on Build GRCh38
rs777025181 Chr.2: 168561334-168561334 on Build GRCh38
rs780835262 Chr.2: 168561357-168561357 on Build GRCh38
rs368138259 Chr.2: 168561267-168561267 on Build GRCh38
rs754075843 Chr.2: 168561321-168561321 on Build GRCh38
rs375149776 Chr.2: 168561318-168561318 on Build GRCh38

View Full Product Details