Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168769368-168769894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS6
Entrez Gene ID
253782
Gene Name
ceramide synthase 6
Gene Aliases
CERS5, LASS6
UniGene
Hs.506829
Gene Chromosome Location
Chr.2: 168456249-168775134 on Build GRCh38
Transcripts
NM_001256126.1 NM_203463.2 XM_005246440.4 XM_017003749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6627380 c.1018C>T p.R340* Chr.2: 168769525-168769525 on Build GRCh38
1009075 c.1086G>A p.A362A Chr.2: 168769593-168769593 on Build GRCh38
5394868 c.1076C>T p.P359L Chr.2: 168769583-168769583 on Build GRCh38
3314543 c.1085C>T p.A362V Chr.2: 168769592-168769592 on Build GRCh38
7361004 c.1146G>T p.M382I Chr.2: 168769653-168769653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776193867 Chr.2: 168769687-168769687 on Build GRCh38
rs775718724 Chr.2: 168769808-168769808 on Build GRCh38
rs764877283 Chr.2: 168769704-168769704 on Build GRCh38
rs775881458 Chr.2: 168769672-168769672 on Build GRCh38
rs913746245 Chr.2: 168769847-168769847 on Build GRCh38
rs773331109 Chr.2: 168769534-168769534 on Build GRCh38
rs778650794 Chr.2: 168769495-168769495 on Build GRCh38
rs760129955 Chr.2: 168769587-168769587 on Build GRCh38
rs756901461 Chr.2: 168769593-168769593 on Build GRCh38
rs571903257 Chr.2: 168769855-168769855 on Build GRCh38
rs748601926 Chr.2: 168769473-168769474 on Build GRCh38
rs757021696 Chr.2: 168769487-168769487 on Build GRCh38
rs1040290041 Chr.2: 168769438-168769438 on Build GRCh38
rs201175362 Chr.2: 168769604-168769604 on Build GRCh38
rs1040340451 Chr.2: 168769721-168769721 on Build GRCh38
rs980494123 Chr.2: 168769697-168769697 on Build GRCh38
rs185848286 Chr.2: 168769859-168769859 on Build GRCh38
rs936642573 Chr.2: 168769712-168769712 on Build GRCh38
rs768121719 Chr.2: 168769684-168769684 on Build GRCh38
rs576654525 Chr.2: 168769801-168769801 on Build GRCh38
rs982340851 Chr.2: 168769568-168769568 on Build GRCh38
rs749676502 Chr.2: 168769844-168769844 on Build GRCh38
rs565003592 Chr.2: 168769500-168769500 on Build GRCh38
rs369195108 Chr.2: 168769677-168769677 on Build GRCh38
rs535039602 Chr.2: 168769600-168769600 on Build GRCh38
rs749539202 Chr.2: 168769644-168769644 on Build GRCh38
rs771460989 Chr.2: 168769540-168769540 on Build GRCh38
rs925250699 Chr.2: 168769407-168769407 on Build GRCh38
rs961750710 Chr.2: 168769620-168769620 on Build GRCh38
rs936490323 Chr.2: 168769442-168769442 on Build GRCh38
rs761258895 Chr.2: 168769698-168769698 on Build GRCh38
rs147015284 Chr.2: 168769531-168769531 on Build GRCh38
rs570312530 Chr.2: 168769456-168769456 on Build GRCh38
rs763655070 Chr.2: 168769459-168769459 on Build GRCh38
rs866198654 Chr.2: 168769646-168769646 on Build GRCh38
rs772096732 Chr.2: 168769526-168769526 on Build GRCh38
rs753487439 Chr.2: 168769479-168769479 on Build GRCh38
rs771262751 Chr.2: 168769649-168769649 on Build GRCh38
rs931837192 Chr.2: 168769751-168769751 on Build GRCh38
rs545674469 Chr.2: 168769823-168769823 on Build GRCh38
rs566046243 Chr.2: 168769572-168769572 on Build GRCh38
rs201799372 Chr.2: 168769628-168769628 on Build GRCh38
rs780637717 Chr.2: 168769415-168769415 on Build GRCh38
rs372041111 Chr.2: 168769622-168769622 on Build GRCh38
rs747811536 Chr.2: 168769590-168769590 on Build GRCh38
rs774657184 Chr.2: 168769651-168769651 on Build GRCh38
rs373049662 Chr.2: 168769709-168769709 on Build GRCh38
rs1006018825 Chr.2: 168769498-168769498 on Build GRCh38
rs199916442 Chr.2: 168769571-168769571 on Build GRCh38
rs944641913 Chr.2: 168769396-168769396 on Build GRCh38
rs771301891 Chr.2: 168769849-168769849 on Build GRCh38
rs575986947 Chr.2: 168769758-168769758 on Build GRCh38
rs987895539 Chr.2: 168769802-168769802 on Build GRCh38
rs530196124 Chr.2: 168769430-168769430 on Build GRCh38
rs372304719 Chr.2: 168769496-168769496 on Build GRCh38
rs748181431 Chr.2: 168769530-168769530 on Build GRCh38
rs181296302 Chr.2: 168769451-168769451 on Build GRCh38
rs1028450765 Chr.2: 168769485-168769485 on Build GRCh38
rs376775078 Chr.2: 168769626-168769626 on Build GRCh38
rs770065647 Chr.2: 168769609-168769617 on Build GRCh38
rs537188689 Chr.2: 168769726-168769726 on Build GRCh38
rs779998285 Chr.2: 168769623-168769623 on Build GRCh38
rs369260392 Chr.2: 168769639-168769639 on Build GRCh38
rs1015138015 Chr.2: 168769610-168769610 on Build GRCh38
rs138914884 Chr.2: 168769527-168769527 on Build GRCh38
rs749506102 Chr.2: 168769535-168769535 on Build GRCh38
rs1048935429 Chr.2: 168769794-168769794 on Build GRCh38
rs7355470 Chr.2: 168769854-168769854 on Build GRCh38
rs891878553 Chr.2: 168769493-168769493 on Build GRCh38
rs746238053 Chr.2: 168769655-168769655 on Build GRCh38
rs900417712 Chr.2: 168769746-168769746 on Build GRCh38
rs200798086 Chr.2: 168769499-168769499 on Build GRCh38
rs772522040 Chr.2: 168769658-168769658 on Build GRCh38
rs760178446 Chr.2: 168769557-168769557 on Build GRCh38
rs774849482 Chr.2: 168769555-168769555 on Build GRCh38
rs753478893 Chr.2: 168769592-168769592 on Build GRCh38

View Full Product Details