Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168907677-168908176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2 XM_011511565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5058238 c.809G>A p.G270D Chr.2: 168907820-168907820 on Build GRCh38
3379851 c.861C>A p.N287K Chr.2: 168907872-168907872 on Build GRCh38
4989086 c.861C>G p.N287K Chr.2: 168907872-168907872 on Build GRCh38
6842944 c.910C>A p.L304M Chr.2: 168907921-168907921 on Build GRCh38
6842939 c.873C>G p.S291R Chr.2: 168907884-168907884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780112185 Chr.2: 168907780-168907780 on Build GRCh38
rs753707145 Chr.2: 168907776-168907776 on Build GRCh38
rs764158428 Chr.2: 168907775-168907775 on Build GRCh38
rs541931958 Chr.2: 168907889-168907889 on Build GRCh38
rs201323443 Chr.2: 168907974-168907974 on Build GRCh38
rs147360987 Chr.2: 168907759-168907759 on Build GRCh38
rs768801451 Chr.2: 168907723-168907723 on Build GRCh38
rs770384960 Chr.2: 168907749-168907749 on Build GRCh38
rs141216504 Chr.2: 168907708-168907708 on Build GRCh38
rs780589028 Chr.2: 168908095-168908095 on Build GRCh38
rs750777548 Chr.2: 168908088-168908088 on Build GRCh38
rs754077966 Chr.2: 168908061-168908061 on Build GRCh38
rs149616455 Chr.2: 168907756-168907756 on Build GRCh38
rs139564794 Chr.2: 168907788-168907788 on Build GRCh38
rs1043706366 Chr.2: 168907733-168907733 on Build GRCh38
rs1043828434 Chr.2: 168907740-168907740 on Build GRCh38
rs189856873 Chr.2: 168908126-168908126 on Build GRCh38
rs141820044 Chr.2: 168907938-168907938 on Build GRCh38
rs201753573 Chr.2: 168907911-168907911 on Build GRCh38
rs779619992 Chr.2: 168907972-168907972 on Build GRCh38
rs745871593 Chr.2: 168907912-168907912 on Build GRCh38
rs760776009 Chr.2: 168908055-168908055 on Build GRCh38
rs2232328 Chr.2: 168908036-168908036 on Build GRCh38
rs774020743 Chr.2: 168907818-168907818 on Build GRCh38
rs923991987 Chr.2: 168908032-168908032 on Build GRCh38
rs765702651 Chr.2: 168908075-168908075 on Build GRCh38
rs760250729 Chr.2: 168907932-168907932 on Build GRCh38
rs759289035 Chr.2: 168907763-168907763 on Build GRCh38
rs549543851 Chr.2: 168907890-168907890 on Build GRCh38
rs137857125 Chr.2: 168907949-168907949 on Build GRCh38
rs781489778 Chr.2: 168907859-168907859 on Build GRCh38
rs200336133 Chr.2: 168907939-168907939 on Build GRCh38
rs367851926 Chr.2: 168908060-168908060 on Build GRCh38
rs559301659 Chr.2: 168908146-168908146 on Build GRCh38
rs139331128 Chr.2: 168907929-168907929 on Build GRCh38
rs770038801 Chr.2: 168907798-168907798 on Build GRCh38
rs752876983 Chr.2: 168907798-168907808 on Build GRCh38
rs746746462 Chr.2: 168908067-168908067 on Build GRCh38
rs755021707 Chr.2: 168907865-168907865 on Build GRCh38
rs2232325 Chr.2: 168907773-168907773 on Build GRCh38
rs146233425 Chr.2: 168908000-168908000 on Build GRCh38
rs756289993 Chr.2: 168907879-168907879 on Build GRCh38
rs777548181 Chr.2: 168907707-168907707 on Build GRCh38
rs2232327 Chr.2: 168908030-168908030 on Build GRCh38
rs143686780 Chr.2: 168907877-168907877 on Build GRCh38
rs766726786 Chr.2: 168907832-168907832 on Build GRCh38
rs146779637 Chr.2: 168907858-168907858 on Build GRCh38
rs766606769 Chr.2: 168907717-168907717 on Build GRCh38
rs777014914 Chr.2: 168907724-168907724 on Build GRCh38
rs767229217 Chr.2: 168907771-168907771 on Build GRCh38
rs775478622 Chr.2: 168908051-168908051 on Build GRCh38
rs779945911 Chr.2: 168907991-168907997 on Build GRCh38
rs759095465 Chr.2: 168907819-168907819 on Build GRCh38
rs754509276 Chr.2: 168908110-168908110 on Build GRCh38
rs766251315 Chr.2: 168908043-168908043 on Build GRCh38
rs767317043 Chr.2: 168907827-168907827 on Build GRCh38
rs768200400 Chr.2: 168907933-168907933 on Build GRCh38
rs781463613 Chr.2: 168907873-168907873 on Build GRCh38
rs148689354 Chr.2: 168907828-168907828 on Build GRCh38
rs752588930 Chr.2: 168907772-168907772 on Build GRCh38
rs758997713 Chr.2: 168908094-168908094 on Build GRCh38
rs751692549 Chr.2: 168907861-168907861 on Build GRCh38
rs577070120 Chr.2: 168907716-168907716 on Build GRCh38
rs772642710 Chr.2: 168907979-168907979 on Build GRCh38
rs778025780 Chr.2: 168907881-168907881 on Build GRCh38
rs757496063 Chr.2: 168908063-168908063 on Build GRCh38
rs748589053 Chr.2: 168907793-168907793 on Build GRCh38
rs374055555 Chr.2: 168907888-168907888 on Build GRCh38
rs773566412 Chr.2: 168907806-168907806 on Build GRCh38
rs748466993 Chr.2: 168907747-168907747 on Build GRCh38
rs749760008 Chr.2: 168907808-168907808 on Build GRCh38
rs769567058 Chr.2: 168908016-168908016 on Build GRCh38
rs925382715 Chr.2: 168907923-168907923 on Build GRCh38
rs995137594 Chr.2: 168907872-168907872 on Build GRCh38
rs373196602 Chr.2: 168907943-168907943 on Build GRCh38
rs369261389 Chr.2: 168908122-168908122 on Build GRCh38
rs781373483 Chr.2: 168907789-168907789 on Build GRCh38
rs777764289 Chr.2: 168907901-168907906 on Build GRCh38
rs145217135 Chr.2: 168907700-168907700 on Build GRCh38
rs11554658 Chr.2: 168908044-168908044 on Build GRCh38
rs150435873 Chr.2: 168907750-168907750 on Build GRCh38
rs144077223 Chr.2: 168907710-168907710 on Build GRCh38
rs574976550 Chr.2: 168907957-168907957 on Build GRCh38
rs777732226 Chr.2: 168908084-168908084 on Build GRCh38
rs2232329 Chr.2: 168908106-168908106 on Build GRCh38
rs150538801 Chr.2: 168907777-168907777 on Build GRCh38
rs972971686 Chr.2: 168907702-168907702 on Build GRCh38
rs753602775 Chr.2: 168907840-168907840 on Build GRCh38
rs780384187 Chr.2: 168907713-168907713 on Build GRCh38
rs768454510 Chr.2: 168908068-168908069 on Build GRCh38
rs376540601 Chr.2: 168908118-168908118 on Build GRCh38
rs370470317 Chr.2: 168907956-168907956 on Build GRCh38
rs768246460 Chr.2: 168908003-168908003 on Build GRCh38
rs1035992308 Chr.2: 168907892-168907892 on Build GRCh38
rs993287550 Chr.2: 168908046-168908046 on Build GRCh38
rs758253628 Chr.2: 168907979-168907979 on Build GRCh38
rs757689124 Chr.2: 168907971-168907971 on Build GRCh38
rs763810954 Chr.2: 168907835-168907835 on Build GRCh38
rs902511652 Chr.2: 168907737-168907737 on Build GRCh38
rs141041285 Chr.2: 168907913-168907913 on Build GRCh38
rs776320333 Chr.2: 168908005-168908005 on Build GRCh38
rs756209003 Chr.2: 168907854-168907886 on Build GRCh38
rs754193378 Chr.2: 168907960-168907960 on Build GRCh38
rs979502964 Chr.2: 168908014-168908014 on Build GRCh38
rs1032327978 Chr.2: 168908132-168908132 on Build GRCh38
rs771693835 Chr.2: 168907917-168907917 on Build GRCh38
rs779531845 Chr.2: 168907893-168907893 on Build GRCh38
rs761665556 Chr.2: 168908013-168908013 on Build GRCh38
rs139397262 Chr.2: 168907758-168907758 on Build GRCh38
rs762536992 Chr.2: 168907813-168907813 on Build GRCh38
rs762906384 Chr.2: 168908041-168908041 on Build GRCh38
rs2232326 Chr.2: 168907981-168907981 on Build GRCh38
rs756530700 Chr.2: 168908073-168908073 on Build GRCh38

View Full Product Details