Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168909105-168909616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2 XM_011511565.2

SNP Detail

SNP ID Chromosome Location
rs1014132497 Chr.2: 168909587-168909587 on Build GRCh38
rs184995085 Chr.2: 168909481-168909481 on Build GRCh38
rs952700319 Chr.2: 168909348-168909348 on Build GRCh38
rs969699186 Chr.2: 168909330-168909330 on Build GRCh38
rs530990162 Chr.2: 168909357-168909357 on Build GRCh38
rs936818832 Chr.2: 168909368-168909368 on Build GRCh38
rs979788873 Chr.2: 168909355-168909355 on Build GRCh38
rs944539472 Chr.2: 168909580-168909580 on Build GRCh38
rs574508763 Chr.2: 168909505-168909505 on Build GRCh38
rs1053029545 Chr.2: 168909207-168909207 on Build GRCh38
rs191485038 Chr.2: 168909540-168909540 on Build GRCh38
rs992979194 Chr.2: 168909406-168909406 on Build GRCh38
rs568907365 Chr.2: 168909156-168909156 on Build GRCh38
rs541789966 Chr.2: 168909453-168909453 on Build GRCh38
rs915668530 Chr.2: 168909410-168909410 on Build GRCh38
rs899888662 Chr.2: 168909280-168909280 on Build GRCh38
rs1052488334 Chr.2: 168909446-168909446 on Build GRCh38
rs779170310 Chr.2: 168909326-168909326 on Build GRCh38
rs1026589469 Chr.2: 168909233-168909233 on Build GRCh38
rs374184887 Chr.2: 168909561-168909561 on Build GRCh38
rs904188557 Chr.2: 168909169-168909169 on Build GRCh38
rs1053465349 Chr.2: 168909575-168909575 on Build GRCh38
rs899228282 Chr.2: 168909527-168909527 on Build GRCh38
rs567243 Chr.2: 168909227-168909227 on Build GRCh38
rs1017707705 Chr.2: 168909205-168909205 on Build GRCh38
rs983461657 Chr.2: 168909351-168909351 on Build GRCh38
rs545990072 Chr.2: 168909234-168909234 on Build GRCh38
rs573216728 Chr.2: 168909436-168909436 on Build GRCh38
rs188608901 Chr.2: 168909485-168909485 on Build GRCh38
rs948491986 Chr.2: 168909438-168909438 on Build GRCh38
rs960494525 Chr.2: 168909391-168909391 on Build GRCh38
rs149207756 Chr.2: 168909369-168909369 on Build GRCh38
rs192894826 Chr.2: 168909216-168909216 on Build GRCh38
rs996884628 Chr.2: 168909282-168909282 on Build GRCh38
rs2685614 Chr.2: 168909199-168909199 on Build GRCh38
rs188809661 Chr.2: 168909180-168909180 on Build GRCh38
rs914087233 Chr.2: 168909382-168909382 on Build GRCh38
rs1051759943 Chr.2: 168909170-168909170 on Build GRCh38
rs995358783 Chr.2: 168909536-168909536 on Build GRCh38
rs978501469 Chr.2: 168909489-168909489 on Build GRCh38
rs925719521 Chr.2: 168909360-168909360 on Build GRCh38
rs1008708188 Chr.2: 168909217-168909217 on Build GRCh38
rs11554657 Chr.2: 168909264-168909264 on Build GRCh38
rs945642603 Chr.2: 168909413-168909413 on Build GRCh38
rs890497241 Chr.2: 168909171-168909171 on Build GRCh38
rs911784307 Chr.2: 168909579-168909579 on Build GRCh38
rs1026975499 Chr.2: 168909334-168909334 on Build GRCh38
rs890425621 Chr.2: 168909451-168909451 on Build GRCh38
rs766405538 Chr.2: 168909201-168909201 on Build GRCh38

View Full Product Details