Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 170843859-170844382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAD1
Entrez Gene ID
2571
Gene Name
glutamate decarboxylase 1
Gene Aliases
CPSQ1, GAD, SCP
UniGene
Hs.420036
Gene Chromosome Location
Chr.2: 170813210-170861151 on Build GRCh38
Transcripts
NM_000817.2 XM_011510922.1 XM_017003756.1 XM_017003757.1 XM_017003758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3990749 c.675G>T p.M225I Chr.2: 170844081-170844081 on Build GRCh38
6668770 c.640T>C p.F214L Chr.2: 170844046-170844046 on Build GRCh38
7094118 c.746C>A p.S249Y Chr.2: 170844152-170844152 on Build GRCh38
6154766 c.699G>C p.M233I Chr.2: 170844105-170844105 on Build GRCh38
6154765 c.713G>A p.G238E Chr.2: 170844119-170844119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144578957 Chr.2: 170844248-170844249 on Build GRCh38
rs970306734 Chr.2: 170844263-170844263 on Build GRCh38
rs532034120 Chr.2: 170843889-170843889 on Build GRCh38
rs1027210617 Chr.2: 170844260-170844260 on Build GRCh38
rs552207880 Chr.2: 170844291-170844291 on Build GRCh38
rs749832102 Chr.2: 170844177-170844177 on Build GRCh38
rs550563922 Chr.2: 170843917-170843917 on Build GRCh38
rs773925203 Chr.2: 170844164-170844168 on Build GRCh38
rs45601834 Chr.2: 170844219-170844219 on Build GRCh38
rs570691657 Chr.2: 170844307-170844307 on Build GRCh38
rs532573219 Chr.2: 170844187-170844187 on Build GRCh38
rs756971067 Chr.2: 170844054-170844054 on Build GRCh38
rs759157388 Chr.2: 170844203-170844203 on Build GRCh38
rs568985468 Chr.2: 170844036-170844036 on Build GRCh38
rs751171177 Chr.2: 170844022-170844022 on Build GRCh38
rs781413601 Chr.2: 170844145-170844145 on Build GRCh38
rs752666609 Chr.2: 170844338-170844338 on Build GRCh38
rs753494401 Chr.2: 170843886-170843886 on Build GRCh38
rs749782313 Chr.2: 170844192-170844192 on Build GRCh38
rs916373488 Chr.2: 170843964-170843964 on Build GRCh38
rs752243201 Chr.2: 170844051-170844051 on Build GRCh38
rs1054883348 Chr.2: 170843957-170843957 on Build GRCh38
rs960520699 Chr.2: 170844337-170844337 on Build GRCh38
rs753819814 Chr.2: 170844097-170844097 on Build GRCh38
rs774745771 Chr.2: 170844198-170844198 on Build GRCh38
rs548295575 Chr.2: 170844116-170844116 on Build GRCh38
rs972682288 Chr.2: 170844313-170844313 on Build GRCh38
rs773521431 Chr.2: 170844159-170844159 on Build GRCh38
rs201444526 Chr.2: 170844067-170844067 on Build GRCh38
rs369730580 Chr.2: 170844078-170844078 on Build GRCh38
rs767216424 Chr.2: 170844208-170844208 on Build GRCh38
rs1034811383 Chr.2: 170844316-170844316 on Build GRCh38
rs560802377 Chr.2: 170844349-170844350 on Build GRCh38
rs1049886733 Chr.2: 170843980-170843980 on Build GRCh38
rs766767856 Chr.2: 170844019-170844019 on Build GRCh38
rs952157648 Chr.2: 170844247-170844247 on Build GRCh38
rs369505970 Chr.2: 170844156-170844156 on Build GRCh38
rs778876826 Chr.2: 170844111-170844111 on Build GRCh38
rs769174498 Chr.2: 170844023-170844025 on Build GRCh38
rs961559218 Chr.2: 170844304-170844304 on Build GRCh38
rs980058885 Chr.2: 170844282-170844282 on Build GRCh38
rs1043428289 Chr.2: 170843975-170843975 on Build GRCh38
rs946358004 Chr.2: 170843966-170843970 on Build GRCh38
rs939048427 Chr.2: 170843934-170843934 on Build GRCh38
rs996534000 Chr.2: 170843951-170843951 on Build GRCh38
rs756066322 Chr.2: 170844052-170844052 on Build GRCh38
rs755294507 Chr.2: 170844143-170844143 on Build GRCh38
rs756348605 Chr.2: 170844241-170844241 on Build GRCh38
rs879248763 Chr.2: 170843937-170843938 on Build GRCh38
rs890869044 Chr.2: 170844049-170844049 on Build GRCh38
rs756964109 Chr.2: 170844106-170844106 on Build GRCh38
rs45566933 Chr.2: 170844088-170844088 on Build GRCh38
rs996180697 Chr.2: 170844184-170844184 on Build GRCh38

View Full Product Details