Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 170845428-170845939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAD1
Entrez Gene ID
2571
Gene Name
glutamate decarboxylase 1
Gene Aliases
CPSQ1, GAD, SCP
UniGene
Hs.420036
Gene Chromosome Location
Chr.2: 170813210-170861151 on Build GRCh38
Transcripts
NM_000817.2 XM_011510922.1 XM_017003756.1 XM_017003757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
208043 c.757G>A p.A253T Chr.2: 170845511-170845511 on Build GRCh38
5820331 c.815A>G p.K272R Chr.2: 170845569-170845569 on Build GRCh38
4450651 c.752-1G>A p.? Chr.2: 170845505-170845505 on Build GRCh38
5517945 c.867+6T>A p.? Chr.2: 170845627-170845627 on Build GRCh38
1401228 c.832G>A p.A278T Chr.2: 170845586-170845586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749735425 Chr.2: 170845509-170845509 on Build GRCh38
rs369785577 Chr.2: 170845630-170845630 on Build GRCh38
rs774940037 Chr.2: 170845768-170845768 on Build GRCh38
rs763492866 Chr.2: 170845535-170845535 on Build GRCh38
rs755325669 Chr.2: 170845823-170845823 on Build GRCh38
rs750313621 Chr.2: 170845488-170845488 on Build GRCh38
rs758897677 Chr.2: 170845806-170845806 on Build GRCh38
rs747831626 Chr.2: 170845722-170845722 on Build GRCh38
rs776247095 Chr.2: 170845543-170845543 on Build GRCh38
rs755233233 Chr.2: 170845493-170845493 on Build GRCh38
rs766305351 Chr.2: 170845567-170845567 on Build GRCh38
rs780279882 Chr.2: 170845687-170845687 on Build GRCh38
rs757270484 Chr.2: 170845480-170845480 on Build GRCh38
rs769377346 Chr.2: 170845728-170845728 on Build GRCh38
rs761345985 Chr.2: 170845615-170845615 on Build GRCh38
rs927451899 Chr.2: 170845453-170845453 on Build GRCh38
rs761656256 Chr.2: 170845544-170845544 on Build GRCh38
rs576560343 Chr.2: 170845603-170845603 on Build GRCh38
rs757287794 Chr.2: 170845650-170845650 on Build GRCh38
rs769428753 Chr.2: 170845629-170845629 on Build GRCh38
rs763003381 Chr.2: 170845631-170845631 on Build GRCh38
rs765319290 Chr.2: 170845558-170845558 on Build GRCh38
rs758947117 Chr.2: 170845683-170845683 on Build GRCh38
rs1052237925 Chr.2: 170845807-170845807 on Build GRCh38
rs753672528 Chr.2: 170845472-170845472 on Build GRCh38
rs763692590 Chr.2: 170845461-170845461 on Build GRCh38
rs768919266 Chr.2: 170845702-170845702 on Build GRCh38
rs188144170 Chr.2: 170845798-170845798 on Build GRCh38
rs756104212 Chr.2: 170845500-170845500 on Build GRCh38
rs767248551 Chr.2: 170845785-170845785 on Build GRCh38
rs201601105 Chr.2: 170845655-170845656 on Build GRCh38
rs752812272 Chr.2: 170845569-170845569 on Build GRCh38
rs1029108102 Chr.2: 170845851-170845851 on Build GRCh38
rs764434464 Chr.2: 170845644-170845644 on Build GRCh38
rs34291719 Chr.2: 170845484-170845485 on Build GRCh38
rs938708639 Chr.2: 170845640-170845640 on Build GRCh38
rs770967698 Chr.2: 170845749-170845749 on Build GRCh38
rs777683337 Chr.2: 170845572-170845572 on Build GRCh38
rs775137917 Chr.2: 170845458-170845458 on Build GRCh38
rs762950875 Chr.2: 170845734-170845734 on Build GRCh38
rs867050800 Chr.2: 170845507-170845507 on Build GRCh38
rs775995716 Chr.2: 170845611-170845611 on Build GRCh38
rs750572051 Chr.2: 170845802-170845802 on Build GRCh38
rs754014995 Chr.2: 170845573-170845573 on Build GRCh38
rs774967412 Chr.2: 170845528-170845528 on Build GRCh38
rs376291126 Chr.2: 170845670-170845670 on Build GRCh38
rs904560859 Chr.2: 170845693-170845693 on Build GRCh38
rs759779673 Chr.2: 170845694-170845697 on Build GRCh38
rs759973203 Chr.2: 170845856-170845856 on Build GRCh38
rs777929911 Chr.2: 170845501-170845501 on Build GRCh38
rs562343816 Chr.2: 170845754-170845754 on Build GRCh38
rs956461027 Chr.2: 170845525-170845525 on Build GRCh38
rs765098259 Chr.2: 170845486-170845486 on Build GRCh38
rs780519382 Chr.2: 170845604-170845604 on Build GRCh38
rs765272565 Chr.2: 170845800-170845800 on Build GRCh38
rs374632004 Chr.2: 170845649-170845649 on Build GRCh38
rs766254022 Chr.2: 170845637-170845637 on Build GRCh38
rs892303802 Chr.2: 170845825-170845825 on Build GRCh38
rs746406697 Chr.2: 170845585-170845585 on Build GRCh38
rs781429378 Chr.2: 170845828-170845828 on Build GRCh38
rs751408796 Chr.2: 170845639-170845639 on Build GRCh38
rs760803337 Chr.2: 170845643-170845643 on Build GRCh38
rs75821413 Chr.2: 170845665-170845665 on Build GRCh38
rs772431542 Chr.2: 170845516-170845516 on Build GRCh38
rs766855756 Chr.2: 170845814-170845814 on Build GRCh38
rs757342630 Chr.2: 170845576-170845576 on Build GRCh38
rs17857148 Chr.2: 170845710-170845710 on Build GRCh38
rs1019452513 Chr.2: 170845897-170845897 on Build GRCh38
rs760398831 Chr.2: 170845529-170845529 on Build GRCh38
rs539657238 Chr.2: 170845510-170845510 on Build GRCh38
rs747778379 Chr.2: 170845829-170845829 on Build GRCh38
rs771470806 Chr.2: 170845504-170845505 on Build GRCh38
rs953508636 Chr.2: 170845873-170845873 on Build GRCh38
rs768426080 Chr.2: 170845606-170845606 on Build GRCh38
rs148633602 Chr.2: 170845565-170845565 on Build GRCh38
rs541810963 Chr.2: 170845790-170845790 on Build GRCh38
rs370456144 Chr.2: 170845720-170845720 on Build GRCh38
rs772896216 Chr.2: 170845733-170845733 on Build GRCh38
rs574505468 Chr.2: 170845769-170845769 on Build GRCh38
rs370906252 Chr.2: 170845691-170845691 on Build GRCh38
rs34100397 Chr.2: 170845561-170845561 on Build GRCh38
rs750908978 Chr.2: 170845672-170845672 on Build GRCh38
rs763126962 Chr.2: 170845562-170845562 on Build GRCh38
rs781571382 Chr.2: 170845903-170845903 on Build GRCh38
rs143319979 Chr.2: 170845584-170845584 on Build GRCh38
rs189949060 Chr.2: 170845890-170845890 on Build GRCh38
rs888889205 Chr.2: 170845913-170845913 on Build GRCh38
rs368994782 Chr.2: 170845467-170845467 on Build GRCh38
rs919438057 Chr.2: 170845636-170845636 on Build GRCh38
rs545577676 Chr.2: 170845848-170845848 on Build GRCh38
rs760039773 Chr.2: 170845459-170845459 on Build GRCh38
rs377405060 Chr.2: 170845498-170845498 on Build GRCh38
rs527332017 Chr.2: 170845819-170845819 on Build GRCh38
rs1057317397 Chr.2: 170845589-170845589 on Build GRCh38
rs373115750 Chr.2: 170845495-170845495 on Build GRCh38
rs746613477 Chr.2: 170845605-170845605 on Build GRCh38
rs756406233 Chr.2: 170845571-170845571 on Build GRCh38

View Full Product Details