Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171443232-171443738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF17
Entrez Gene ID
80067
Gene Name
DDB1 and CUL4 associated factor 17
Gene Aliases
C2orf37
UniGene
Hs.659439
Gene Chromosome Location
Chr.2: 171434166-171491029 on Build GRCh38
Transcripts
NM_001164821.1 NM_025000.3 NR_028482.1 XM_006712767.1 XM_006712768.1 XM_011511881.1 XM_011511882.1 XM_017004995.1 XM_017004996.1 XM_017004997.1 XM_017004998.1 XM_017004999.1 XM_017005000.1 XM_017005001.1 XM_017005002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5983259 c.240T>A p.S80S Chr.2: 171443532-171443532 on Build GRCh38
6640479 c.283_284insA p.I97fs*22 Chr.2: 171443575-171443576 on Build GRCh38
441499 c.265C>G p.P89A Chr.2: 171443557-171443557 on Build GRCh38
1494643 c.277A>C p.K93Q Chr.2: 171443569-171443569 on Build GRCh38
6379925 c.244C>G p.P82A Chr.2: 171443536-171443536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949029675 Chr.2: 171443344-171443344 on Build GRCh38
rs781164465 Chr.2: 171443619-171443619 on Build GRCh38
rs746564010 Chr.2: 171443596-171443596 on Build GRCh38
rs758012127 Chr.2: 171443586-171443586 on Build GRCh38
rs1052861109 Chr.2: 171443363-171443363 on Build GRCh38
rs983855241 Chr.2: 171443621-171443621 on Build GRCh38
rs759763006 Chr.2: 171443634-171443634 on Build GRCh38
rs906890077 Chr.2: 171443431-171443431 on Build GRCh38
rs748647626 Chr.2: 171443490-171443491 on Build GRCh38
rs892006438 Chr.2: 171443452-171443452 on Build GRCh38
rs879253799 Chr.2: 171443562-171443562 on Build GRCh38
rs993310968 Chr.2: 171443712-171443712 on Build GRCh38
rs1038571948 Chr.2: 171443345-171443345 on Build GRCh38
rs948421544 Chr.2: 171443366-171443366 on Build GRCh38
rs796246351 Chr.2: 171443350-171443350 on Build GRCh38
rs898255939 Chr.2: 171443348-171443348 on Build GRCh38
rs574062680 Chr.2: 171443673-171443673 on Build GRCh38
rs774566117 Chr.2: 171443515-171443515 on Build GRCh38
rs768613603 Chr.2: 171443482-171443482 on Build GRCh38
rs863224865 Chr.2: 171443581-171443582 on Build GRCh38
rs1026272174 Chr.2: 171443696-171443696 on Build GRCh38
rs369859810 Chr.2: 171443661-171443661 on Build GRCh38
rs964785568 Chr.2: 171443501-171443501 on Build GRCh38
rs754236793 Chr.2: 171443550-171443550 on Build GRCh38
rs889604735 Chr.2: 171443558-171443558 on Build GRCh38
rs148401399 Chr.2: 171443532-171443532 on Build GRCh38
rs77284169 Chr.2: 171443674-171443674 on Build GRCh38
rs779579578 Chr.2: 171443595-171443595 on Build GRCh38
rs763076918 Chr.2: 171443415-171443415 on Build GRCh38
rs933306155 Chr.2: 171443272-171443272 on Build GRCh38
rs756655038 Chr.2: 171443544-171443544 on Build GRCh38
rs960185811 Chr.2: 171443697-171443697 on Build GRCh38
rs1049615647 Chr.2: 171443449-171443449 on Build GRCh38
rs60392491 Chr.2: 171443349-171443350 on Build GRCh38
rs554684015 Chr.2: 171443255-171443255 on Build GRCh38
rs757760166 Chr.2: 171443565-171443565 on Build GRCh38
rs1031954686 Chr.2: 171443435-171443435 on Build GRCh38
rs202131890 Chr.2: 171443633-171443633 on Build GRCh38
rs776380751 Chr.2: 171443489-171443489 on Build GRCh38
rs563816056 Chr.2: 171443368-171443368 on Build GRCh38
rs772220298 Chr.2: 171443656-171443656 on Build GRCh38
rs183914118 Chr.2: 171443356-171443356 on Build GRCh38
rs774136261 Chr.2: 171443383-171443383 on Build GRCh38
rs1005486739 Chr.2: 171443632-171443632 on Build GRCh38
rs1002937703 Chr.2: 171443433-171443433 on Build GRCh38
rs766497476 Chr.2: 171443497-171443497 on Build GRCh38
rs779152661 Chr.2: 171443568-171443568 on Build GRCh38
rs199808149 Chr.2: 171443283-171443283 on Build GRCh38
rs1043681417 Chr.2: 171443347-171443348 on Build GRCh38
rs1011757144 Chr.2: 171443454-171443454 on Build GRCh38
rs764744666 Chr.2: 171443548-171443548 on Build GRCh38
rs759690510 Chr.2: 171443516-171443516 on Build GRCh38
rs574838124 Chr.2: 171443348-171443348 on Build GRCh38
rs1030361656 Chr.2: 171443522-171443522 on Build GRCh38
rs768558594 Chr.2: 171443598-171443598 on Build GRCh38
rs768442815 Chr.2: 171443256-171443256 on Build GRCh38
rs761646871 Chr.2: 171443494-171443494 on Build GRCh38
rs192321411 Chr.2: 171443335-171443335 on Build GRCh38
rs750038271 Chr.2: 171443583-171443583 on Build GRCh38
rs76137360 Chr.2: 171443292-171443292 on Build GRCh38
rs752749215 Chr.2: 171443543-171443543 on Build GRCh38
rs1021721968 Chr.2: 171443462-171443462 on Build GRCh38
rs1044101422 Chr.2: 171443418-171443418 on Build GRCh38

View Full Product Details