Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171449525-171450064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF17
Entrez Gene ID
80067
Gene Name
DDB1 and CUL4 associated factor 17
Gene Aliases
C2orf37
UniGene
Hs.659439
Gene Chromosome Location
Chr.2: 171434166-171491029 on Build GRCh38
Transcripts
NM_001164821.1 NM_025000.3 NR_028482.1 XM_006712767.1 XM_006712768.1 XM_011511881.1 XM_011511882.1 XM_017004995.1 XM_017004996.1 XM_017004997.1 XM_017004998.1 XM_017004999.1 XM_017005000.1 XM_017005001.1 XM_017005002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4551013 c.515G>C p.R172T Chr.2: 171449935-171449935 on Build GRCh38
7363727 c.533G>A p.R178Q Chr.2: 171449953-171449953 on Build GRCh38
3569987 c.532C>T p.R178W Chr.2: 171449952-171449952 on Build GRCh38
4154598 c.523G>C p.A175P Chr.2: 171449943-171449943 on Build GRCh38
7287078 c.537+3A>G p.? Chr.2: 171449960-171449960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550953804 Chr.2: 171449927-171449927 on Build GRCh38
rs773914315 Chr.2: 171449822-171449822 on Build GRCh38
rs973851184 Chr.2: 171449700-171449700 on Build GRCh38
rs879255422 Chr.2: 171449929-171449929 on Build GRCh38
rs763660064 Chr.2: 171449830-171449830 on Build GRCh38
rs760384569 Chr.2: 171449963-171449963 on Build GRCh38
rs747625728 Chr.2: 171449963-171449976 on Build GRCh38
rs750231739 Chr.2: 171450000-171450000 on Build GRCh38
rs777550757 Chr.2: 171449957-171449957 on Build GRCh38
rs745701144 Chr.2: 171449847-171449847 on Build GRCh38
rs986651760 Chr.2: 171449645-171449645 on Build GRCh38
rs928893194 Chr.2: 171449874-171449874 on Build GRCh38
rs113215140 Chr.2: 171449866-171449866 on Build GRCh38
rs867176926 Chr.2: 171449659-171449659 on Build GRCh38
rs786205638 Chr.2: 171449872-171449919 on Build GRCh38
rs774674106 Chr.2: 171449933-171449933 on Build GRCh38
rs778370006 Chr.2: 171449986-171449988 on Build GRCh38
rs377364475 Chr.2: 171449952-171449952 on Build GRCh38
rs546878583 Chr.2: 171449860-171449862 on Build GRCh38
rs781622815 Chr.2: 171449863-171449863 on Build GRCh38
rs757047969 Chr.2: 171449870-171449870 on Build GRCh38
rs569143715 Chr.2: 171449683-171449683 on Build GRCh38
rs55987027 Chr.2: 171449689-171449689 on Build GRCh38
rs1023181187 Chr.2: 171449570-171449570 on Build GRCh38
rs566237798 Chr.2: 171449736-171449736 on Build GRCh38
rs770432686 Chr.2: 171449974-171449975 on Build GRCh38
rs922440745 Chr.2: 171449754-171449754 on Build GRCh38
rs200433540 Chr.2: 171449964-171449964 on Build GRCh38
rs140913375 Chr.2: 171449709-171449709 on Build GRCh38
rs183473333 Chr.2: 171449737-171449737 on Build GRCh38
rs759948524 Chr.2: 171449950-171449950 on Build GRCh38
rs761814031 Chr.2: 171449991-171449991 on Build GRCh38
rs202231211 Chr.2: 171449953-171449953 on Build GRCh38
rs1014471863 Chr.2: 171450024-171450024 on Build GRCh38
rs755232804 Chr.2: 171449850-171449850 on Build GRCh38
rs748257732 Chr.2: 171449871-171449871 on Build GRCh38
rs753423974 Chr.2: 171449978-171449978 on Build GRCh38
rs1028669334 Chr.2: 171449633-171449633 on Build GRCh38
rs988555115 Chr.2: 171449730-171449730 on Build GRCh38
rs1038130493 Chr.2: 171449898-171449898 on Build GRCh38
rs780590311 Chr.2: 171449874-171449874 on Build GRCh38
rs778815572 Chr.2: 171449845-171449845 on Build GRCh38
rs757286307 Chr.2: 171449840-171449840 on Build GRCh38
rs373466341 Chr.2: 171449932-171449932 on Build GRCh38
rs182481451 Chr.2: 171449699-171449699 on Build GRCh38
rs933792638 Chr.2: 171449787-171449787 on Build GRCh38
rs17220440 Chr.2: 171449638-171449638 on Build GRCh38
rs979757217 Chr.2: 171449849-171449849 on Build GRCh38
rs552031635 Chr.2: 171449916-171449916 on Build GRCh38
rs537583175 Chr.2: 171450026-171450026 on Build GRCh38
rs935361609 Chr.2: 171449987-171449987 on Build GRCh38
rs1015461863 Chr.2: 171449661-171449661 on Build GRCh38
rs1014520876 Chr.2: 171449566-171449566 on Build GRCh38
rs778769146 Chr.2: 171449986-171449986 on Build GRCh38
rs753657865 Chr.2: 171449835-171449835 on Build GRCh38
rs773362482 Chr.2: 171449924-171449924 on Build GRCh38
rs533581220 Chr.2: 171449807-171449807 on Build GRCh38
rs948392932 Chr.2: 171449828-171449828 on Build GRCh38
rs947555057 Chr.2: 171450022-171450022 on Build GRCh38
rs765317190 Chr.2: 171449995-171449995 on Build GRCh38
rs919680844 Chr.2: 171449777-171449777 on Build GRCh38
rs142462938 Chr.2: 171449973-171449973 on Build GRCh38

View Full Product Details