Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171458152-171458657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF17
Entrez Gene ID
80067
Gene Name
DDB1 and CUL4 associated factor 17
Gene Aliases
C2orf37
UniGene
Hs.659439
Gene Chromosome Location
Chr.2: 171434166-171491029 on Build GRCh38
Transcripts
NM_001164821.1 NM_025000.3 NR_028482.1 XM_006712767.1 XM_006712768.1 XM_011511881.1 XM_011511882.1 XM_017004995.1 XM_017004996.1 XM_017004997.1 XM_017004998.1 XM_017004999.1 XM_017005000.1 XM_017005001.1 XM_017005002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249734 c.818C>T p.P273L Chr.2: 171458457-171458457 on Build GRCh38
5917589 c.817C>A p.P273T Chr.2: 171458456-171458456 on Build GRCh38
2893272 c.751G>C p.D251H Chr.2: 171458390-171458390 on Build GRCh38
1494642 c.733-2A>T p.? Chr.2: 171458370-171458370 on Build GRCh38
4624465 c.736A>T p.M246L Chr.2: 171458375-171458375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897980063 Chr.2: 171458559-171458559 on Build GRCh38
rs199742600 Chr.2: 171458431-171458431 on Build GRCh38
rs745776305 Chr.2: 171458342-171458342 on Build GRCh38
rs745422590 Chr.2: 171458459-171458459 on Build GRCh38
rs140025362 Chr.2: 171458595-171458595 on Build GRCh38
rs746956061 Chr.2: 171458488-171458488 on Build GRCh38
rs770003540 Chr.2: 171458370-171458370 on Build GRCh38
rs900105100 Chr.2: 171458294-171458294 on Build GRCh38
rs768194770 Chr.2: 171458403-171458403 on Build GRCh38
rs958798022 Chr.2: 171458252-171458252 on Build GRCh38
rs763098025 Chr.2: 171458381-171458381 on Build GRCh38
rs753197965 Chr.2: 171458517-171458517 on Build GRCh38
rs1049093420 Chr.2: 171458599-171458599 on Build GRCh38
rs753477159 Chr.2: 171458425-171458425 on Build GRCh38
rs549506154 Chr.2: 171458261-171458261 on Build GRCh38
rs774474650 Chr.2: 171458507-171458507 on Build GRCh38
rs948047652 Chr.2: 171458246-171458246 on Build GRCh38
rs947476741 Chr.2: 171458240-171458240 on Build GRCh38
rs16859390 Chr.2: 171458603-171458603 on Build GRCh38
rs992649307 Chr.2: 171458262-171458262 on Build GRCh38
rs369164414 Chr.2: 171458344-171458344 on Build GRCh38
rs374359784 Chr.2: 171458491-171458491 on Build GRCh38
rs547306608 Chr.2: 171458414-171458414 on Build GRCh38
rs766434466 Chr.2: 171458382-171458382 on Build GRCh38
rs771100235 Chr.2: 171458503-171458503 on Build GRCh38
rs764509776 Chr.2: 171458523-171458523 on Build GRCh38
rs753106369 Chr.2: 171458406-171458406 on Build GRCh38
rs767709440 Chr.2: 171458513-171458513 on Build GRCh38
rs535322644 Chr.2: 171458285-171458285 on Build GRCh38
rs754191203 Chr.2: 171458526-171458526 on Build GRCh38
rs773360559 Chr.2: 171458374-171458374 on Build GRCh38
rs947429852 Chr.2: 171458539-171458539 on Build GRCh38
rs749277004 Chr.2: 171458238-171458238 on Build GRCh38
rs557969150 Chr.2: 171458568-171458568 on Build GRCh38
rs759755757 Chr.2: 171458508-171458508 on Build GRCh38
rs879240573 Chr.2: 171458241-171458241 on Build GRCh38
rs903257915 Chr.2: 171458259-171458259 on Build GRCh38
rs1015369709 Chr.2: 171458405-171458405 on Build GRCh38
rs775246401 Chr.2: 171458366-171458366 on Build GRCh38
rs575798043 Chr.2: 171458251-171458254 on Build GRCh38
rs925649812 Chr.2: 171458335-171458335 on Build GRCh38
rs751624341 Chr.2: 171458394-171458394 on Build GRCh38
rs952319928 Chr.2: 171458365-171458365 on Build GRCh38
rs760524776 Chr.2: 171458369-171458369 on Build GRCh38
rs773752168 Chr.2: 171458325-171458325 on Build GRCh38
rs760059288 Chr.2: 171458398-171458398 on Build GRCh38
rs185405017 Chr.2: 171458229-171458229 on Build GRCh38
rs768382294 Chr.2: 171458489-171458489 on Build GRCh38
rs372481846 Chr.2: 171458409-171458409 on Build GRCh38
rs778451530 Chr.2: 171458445-171458445 on Build GRCh38
rs981983769 Chr.2: 171458420-171458420 on Build GRCh38
rs1002130216 Chr.2: 171458624-171458624 on Build GRCh38
rs370438709 Chr.2: 171458465-171458465 on Build GRCh38
rs191001101 Chr.2: 171458265-171458265 on Build GRCh38
rs780036993 Chr.2: 171458473-171458473 on Build GRCh38
rs376182176 Chr.2: 171458493-171458493 on Build GRCh38
rs967184525 Chr.2: 171458286-171458286 on Build GRCh38
rs539237511 Chr.2: 171458555-171458555 on Build GRCh38
rs775577850 Chr.2: 171458350-171458354 on Build GRCh38

View Full Product Details