Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171687380-171687877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I2
Entrez Gene ID
1781
Gene Name
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases
DIC74, DNCI2, IC2
UniGene
Hs.546250
Gene Chromosome Location
Chr.2: 171687409-171750158 on Build GRCh38
Transcripts
NM_001271785.1 NM_001271786.1 NM_001271787.1 NM_001271788.1 NM_001271789.1 NM_001271790.1 NM_001320882.1 NM_001320883.1 NM_001320884.1 NM_001378.2 XM_006712347.3 XM_017003525.1

SNP Detail

SNP ID Chromosome Location
rs931042416 Chr.2: 171687823-171687823 on Build GRCh38
rs896876237 Chr.2: 171687644-171687644 on Build GRCh38
rs747024919 Chr.2: 171687607-171687607 on Build GRCh38
rs1048203464 Chr.2: 171687832-171687832 on Build GRCh38
rs1020965363 Chr.2: 171687420-171687420 on Build GRCh38
rs777232459 Chr.2: 171687776-171687776 on Build GRCh38
rs757966929 Chr.2: 171687543-171687543 on Build GRCh38
rs553582930 Chr.2: 171687786-171687786 on Build GRCh38
rs779775704 Chr.2: 171687611-171687611 on Build GRCh38
rs905730434 Chr.2: 171687708-171687708 on Build GRCh38
rs533120527 Chr.2: 171687749-171687749 on Build GRCh38
rs979517209 Chr.2: 171687764-171687764 on Build GRCh38
rs936342964 Chr.2: 171687789-171687789 on Build GRCh38
rs918295573 Chr.2: 171687792-171687792 on Build GRCh38
rs545506706 Chr.2: 171687400-171687400 on Build GRCh38
rs966760801 Chr.2: 171687761-171687761 on Build GRCh38
rs954377547 Chr.2: 171687766-171687766 on Build GRCh38
rs932908310 Chr.2: 171687555-171687555 on Build GRCh38
rs6721680 Chr.2: 171687403-171687403 on Build GRCh38
rs748006529 Chr.2: 171687790-171687790 on Build GRCh38
rs751190812 Chr.2: 171687723-171687723 on Build GRCh38
rs1013970744 Chr.2: 171687758-171687758 on Build GRCh38
rs754247926 Chr.2: 171687440-171687440 on Build GRCh38
rs930088124 Chr.2: 171687693-171687693 on Build GRCh38
rs1029680314 Chr.2: 171687550-171687550 on Build GRCh38
rs1048255578 Chr.2: 171687684-171687684 on Build GRCh38
rs765741993 Chr.2: 171687534-171687534 on Build GRCh38
rs769519815 Chr.2: 171687818-171687818 on Build GRCh38
rs1030108084 Chr.2: 171687831-171687831 on Build GRCh38
rs754672277 Chr.2: 171687773-171687773 on Build GRCh38
rs997276286 Chr.2: 171687827-171687827 on Build GRCh38
rs892265474 Chr.2: 171687532-171687532 on Build GRCh38
rs1035992160 Chr.2: 171687748-171687748 on Build GRCh38
rs931053790 Chr.2: 171687671-171687671 on Build GRCh38
rs912213332 Chr.2: 171687581-171687581 on Build GRCh38
rs541293833 Chr.2: 171687655-171687655 on Build GRCh38
rs192066085 Chr.2: 171687622-171687622 on Build GRCh38
rs985666949 Chr.2: 171687771-171687771 on Build GRCh38
rs551909227 Chr.2: 171687765-171687765 on Build GRCh38
rs943680737 Chr.2: 171687591-171687591 on Build GRCh38
rs1020913324 Chr.2: 171687760-171687760 on Build GRCh38
rs966773765 Chr.2: 171687432-171687432 on Build GRCh38
rs141792734 Chr.2: 171687559-171687559 on Build GRCh38
rs374950408 Chr.2: 171687734-171687734 on Build GRCh38
rs963072437 Chr.2: 171687609-171687609 on Build GRCh38
rs561710233 Chr.2: 171687580-171687580 on Build GRCh38
rs944313477 Chr.2: 171687785-171687785 on Build GRCh38
rs747991518 Chr.2: 171687567-171687567 on Build GRCh38
rs974341536 Chr.2: 171687632-171687632 on Build GRCh38
rs144938072 Chr.2: 171687500-171687500 on Build GRCh38

View Full Product Details