Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171726052-171726550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I2
Entrez Gene ID
1781
Gene Name
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases
DIC74, DNCI2, IC2
UniGene
Hs.546250
Gene Chromosome Location
Chr.2: 171687409-171750158 on Build GRCh38
Transcripts
NM_001271785.1 NM_001271786.1 NM_001271787.1 NM_001271788.1 NM_001271789.1 NM_001271790.1 NM_001320882.1 NM_001320883.1 NM_001320884.1 NM_001378.2 XM_006712347.3 XM_017003525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5883290 c.754C>T p.Q252* Chr.2: 171726201-171726201 on Build GRCh38
6337670 c.781C>T p.R261* Chr.2: 171726228-171726228 on Build GRCh38
6996466 c.800G>A p.R267H Chr.2: 171726247-171726247 on Build GRCh38
6337669 c.805C>T p.R269* Chr.2: 171726228-171726228 on Build GRCh38
5883289 c.778C>T p.Q260* Chr.2: 171726201-171726201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749194010 Chr.2: 171726294-171726294 on Build GRCh38
rs954225563 Chr.2: 171726471-171726471 on Build GRCh38
rs757349085 Chr.2: 171726198-171726198 on Build GRCh38
rs149580450 Chr.2: 171726324-171726324 on Build GRCh38
rs751933536 Chr.2: 171726325-171726325 on Build GRCh38
rs780891314 Chr.2: 171726246-171726246 on Build GRCh38
rs373783701 Chr.2: 171726299-171726299 on Build GRCh38
rs537702121 Chr.2: 171726131-171726131 on Build GRCh38
rs746625267 Chr.2: 171726151-171726151 on Build GRCh38
rs748033252 Chr.2: 171726296-171726296 on Build GRCh38
rs752995751 Chr.2: 171726176-171726176 on Build GRCh38
rs574586191 Chr.2: 171726234-171726234 on Build GRCh38
rs371436419 Chr.2: 171726247-171726247 on Build GRCh38
rs757640523 Chr.2: 171726108-171726108 on Build GRCh38
rs756299361 Chr.2: 171726091-171726091 on Build GRCh38
rs780933857 Chr.2: 171726271-171726273 on Build GRCh38
rs766851656 Chr.2: 171726437-171726437 on Build GRCh38
rs776893809 Chr.2: 171726314-171726314 on Build GRCh38
rs985554110 Chr.2: 171726477-171726477 on Build GRCh38
rs112612557 Chr.2: 171726375-171726375 on Build GRCh38
rs765384526 Chr.2: 171726183-171726183 on Build GRCh38
rs768128306 Chr.2: 171726243-171726243 on Build GRCh38
rs1047672191 Chr.2: 171726236-171726236 on Build GRCh38
rs777671224 Chr.2: 171726092-171726092 on Build GRCh38
rs370761161 Chr.2: 171726128-171726128 on Build GRCh38
rs370187651 Chr.2: 171726316-171726316 on Build GRCh38
rs769686099 Chr.2: 171726170-171726170 on Build GRCh38
rs765557168 Chr.2: 171726368-171726368 on Build GRCh38
rs764598311 Chr.2: 171726462-171726466 on Build GRCh38
rs755860143 Chr.2: 171726339-171726341 on Build GRCh38
rs1032325057 Chr.2: 171726443-171726443 on Build GRCh38
rs757451212 Chr.2: 171726169-171726170 on Build GRCh38
rs564811523 Chr.2: 171726468-171726468 on Build GRCh38
rs755355906 Chr.2: 171726327-171726327 on Build GRCh38
rs750491632 Chr.2: 171726192-171726193 on Build GRCh38
rs775948802 Chr.2: 171726162-171726162 on Build GRCh38
rs895743961 Chr.2: 171726429-171726429 on Build GRCh38
rs750902846 Chr.2: 171726208-171726208 on Build GRCh38
rs144273879 Chr.2: 171726378-171726378 on Build GRCh38
rs138226344 Chr.2: 171726319-171726319 on Build GRCh38
rs754949522 Chr.2: 171726250-171726250 on Build GRCh38
rs778255495 Chr.2: 171726395-171726395 on Build GRCh38
rs779722228 Chr.2: 171726146-171726146 on Build GRCh38
rs910306765 Chr.2: 171726138-171726138 on Build GRCh38
rs1047595987 Chr.2: 171726425-171726425 on Build GRCh38
rs774227906 Chr.2: 171726304-171726304 on Build GRCh38
rs912603005 Chr.2: 171726486-171726486 on Build GRCh38
rs558103422 Chr.2: 171726093-171726093 on Build GRCh38
rs373010290 Chr.2: 171726215-171726215 on Build GRCh38
rs773267424 Chr.2: 171726174-171726174 on Build GRCh38
rs201480046 Chr.2: 171726190-171726190 on Build GRCh38
rs925287988 Chr.2: 171726262-171726262 on Build GRCh38
rs367747207 Chr.2: 171726315-171726315 on Build GRCh38
rs1021256977 Chr.2: 171726347-171726347 on Build GRCh38
rs1032464421 Chr.2: 171726506-171726506 on Build GRCh38
rs769325251 Chr.2: 171726250-171726250 on Build GRCh38
rs560594412 Chr.2: 171726305-171726305 on Build GRCh38
rs376565643 Chr.2: 171726441-171726441 on Build GRCh38
rs377058205 Chr.2: 171726242-171726242 on Build GRCh38
rs768237714 Chr.2: 171726161-171726161 on Build GRCh38
rs1020148429 Chr.2: 171726418-171726418 on Build GRCh38
rs746293983 Chr.2: 171726114-171726114 on Build GRCh38
rs566253747 Chr.2: 171726513-171726513 on Build GRCh38
rs532037346 Chr.2: 171726331-171726331 on Build GRCh38
rs893796453 Chr.2: 171726527-171726527 on Build GRCh38
rs932739237 Chr.2: 171726270-171726270 on Build GRCh38
rs764244661 Chr.2: 171726188-171726188 on Build GRCh38
rs779340708 Chr.2: 171726201-171726201 on Build GRCh38
rs62184167 Chr.2: 171726109-171726109 on Build GRCh38
rs895576606 Chr.2: 171726383-171726383 on Build GRCh38
rs754107906 Chr.2: 171726164-171726165 on Build GRCh38
rs755041582 Chr.2: 171726130-171726135 on Build GRCh38
rs929011654 Chr.2: 171726412-171726412 on Build GRCh38
rs750951727 Chr.2: 171726317-171726317 on Build GRCh38
rs761813083 Chr.2: 171726163-171726163 on Build GRCh38
rs189123828 Chr.2: 171726391-171726391 on Build GRCh38
rs780755896 Chr.2: 171726328-171726328 on Build GRCh38
rs776906142 Chr.2: 171726306-171726306 on Build GRCh38
rs987646770 Chr.2: 171726376-171726376 on Build GRCh38
rs1035883773 Chr.2: 171726382-171726382 on Build GRCh38
rs1020181059 Chr.2: 171726218-171726218 on Build GRCh38
rs1002688591 Chr.2: 171726480-171726480 on Build GRCh38
rs773318375 Chr.2: 171726261-171726261 on Build GRCh38

View Full Product Details