Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 172087925-172088430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX1
Entrez Gene ID
1745
Gene Name
distal-less homeobox 1
Gene Aliases
-
UniGene
Hs.407015
Gene Chromosome Location
Chr.2: 172085480-172089674 on Build GRCh38
Transcripts
NM_001038493.1 NM_178120.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3570014 c.683G>A p.G228E Chr.2: 172088172-172088172 on Build GRCh38
6333676 c.592G>T p.A198S Chr.2: 172088081-172088081 on Build GRCh38
3570015 c.696C>T p.S232S Chr.2: 172088185-172088185 on Build GRCh38
6088357 c.574G>C p.A192P Chr.2: 172088063-172088063 on Build GRCh38
6154742 c.739G>C p.E247Q Chr.2: 172088228-172088228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747883157 Chr.2: 172088127-172088127 on Build GRCh38
rs752389190 Chr.2: 172088158-172088158 on Build GRCh38
rs777348613 Chr.2: 172088243-172088243 on Build GRCh38
rs199592485 Chr.2: 172088141-172088141 on Build GRCh38
rs765790329 Chr.2: 172088294-172088294 on Build GRCh38
rs201352974 Chr.2: 172087964-172087964 on Build GRCh38
rs775357521 Chr.2: 172088279-172088279 on Build GRCh38
rs773487262 Chr.2: 172088294-172088294 on Build GRCh38
rs760394646 Chr.2: 172088046-172088047 on Build GRCh38
rs867218613 Chr.2: 172088151-172088151 on Build GRCh38
rs780278640 Chr.2: 172088291-172088291 on Build GRCh38
rs758527146 Chr.2: 172088191-172088191 on Build GRCh38
rs748395831 Chr.2: 172088231-172088231 on Build GRCh38
rs953905385 Chr.2: 172088347-172088347 on Build GRCh38
rs960341904 Chr.2: 172088172-172088172 on Build GRCh38
rs979350490 Chr.2: 172088306-172088306 on Build GRCh38
rs750525757 Chr.2: 172088189-172088189 on Build GRCh38
rs898067787 Chr.2: 172088081-172088081 on Build GRCh38
rs766492924 Chr.2: 172088300-172088300 on Build GRCh38
rs995032862 Chr.2: 172088370-172088370 on Build GRCh38
rs374088146 Chr.2: 172088155-172088155 on Build GRCh38
rs768152552 Chr.2: 172088048-172088049 on Build GRCh38
rs779074889 Chr.2: 172088088-172088088 on Build GRCh38
rs762123407 Chr.2: 172088170-172088170 on Build GRCh38
rs562265867 Chr.2: 172087996-172087996 on Build GRCh38
rs755806324 Chr.2: 172088233-172088233 on Build GRCh38
rs910649773 Chr.2: 172088391-172088391 on Build GRCh38
rs531211683 Chr.2: 172088369-172088369 on Build GRCh38
rs747497909 Chr.2: 172088117-172088117 on Build GRCh38
rs769293085 Chr.2: 172088128-172088128 on Build GRCh38
rs376171844 Chr.2: 172088050-172088050 on Build GRCh38
rs759519725 Chr.2: 172088150-172088150 on Build GRCh38
rs368867772 Chr.2: 172088089-172088089 on Build GRCh38
rs1040620446 Chr.2: 172088064-172088064 on Build GRCh38
rs748734476 Chr.2: 172088298-172088298 on Build GRCh38
rs377685238 Chr.2: 172088197-172088197 on Build GRCh38
rs765307778 Chr.2: 172088185-172088185 on Build GRCh38
rs200336628 Chr.2: 172088122-172088122 on Build GRCh38
rs112033071 Chr.2: 172087978-172087978 on Build GRCh38
rs1024356787 Chr.2: 172088402-172088402 on Build GRCh38
rs748944355 Chr.2: 172088245-172088245 on Build GRCh38
rs754133482 Chr.2: 172088068-172088068 on Build GRCh38
rs775328313 Chr.2: 172087980-172087981 on Build GRCh38
rs986596825 Chr.2: 172088380-172088380 on Build GRCh38
rs772572784 Chr.2: 172087982-172087982 on Build GRCh38
rs1033439943 Chr.2: 172088166-172088166 on Build GRCh38
rs751588540 Chr.2: 172088305-172088305 on Build GRCh38
rs991770652 Chr.2: 172088188-172088188 on Build GRCh38
rs776207917 Chr.2: 172088271-172088271 on Build GRCh38
rs927180879 Chr.2: 172088156-172088156 on Build GRCh38
rs762494940 Chr.2: 172088140-172088140 on Build GRCh38
rs755452975 Chr.2: 172088203-172088203 on Build GRCh38
rs758559781 Chr.2: 172088101-172088101 on Build GRCh38
rs201733130 Chr.2: 172088262-172088262 on Build GRCh38
rs769384702 Chr.2: 172087971-172087971 on Build GRCh38
rs759462574 Chr.2: 172088027-172088027 on Build GRCh38
rs376188162 Chr.2: 172087963-172087963 on Build GRCh38
rs751413970 Chr.2: 172088003-172088003 on Build GRCh38
rs1027881660 Chr.2: 172088384-172088384 on Build GRCh38
rs771866816 Chr.2: 172088270-172088270 on Build GRCh38
rs370539046 Chr.2: 172088308-172088308 on Build GRCh38
rs760326361 Chr.2: 172088289-172088289 on Build GRCh38
rs761169927 Chr.2: 172088275-172088277 on Build GRCh38
rs776403160 Chr.2: 172087955-172087955 on Build GRCh38
rs900699285 Chr.2: 172088330-172088330 on Build GRCh38
rs773980605 Chr.2: 172088266-172088266 on Build GRCh38
rs925073235 Chr.2: 172088326-172088326 on Build GRCh38
rs138502787 Chr.2: 172088078-172088078 on Build GRCh38
rs758908128 Chr.2: 172088097-172088097 on Build GRCh38
rs73976550 Chr.2: 172088131-172088131 on Build GRCh38
rs950692693 Chr.2: 172088389-172088389 on Build GRCh38
rs774501591 Chr.2: 172088144-172088144 on Build GRCh38
rs750521498 Chr.2: 172087991-172087991 on Build GRCh38
rs770608566 Chr.2: 172088248-172088248 on Build GRCh38
rs752100178 Chr.2: 172088196-172088196 on Build GRCh38
rs866378513 Chr.2: 172088133-172088133 on Build GRCh38
rs574239254 Chr.2: 172088011-172088011 on Build GRCh38
rs781635559 Chr.2: 172088221-172088221 on Build GRCh38
rs745825806 Chr.2: 172088269-172088269 on Build GRCh38
rs767472300 Chr.2: 172088153-172088153 on Build GRCh38
rs541299088 Chr.2: 172088092-172088092 on Build GRCh38

View Full Product Details