Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173364715-173365223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA7
Entrez Gene ID
83879
Gene Name
cell division cycle associated 7
Gene Aliases
ICF3, JPO1
UniGene
Hs.470654
Gene Chromosome Location
Chr.2: 173354017-173368992 on Build GRCh38
Transcripts
NM_031942.4 NM_145810.2 XM_011511957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4086824 c.715C>T p.R239* Chr.2: 173364810-173364810 on Build GRCh38
5860624 c.825_827delGGA p.E281delE Chr.2: 173364920-173364922 on Build GRCh38
4086825 c.850T>C p.Y284H Chr.2: 173364945-173364945 on Build GRCh38
6427615 c.838G>T p.E280* Chr.2: 173364933-173364933 on Build GRCh38
3570095 c.719G>A p.R240K Chr.2: 173364814-173364814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029961208 Chr.2: 173364919-173364919 on Build GRCh38
rs535908062 Chr.2: 173364922-173364922 on Build GRCh38
rs145479884 Chr.2: 173364855-173364855 on Build GRCh38
rs201391282 Chr.2: 173364908-173364908 on Build GRCh38
rs766427795 Chr.2: 173364881-173364881 on Build GRCh38
rs931388886 Chr.2: 173365145-173365145 on Build GRCh38
rs772539390 Chr.2: 173364778-173364778 on Build GRCh38
rs757432537 Chr.2: 173364919-173364920 on Build GRCh38
rs781543958 Chr.2: 173365017-173365017 on Build GRCh38
rs756436135 Chr.2: 173364750-173364751 on Build GRCh38
rs534928780 Chr.2: 173364817-173364817 on Build GRCh38
rs373671271 Chr.2: 173365190-173365191 on Build GRCh38
rs13418393 Chr.2: 173364743-173364743 on Build GRCh38
rs777875556 Chr.2: 173364818-173364818 on Build GRCh38
rs148404814 Chr.2: 173364857-173364857 on Build GRCh38
rs540705403 Chr.2: 173364827-173364827 on Build GRCh38
rs754798292 Chr.2: 173365053-173365053 on Build GRCh38
rs147411652 Chr.2: 173364899-173364899 on Build GRCh38
rs777100041 Chr.2: 173364940-173364940 on Build GRCh38
rs769558563 Chr.2: 173364964-173364964 on Build GRCh38
rs914480908 Chr.2: 173364859-173364859 on Build GRCh38
rs765456341 Chr.2: 173365001-173365001 on Build GRCh38
rs780128145 Chr.2: 173364811-173364811 on Build GRCh38
rs953939520 Chr.2: 173365026-173365026 on Build GRCh38
rs200556185 Chr.2: 173364856-173364856 on Build GRCh38
rs772146602 Chr.2: 173365067-173365067 on Build GRCh38
rs769997632 Chr.2: 173365025-173365025 on Build GRCh38
rs371303570 Chr.2: 173364782-173364782 on Build GRCh38
rs147594902 Chr.2: 173364864-173364864 on Build GRCh38
rs556663937 Chr.2: 173364897-173364897 on Build GRCh38
rs751469184 Chr.2: 173364812-173364812 on Build GRCh38
rs767491623 Chr.2: 173365036-173365036 on Build GRCh38
rs751487627 Chr.2: 173364883-173364883 on Build GRCh38
rs772080685 Chr.2: 173364909-173364909 on Build GRCh38
rs776323018 Chr.2: 173364779-173364779 on Build GRCh38
rs755565561 Chr.2: 173365013-173365013 on Build GRCh38
rs764760592 Chr.2: 173364803-173364803 on Build GRCh38
rs770680049 Chr.2: 173365030-173365030 on Build GRCh38
rs758456042 Chr.2: 173364810-173364810 on Build GRCh38
rs369071982 Chr.2: 173365000-173365000 on Build GRCh38
rs950446756 Chr.2: 173364942-173364942 on Build GRCh38
rs752598179 Chr.2: 173364997-173364997 on Build GRCh38
rs747471184 Chr.2: 173364916-173364916 on Build GRCh38
rs895361730 Chr.2: 173364944-173364944 on Build GRCh38
rs140418017 Chr.2: 173364896-173364896 on Build GRCh38
rs13382180 Chr.2: 173365003-173365003 on Build GRCh38
rs770948684 Chr.2: 173364826-173364826 on Build GRCh38
rs748552199 Chr.2: 173365023-173365023 on Build GRCh38
rs774057496 Chr.2: 173365031-173365031 on Build GRCh38
rs371635516 Chr.2: 173365131-173365131 on Build GRCh38
rs774465144 Chr.2: 173364975-173364975 on Build GRCh38
rs374816306 Chr.2: 173364792-173364792 on Build GRCh38
rs749477773 Chr.2: 173364824-173364824 on Build GRCh38
rs764345710 Chr.2: 173364885-173364886 on Build GRCh38
rs549258132 Chr.2: 173364744-173364744 on Build GRCh38
rs369310070 Chr.2: 173364758-173364758 on Build GRCh38
rs775230836 Chr.2: 173365041-173365041 on Build GRCh38
rs544697691 Chr.2: 173365098-173365098 on Build GRCh38
rs965595664 Chr.2: 173364915-173364915 on Build GRCh38
rs373468351 Chr.2: 173364888-173364888 on Build GRCh38
rs1000128877 Chr.2: 173364985-173364985 on Build GRCh38
rs779058449 Chr.2: 173364929-173364930 on Build GRCh38
rs893880946 Chr.2: 173365068-173365068 on Build GRCh38
rs55847722 Chr.2: 173365088-173365088 on Build GRCh38
rs750009083 Chr.2: 173364873-173364873 on Build GRCh38
rs148421045 Chr.2: 173364971-173364971 on Build GRCh38
rs1012387692 Chr.2: 173364808-173364808 on Build GRCh38
rs758309071 Chr.2: 173364918-173364918 on Build GRCh38
rs935349664 Chr.2: 173365040-173365040 on Build GRCh38
rs762550271 Chr.2: 173364875-173364875 on Build GRCh38
rs761247476 Chr.2: 173364797-173364797 on Build GRCh38
rs753472878 Chr.2: 173364789-173364789 on Build GRCh38
rs146148251 Chr.2: 173364840-173364840 on Build GRCh38
rs765939337 Chr.2: 173364972-173364972 on Build GRCh38
rs368968744 Chr.2: 173364889-173364889 on Build GRCh38
rs748445001 Chr.2: 173364945-173364945 on Build GRCh38
rs538328863 Chr.2: 173364860-173364860 on Build GRCh38
rs749865667 Chr.2: 173364809-173364809 on Build GRCh38
rs376689912 Chr.2: 173364982-173364982 on Build GRCh38
rs567492256 Chr.2: 173364777-173364777 on Build GRCh38
rs749574601 Chr.2: 173365132-173365132 on Build GRCh38
rs943883656 Chr.2: 173365080-173365080 on Build GRCh38
rs758958300 Chr.2: 173365033-173365033 on Build GRCh38
rs918645308 Chr.2: 173365093-173365093 on Build GRCh38
rs908943345 Chr.2: 173364953-173364953 on Build GRCh38
rs780146971 Chr.2: 173365005-173365005 on Build GRCh38
rs770986071 Chr.2: 173364749-173364749 on Build GRCh38
rs1006964770 Chr.2: 173365076-173365076 on Build GRCh38
rs563556904 Chr.2: 173365187-173365188 on Build GRCh38
rs138353896 Chr.2: 173364976-173364976 on Build GRCh38
rs747068213 Chr.2: 173365010-173365010 on Build GRCh38
rs754843372 Chr.2: 173364884-173364884 on Build GRCh38
rs761021685 Chr.2: 173364781-173364781 on Build GRCh38
rs79281910 Chr.2: 173365028-173365028 on Build GRCh38
rs764771856 Chr.2: 173364865-173364865 on Build GRCh38
rs916458005 Chr.2: 173364989-173364989 on Build GRCh38
rs754083976 Chr.2: 173364920-173364922 on Build GRCh38

View Full Product Details