Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174846738-174847246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHN1
Entrez Gene ID
1123
Gene Name
chimerin 1
Gene Aliases
ARHGAP2, CHN, DURS2, NC, RHOGAP2
UniGene
Hs.380138
Gene Chromosome Location
Chr.2: 174799314-175005379 on Build GRCh38
Transcripts
NM_001025201.3 NM_001206602.1 NM_001822.5 NR_038133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3694992 c.82G>T p.D28Y Chr.2: 174847050-174847050 on Build GRCh38
3414327 c.213C>T p.N71N Chr.2: 174846919-174846919 on Build GRCh38
290781 c.570A>T p.R190S Chr.2: 174846937-174846937 on Build GRCh38
3693877 c.108G>T p.M36I Chr.2: 174847024-174847024 on Build GRCh38
2894174 c.223C>T p.P75S Chr.2: 174846909-174846909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756474859 Chr.2: 174847151-174847151 on Build GRCh38
rs141304713 Chr.2: 174847111-174847111 on Build GRCh38
rs1049626009 Chr.2: 174847046-174847046 on Build GRCh38
rs182176073 Chr.2: 174846794-174846794 on Build GRCh38
rs748355372 Chr.2: 174847170-174847170 on Build GRCh38
rs927474391 Chr.2: 174847166-174847166 on Build GRCh38
rs773684607 Chr.2: 174846918-174846918 on Build GRCh38
rs930794325 Chr.2: 174847004-174847004 on Build GRCh38
rs574926915 Chr.2: 174846845-174846845 on Build GRCh38
rs1000358054 Chr.2: 174846775-174846775 on Build GRCh38
rs765565238 Chr.2: 174846828-174846828 on Build GRCh38
rs905990563 Chr.2: 174846827-174846827 on Build GRCh38
rs951274318 Chr.2: 174847093-174847093 on Build GRCh38
rs543364007 Chr.2: 174847146-174847146 on Build GRCh38
rs1014166345 Chr.2: 174847123-174847123 on Build GRCh38
rs533914410 Chr.2: 174846763-174846763 on Build GRCh38
rs985815640 Chr.2: 174846800-174846800 on Build GRCh38
rs190390221 Chr.2: 174847097-174847097 on Build GRCh38
rs751564472 Chr.2: 174847101-174847101 on Build GRCh38
rs764454922 Chr.2: 174846970-174846971 on Build GRCh38
rs148302567 Chr.2: 174847109-174847109 on Build GRCh38
rs972339540 Chr.2: 174846961-174846961 on Build GRCh38
rs992784178 Chr.2: 174846826-174846826 on Build GRCh38
rs911545993 Chr.2: 174846842-174846842 on Build GRCh38
rs946256450 Chr.2: 174847210-174847210 on Build GRCh38
rs1020357813 Chr.2: 174846957-174846957 on Build GRCh38
rs770197850 Chr.2: 174847189-174847189 on Build GRCh38
rs1037910546 Chr.2: 174846802-174846802 on Build GRCh38
rs907979866 Chr.2: 174846784-174846784 on Build GRCh38
rs778071898 Chr.2: 174847155-174847155 on Build GRCh38
rs750860565 Chr.2: 174846879-174846879 on Build GRCh38
rs781458396 Chr.2: 174847145-174847145 on Build GRCh38
rs14228 Chr.2: 174847214-174847214 on Build GRCh38
rs186717784 Chr.2: 174846893-174846893 on Build GRCh38
rs147990577 Chr.2: 174846878-174846878 on Build GRCh38
rs1000008152 Chr.2: 174847182-174847182 on Build GRCh38
rs570781043 Chr.2: 174846791-174846791 on Build GRCh38
rs770505558 Chr.2: 174846902-174846902 on Build GRCh38
rs919421851 Chr.2: 174846965-174846965 on Build GRCh38
rs562060369 Chr.2: 174847193-174847193 on Build GRCh38
rs747720875 Chr.2: 174847039-174847039 on Build GRCh38
rs11556877 Chr.2: 174846947-174846947 on Build GRCh38
rs763040490 Chr.2: 174847081-174847081 on Build GRCh38
rs11556874 Chr.2: 174846979-174846979 on Build GRCh38
rs776409235 Chr.2: 174846983-174846983 on Build GRCh38
rs775489561 Chr.2: 174846935-174846935 on Build GRCh38
rs189918995 Chr.2: 174846786-174846786 on Build GRCh38
rs770239851 Chr.2: 174847078-174847078 on Build GRCh38
rs760529847 Chr.2: 174846962-174846962 on Build GRCh38
rs917517574 Chr.2: 174847203-174847203 on Build GRCh38
rs953135629 Chr.2: 174847194-174847194 on Build GRCh38
rs572102406 Chr.2: 174847057-174847057 on Build GRCh38
rs938079645 Chr.2: 174847103-174847103 on Build GRCh38
rs373949870 Chr.2: 174846995-174846995 on Build GRCh38
rs572206494 Chr.2: 174847003-174847003 on Build GRCh38
rs768036014 Chr.2: 174846848-174846848 on Build GRCh38
rs146313212 Chr.2: 174846919-174846919 on Build GRCh38
rs1032730516 Chr.2: 174846779-174846779 on Build GRCh38
rs370550062 Chr.2: 174846928-174846928 on Build GRCh38
rs556676655 Chr.2: 174846844-174846844 on Build GRCh38

View Full Product Details