Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175072538-175073048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF2
Entrez Gene ID
1386
Gene Name
activating transcription factor 2
Gene Aliases
CRE-BP1, CREB-2, CREB2, HB16, TREB7
UniGene
Hs.592510
Gene Chromosome Location
Chr.2: 175072250-175168206 on Build GRCh38
Transcripts
NM_001256090.1 NM_001256091.1 NM_001256092.1 NM_001256093.1 NM_001880.3 NR_045768.1 NR_045769.1 NR_045770.1 NR_045771.1 NR_045772.1 NR_045773.1 NR_045774.1

SNP Detail

SNP ID Chromosome Location
rs778495459 Chr.2: 175072685-175072685 on Build GRCh38
rs956958975 Chr.2: 175072580-175072580 on Build GRCh38
rs976165560 Chr.2: 175072694-175072694 on Build GRCh38
rs910191097 Chr.2: 175072659-175072660 on Build GRCh38
rs141685111 Chr.2: 175072666-175072666 on Build GRCh38
rs948559743 Chr.2: 175073011-175073011 on Build GRCh38
rs975736471 Chr.2: 175072978-175072978 on Build GRCh38
rs535514500 Chr.2: 175072863-175072863 on Build GRCh38
rs913414819 Chr.2: 175072725-175072725 on Build GRCh38
rs186084026 Chr.2: 175072789-175072789 on Build GRCh38
rs931665148 Chr.2: 175072801-175072801 on Build GRCh38
rs980357134 Chr.2: 175073019-175073019 on Build GRCh38
rs966196924 Chr.2: 175072976-175072977 on Build GRCh38
rs558415961 Chr.2: 175073016-175073016 on Build GRCh38
rs984737090 Chr.2: 175072999-175072999 on Build GRCh38
rs909080494 Chr.2: 175072769-175072772 on Build GRCh38
rs1022404290 Chr.2: 175072903-175072903 on Build GRCh38
rs909029948 Chr.2: 175073004-175073004 on Build GRCh38
rs774545646 Chr.2: 175072920-175072920 on Build GRCh38
rs1027604749 Chr.2: 175072980-175072980 on Build GRCh38
rs1045180144 Chr.2: 175072775-175072775 on Build GRCh38
rs1022458017 Chr.2: 175072603-175072603 on Build GRCh38
rs116222057 Chr.2: 175072767-175072767 on Build GRCh38
rs1034423834 Chr.2: 175072576-175072576 on Build GRCh38
rs929211025 Chr.2: 175072717-175072717 on Build GRCh38
rs145241336 Chr.2: 175072890-175072890 on Build GRCh38
rs926687380 Chr.2: 175073024-175073024 on Build GRCh38
rs35507277 Chr.2: 175072934-175072934 on Build GRCh38
rs1009650603 Chr.2: 175072600-175072600 on Build GRCh38
rs939339381 Chr.2: 175072787-175072787 on Build GRCh38
rs967763471 Chr.2: 175072615-175072615 on Build GRCh38
rs567454853 Chr.2: 175072790-175072790 on Build GRCh38
rs537367448 Chr.2: 175072770-175072770 on Build GRCh38
rs944450346 Chr.2: 175072895-175072895 on Build GRCh38
rs1026025509 Chr.2: 175072979-175072979 on Build GRCh38
rs985924834 Chr.2: 175072620-175072620 on Build GRCh38
rs1050153769 Chr.2: 175072859-175072859 on Build GRCh38
rs904053342 Chr.2: 175072909-175072909 on Build GRCh38
rs995158084 Chr.2: 175072969-175072969 on Build GRCh38
rs548534976 Chr.2: 175072923-175072923 on Build GRCh38
rs572106494 Chr.2: 175072926-175072926 on Build GRCh38
rs777112428 Chr.2: 175072956-175072956 on Build GRCh38
rs551678938 Chr.2: 175072738-175072738 on Build GRCh38
rs1009806941 Chr.2: 175072866-175072866 on Build GRCh38
rs919142896 Chr.2: 175072695-175072695 on Build GRCh38
rs749851574 Chr.2: 175072614-175072614 on Build GRCh38
rs941414768 Chr.2: 175072773-175072773 on Build GRCh38

View Full Product Details