Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175073266-175073796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF2
Entrez Gene ID
1386
Gene Name
activating transcription factor 2
Gene Aliases
CRE-BP1, CREB-2, CREB2, HB16, TREB7
UniGene
Hs.592510
Gene Chromosome Location
Chr.2: 175072250-175168206 on Build GRCh38
Transcripts
NM_001256090.1 NM_001256091.1 NM_001256092.1 NM_001256093.1 NM_001880.3 NR_045768.1 NR_045769.1 NR_045770.1 NR_045771.1 NR_045772.1 NR_045773.1 NR_045774.1

SNP Detail

SNP ID Chromosome Location
rs997250587 Chr.2: 175073498-175073499 on Build GRCh38
rs970389598 Chr.2: 175073453-175073453 on Build GRCh38
rs751769950 Chr.2: 175073389-175073389 on Build GRCh38
rs903238798 Chr.2: 175073731-175073731 on Build GRCh38
rs767209691 Chr.2: 175073538-175073538 on Build GRCh38
rs1035507890 Chr.2: 175073550-175073550 on Build GRCh38
rs1014286417 Chr.2: 175073656-175073656 on Build GRCh38
rs1002091114 Chr.2: 175073330-175073330 on Build GRCh38
rs539612642 Chr.2: 175073766-175073768 on Build GRCh38
rs901590776 Chr.2: 175073314-175073314 on Build GRCh38
rs972208975 Chr.2: 175073510-175073510 on Build GRCh38
rs965772380 Chr.2: 175073504-175073504 on Build GRCh38
rs1048854192 Chr.2: 175073620-175073620 on Build GRCh38
rs183206457 Chr.2: 175073484-175073484 on Build GRCh38
rs879304201 Chr.2: 175073458-175073459 on Build GRCh38
rs886204164 Chr.2: 175073519-175073519 on Build GRCh38
rs369213804 Chr.2: 175073527-175073527 on Build GRCh38
rs191180251 Chr.2: 175073619-175073619 on Build GRCh38
rs532941648 Chr.2: 175073299-175073299 on Build GRCh38
rs943621501 Chr.2: 175073696-175073696 on Build GRCh38
rs1001885038 Chr.2: 175073543-175073543 on Build GRCh38
rs1025972219 Chr.2: 175073401-175073401 on Build GRCh38
rs988617486 Chr.2: 175073573-175073573 on Build GRCh38
rs925310393 Chr.2: 175073683-175073683 on Build GRCh38
rs1049434852 Chr.2: 175073303-175073303 on Build GRCh38
rs565873129 Chr.2: 175073628-175073628 on Build GRCh38
rs895824321 Chr.2: 175073405-175073405 on Build GRCh38
rs530116480 Chr.2: 175073499-175073499 on Build GRCh38
rs950389893 Chr.2: 175073411-175073411 on Build GRCh38
rs563314616 Chr.2: 175073471-175073471 on Build GRCh38
rs1014752101 Chr.2: 175073406-175073406 on Build GRCh38
rs913953751 Chr.2: 175073584-175073584 on Build GRCh38
rs539563388 Chr.2: 175073723-175073723 on Build GRCh38
rs190992180 Chr.2: 175073482-175073482 on Build GRCh38
rs1016162114 Chr.2: 175073437-175073437 on Build GRCh38
rs879684528 Chr.2: 175073335-175073335 on Build GRCh38
rs372950176 Chr.2: 175073640-175073640 on Build GRCh38
rs1039195111 Chr.2: 175073496-175073499 on Build GRCh38
rs895797598 Chr.2: 175073654-175073654 on Build GRCh38
rs943658104 Chr.2: 175073485-175073485 on Build GRCh38
rs960119072 Chr.2: 175073560-175073560 on Build GRCh38
rs775716914 Chr.2: 175073737-175073737 on Build GRCh38
rs889325923 Chr.2: 175073769-175073769 on Build GRCh38
rs754154186 Chr.2: 175073489-175073492 on Build GRCh38
rs969991232 Chr.2: 175073541-175073541 on Build GRCh38
rs186067337 Chr.2: 175073496-175073496 on Build GRCh38
rs368554187 Chr.2: 175073390-175073390 on Build GRCh38
rs1042129538 Chr.2: 175073700-175073700 on Build GRCh38
rs967932343 Chr.2: 175073469-175073469 on Build GRCh38
rs937891633 Chr.2: 175073630-175073630 on Build GRCh38
rs1020402151 Chr.2: 175073462-175073462 on Build GRCh38
rs1005911279 Chr.2: 175073415-175073415 on Build GRCh38
rs780109292 Chr.2: 175073300-175073300 on Build GRCh38
rs755981157 Chr.2: 175073611-175073611 on Build GRCh38
rs1015975618 Chr.2: 175073530-175073530 on Build GRCh38
rs930409468 Chr.2: 175073597-175073597 on Build GRCh38
rs914005869 Chr.2: 175073473-175073474 on Build GRCh38
rs186596625 Chr.2: 175073472-175073472 on Build GRCh38
rs544986561 Chr.2: 175073392-175073392 on Build GRCh38
rs866146125 Chr.2: 175073740-175073740 on Build GRCh38
rs966767259 Chr.2: 175073608-175073608 on Build GRCh38
rs765503202 Chr.2: 175073498-175073501 on Build GRCh38
rs550040904 Chr.2: 175073475-175073475 on Build GRCh38
rs979955591 Chr.2: 175073459-175073460 on Build GRCh38
rs557838414 Chr.2: 175073744-175073744 on Build GRCh38
rs752575881 Chr.2: 175073554-175073554 on Build GRCh38
rs928491196 Chr.2: 175073467-175073467 on Build GRCh38
rs766568091 Chr.2: 175073321-175073321 on Build GRCh38

View Full Product Details