Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176080516-176081015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVX2
Entrez Gene ID
344191
Gene Name
even-skipped homeobox 2
Gene Aliases
EVX-2
UniGene
Hs.452341
Gene Chromosome Location
Chr.2: 176080107-176083962 on Build GRCh38
Transcripts
NM_001080458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4136580 c.797C>A p.T266K Chr.2: 176080741-176080741 on Build GRCh38
6155489 c.864G>A p.P288P Chr.2: 176080674-176080674 on Build GRCh38
1009824 c.766G>T p.A256S Chr.2: 176080772-176080772 on Build GRCh38
4956559 c.937G>A p.A313T Chr.2: 176080601-176080601 on Build GRCh38
7121381 c.723G>A p.M241I Chr.2: 176080815-176080815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761946844 Chr.2: 176080782-176080782 on Build GRCh38
rs988561235 Chr.2: 176080786-176080786 on Build GRCh38
rs377296091 Chr.2: 176080862-176080862 on Build GRCh38
rs754466176 Chr.2: 176080570-176080570 on Build GRCh38
rs775548500 Chr.2: 176080624-176080624 on Build GRCh38
rs374162005 Chr.2: 176080827-176080827 on Build GRCh38
rs746551317 Chr.2: 176080544-176080544 on Build GRCh38
rs992176471 Chr.2: 176080907-176080907 on Build GRCh38
rs749153969 Chr.2: 176080716-176080716 on Build GRCh38
rs111993318 Chr.2: 176080661-176080661 on Build GRCh38
rs760847501 Chr.2: 176080625-176080625 on Build GRCh38
rs777517967 Chr.2: 176080700-176080700 on Build GRCh38
rs563733909 Chr.2: 176080788-176080788 on Build GRCh38
rs146229022 Chr.2: 176080867-176080867 on Build GRCh38
rs776936495 Chr.2: 176080647-176080648 on Build GRCh38
rs548473434 Chr.2: 176080963-176080963 on Build GRCh38
rs777802417 Chr.2: 176080856-176080856 on Build GRCh38
rs369040390 Chr.2: 176080744-176080744 on Build GRCh38
rs73029923 Chr.2: 176080939-176080939 on Build GRCh38
rs759202546 Chr.2: 176080836-176080836 on Build GRCh38
rs769735456 Chr.2: 176080882-176080882 on Build GRCh38
rs567088329 Chr.2: 176080979-176080979 on Build GRCh38
rs777332878 Chr.2: 176080636-176080636 on Build GRCh38
rs993175574 Chr.2: 176080749-176080749 on Build GRCh38
rs1007453298 Chr.2: 176080695-176080695 on Build GRCh38
rs373960757 Chr.2: 176080689-176080689 on Build GRCh38
rs766108943 Chr.2: 176080670-176080670 on Build GRCh38
rs915222621 Chr.2: 176080850-176080850 on Build GRCh38
rs768810964 Chr.2: 176080621-176080653 on Build GRCh38
rs200844347 Chr.2: 176080575-176080575 on Build GRCh38
rs763179583 Chr.2: 176080665-176080665 on Build GRCh38
rs755595195 Chr.2: 176080854-176080854 on Build GRCh38
rs747369034 Chr.2: 176080614-176080640 on Build GRCh38
rs777500581 Chr.2: 176080919-176080919 on Build GRCh38
rs779533275 Chr.2: 176080542-176080542 on Build GRCh38
rs772992433 Chr.2: 176080605-176080605 on Build GRCh38
rs144229189 Chr.2: 176080690-176080690 on Build GRCh38
rs772357531 Chr.2: 176080617-176080617 on Build GRCh38
rs747219076 Chr.2: 176080734-176080734 on Build GRCh38
rs868442087 Chr.2: 176080710-176080710 on Build GRCh38
rs770599533 Chr.2: 176080719-176080719 on Build GRCh38
rs763418729 Chr.2: 176080799-176080799 on Build GRCh38
rs1026491399 Chr.2: 176080752-176080752 on Build GRCh38
rs752224496 Chr.2: 176080852-176080852 on Build GRCh38
rs549373633 Chr.2: 176080924-176080924 on Build GRCh38
rs200426346 Chr.2: 176080691-176080691 on Build GRCh38
rs529005443 Chr.2: 176080943-176080943 on Build GRCh38
rs751286050 Chr.2: 176080812-176080812 on Build GRCh38
rs764264424 Chr.2: 176080628-176080628 on Build GRCh38
rs1040169370 Chr.2: 176080921-176080921 on Build GRCh38
rs541628257 Chr.2: 176080535-176080535 on Build GRCh38
rs745590834 Chr.2: 176080877-176080877 on Build GRCh38
rs769638449 Chr.2: 176080602-176080602 on Build GRCh38
rs867299349 Chr.2: 176080619-176080619 on Build GRCh38
rs369068866 Chr.2: 176080677-176080677 on Build GRCh38
rs780390509 Chr.2: 176080724-176080724 on Build GRCh38
rs766652738 Chr.2: 176080810-176080810 on Build GRCh38
rs778583823 Chr.2: 176080871-176080871 on Build GRCh38
rs984273502 Chr.2: 176080822-176080822 on Build GRCh38
rs376810083 Chr.2: 176080692-176080692 on Build GRCh38
rs944563133 Chr.2: 176080916-176080916 on Build GRCh38
rs767114072 Chr.2: 176080843-176080843 on Build GRCh38
rs748994507 Chr.2: 176080608-176080608 on Build GRCh38
rs769032157 Chr.2: 176080881-176080881 on Build GRCh38
rs765774536 Chr.2: 176080651-176080651 on Build GRCh38
rs1018848633 Chr.2: 176080742-176080742 on Build GRCh38
rs750831697 Chr.2: 176080653-176080653 on Build GRCh38
rs776856614 Chr.2: 176080780-176080780 on Build GRCh38
rs770446959 Chr.2: 176080784-176080784 on Build GRCh38

View Full Product Details