Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176083263-176083761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVX2
Entrez Gene ID
344191
Gene Name
even-skipped homeobox 2
Gene Aliases
EVX-2
UniGene
Hs.452341
Gene Chromosome Location
Chr.2: 176080107-176083962 on Build GRCh38
Transcripts
NM_001080458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
717664 c.181G>A p.A61T Chr.2: 176083596-176083596 on Build GRCh38
4086911 c.43G>T p.G15W Chr.2: 176083734-176083734 on Build GRCh38
3379866 c.58A>G p.T20A Chr.2: 176083719-176083719 on Build GRCh38
4086909 c.100A>G p.N34D Chr.2: 176083677-176083677 on Build GRCh38
6155486 c.261C>A p.T87T Chr.2: 176083516-176083516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775740903 Chr.2: 176083624-176083624 on Build GRCh38
rs780971594 Chr.2: 176083541-176083541 on Build GRCh38
rs757814311 Chr.2: 176083514-176083514 on Build GRCh38
rs541323509 Chr.2: 176083705-176083705 on Build GRCh38
rs771211020 Chr.2: 176083551-176083551 on Build GRCh38
rs753138757 Chr.2: 176083434-176083434 on Build GRCh38
rs759607919 Chr.2: 176083604-176083605 on Build GRCh38
rs775266360 Chr.2: 176083449-176083449 on Build GRCh38
rs140560927 Chr.2: 176083471-176083471 on Build GRCh38
rs151290628 Chr.2: 176083436-176083436 on Build GRCh38
rs533201192 Chr.2: 176083723-176083723 on Build GRCh38
rs751006473 Chr.2: 176083607-176083607 on Build GRCh38
rs750742006 Chr.2: 176083641-176083641 on Build GRCh38
rs765414616 Chr.2: 176083495-176083496 on Build GRCh38
rs774418360 Chr.2: 176083552-176083552 on Build GRCh38
rs1012774356 Chr.2: 176083335-176083335 on Build GRCh38
rs562116152 Chr.2: 176083591-176083591 on Build GRCh38
rs61731351 Chr.2: 176083717-176083717 on Build GRCh38
rs899534511 Chr.2: 176083329-176083329 on Build GRCh38
rs777245036 Chr.2: 176083635-176083635 on Build GRCh38
rs781151618 Chr.2: 176083654-176083654 on Build GRCh38
rs1037679297 Chr.2: 176083581-176083581 on Build GRCh38
rs367856760 Chr.2: 176083463-176083463 on Build GRCh38
rs756963433 Chr.2: 176083502-176083502 on Build GRCh38
rs199567327 Chr.2: 176083453-176083453 on Build GRCh38
rs748616817 Chr.2: 176083700-176083700 on Build GRCh38
rs752105450 Chr.2: 176083343-176083343 on Build GRCh38
rs765491751 Chr.2: 176083397-176083397 on Build GRCh38
rs764979508 Chr.2: 176083597-176083597 on Build GRCh38
rs769448659 Chr.2: 176083616-176083616 on Build GRCh38
rs200378788 Chr.2: 176083396-176083396 on Build GRCh38
rs939182040 Chr.2: 176083668-176083668 on Build GRCh38
rs750041037 Chr.2: 176083604-176083604 on Build GRCh38
rs747124965 Chr.2: 176083687-176083687 on Build GRCh38
rs748245928 Chr.2: 176083559-176083559 on Build GRCh38
rs565703140 Chr.2: 176083311-176083311 on Build GRCh38
rs768525510 Chr.2: 176083532-176083532 on Build GRCh38
rs181478435 Chr.2: 176083413-176083413 on Build GRCh38
rs773371574 Chr.2: 176083496-176083497 on Build GRCh38
rs370365842 Chr.2: 176083339-176083339 on Build GRCh38
rs780106623 Chr.2: 176083529-176083529 on Build GRCh38
rs755516509 Chr.2: 176083426-176083426 on Build GRCh38
rs201829860 Chr.2: 176083497-176083497 on Build GRCh38
rs149488308 Chr.2: 176083412-176083412 on Build GRCh38
rs925804994 Chr.2: 176083447-176083447 on Build GRCh38
rs201278449 Chr.2: 176083383-176083383 on Build GRCh38
rs747901985 Chr.2: 176083544-176083544 on Build GRCh38
rs575398101 Chr.2: 176083486-176083486 on Build GRCh38
rs758714494 Chr.2: 176083677-176083677 on Build GRCh38
rs768852240 Chr.2: 176083691-176083691 on Build GRCh38
rs779555868 Chr.2: 176083606-176083606 on Build GRCh38
rs766072773 Chr.2: 176083720-176083720 on Build GRCh38
rs569811881 Chr.2: 176083368-176083368 on Build GRCh38
rs536885330 Chr.2: 176083371-176083371 on Build GRCh38
rs762518774 Chr.2: 176083642-176083642 on Build GRCh38
rs774210172 Chr.2: 176083378-176083378 on Build GRCh38
rs748397665 Chr.2: 176083353-176083353 on Build GRCh38
rs375784099 Chr.2: 176083579-176083579 on Build GRCh38
rs749069138 Chr.2: 176083676-176083676 on Build GRCh38
rs372922973 Chr.2: 176083448-176083448 on Build GRCh38
rs773482435 Chr.2: 176083713-176083713 on Build GRCh38
rs945960982 Chr.2: 176083415-176083415 on Build GRCh38
rs748028727 Chr.2: 176083615-176083615 on Build GRCh38
rs765403519 Chr.2: 176083338-176083338 on Build GRCh38
rs781632361 Chr.2: 176083344-176083344 on Build GRCh38
rs368943145 Chr.2: 176083662-176083662 on Build GRCh38
rs767874850 Chr.2: 176083430-176083430 on Build GRCh38
rs772267515 Chr.2: 176083621-176083621 on Build GRCh38
rs377471251 Chr.2: 176083323-176083323 on Build GRCh38
rs765978599 Chr.2: 176083644-176083644 on Build GRCh38
rs781619146 Chr.2: 176083472-176083472 on Build GRCh38
rs756017605 Chr.2: 176083669-176083669 on Build GRCh38
rs764564856 Chr.2: 176083501-176083501 on Build GRCh38
rs759724204 Chr.2: 176083553-176083553 on Build GRCh38
rs999970199 Chr.2: 176083305-176083305 on Build GRCh38
rs757984386 Chr.2: 176083605-176083605 on Build GRCh38
rs775384369 Chr.2: 176083392-176083392 on Build GRCh38
rs746936014 Chr.2: 176083531-176083531 on Build GRCh38
rs748932110 Chr.2: 176083437-176083437 on Build GRCh38
rs781061588 Chr.2: 176083609-176083609 on Build GRCh38
rs200599713 Chr.2: 176083573-176083573 on Build GRCh38
rs765506969 Chr.2: 176083640-176083640 on Build GRCh38
rs754465827 Chr.2: 176083649-176083649 on Build GRCh38
rs992807810 Chr.2: 176083393-176083393 on Build GRCh38
rs779649535 Chr.2: 176083388-176083388 on Build GRCh38
rs973490271 Chr.2: 176083385-176083385 on Build GRCh38
rs745431749 Chr.2: 176083504-176083504 on Build GRCh38
rs780579197 Chr.2: 176083340-176083340 on Build GRCh38
rs1015118905 Chr.2: 176083602-176083602 on Build GRCh38
rs769605061 Chr.2: 176083354-176083354 on Build GRCh38
rs760785672 Chr.2: 176083625-176083625 on Build GRCh38
rs138591857 Chr.2: 176083697-176083697 on Build GRCh38
rs763176589 Chr.2: 176083466-176083466 on Build GRCh38
rs763087967 Chr.2: 176083419-176083419 on Build GRCh38
rs374328467 Chr.2: 176083355-176083355 on Build GRCh38
rs777432711 Chr.2: 176083671-176083671 on Build GRCh38
rs201090389 Chr.2: 176083384-176083384 on Build GRCh38
rs1039458114 Chr.2: 176083316-176083316 on Build GRCh38
rs1010860146 Chr.2: 176083698-176083698 on Build GRCh38
rs751093613 Chr.2: 176083648-176083648 on Build GRCh38
rs368061397 Chr.2: 176083330-176083330 on Build GRCh38
rs538737983 Chr.2: 176083425-176083425 on Build GRCh38
rs762025245 Chr.2: 176083331-176083331 on Build GRCh38
rs201905415 Chr.2: 176083450-176083450 on Build GRCh38
rs1045591214 Chr.2: 176083618-176083618 on Build GRCh38
rs987566992 Chr.2: 176083481-176083481 on Build GRCh38
rs35200230 Chr.2: 176083723-176083723 on Build GRCh38
rs773965714 Chr.2: 176083727-176083727 on Build GRCh38
rs927376477 Chr.2: 176083387-176083387 on Build GRCh38
rs955080608 Chr.2: 176083479-176083479 on Build GRCh38
rs371667432 Chr.2: 176083468-176083468 on Build GRCh38
rs200000504 Chr.2: 176083679-176083679 on Build GRCh38

View Full Product Details