Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178453149-178453646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFNB59
Entrez Gene ID
494513
Gene Name
deafness, autosomal recessive 59
Gene Aliases
PJVK
UniGene
Hs.87734
Gene Chromosome Location
Chr.2: 178450592-178461390 on Build GRCh38
Transcripts
NM_001042702.3 XM_005246627.2 XM_005246628.3 XM_005246629.3 XM_011511247.2 XM_011511249.2 XM_011511250.2 XM_011511251.1 XM_017004221.1 XM_017004222.1 XM_017004223.1 XM_017004224.1 XM_017004225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4621451 c.86A>G p.D29G Chr.2: 178453495-178453495 on Build GRCh38
7220139 c.182T>C p.I61T Chr.2: 178453591-178453591 on Build GRCh38
1249992 c.4T>A p.F2I Chr.2: 178453413-178453413 on Build GRCh38
5024575 c.97C>A p.P33T Chr.2: 178453506-178453506 on Build GRCh38
5638224 c.21G>T p.K7N Chr.2: 178453430-178453430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575089285 Chr.2: 178453408-178453408 on Build GRCh38
rs201772080 Chr.2: 178453496-178453496 on Build GRCh38
rs966441329 Chr.2: 178453320-178453320 on Build GRCh38
rs960248519 Chr.2: 178453193-178453193 on Build GRCh38
rs1033825654 Chr.2: 178453215-178453216 on Build GRCh38
rs1051230408 Chr.2: 178453432-178453432 on Build GRCh38
rs36048436 Chr.2: 178453175-178453176 on Build GRCh38
rs546673221 Chr.2: 178453342-178453342 on Build GRCh38
rs886043614 Chr.2: 178453458-178453458 on Build GRCh38
rs754576211 Chr.2: 178453602-178453602 on Build GRCh38
rs916576005 Chr.2: 178453199-178453199 on Build GRCh38
rs199572377 Chr.2: 178453365-178453365 on Build GRCh38
rs753312914 Chr.2: 178453556-178453556 on Build GRCh38
rs118203988 Chr.2: 178453570-178453570 on Build GRCh38
rs764862274 Chr.2: 178453490-178453490 on Build GRCh38
rs761206757 Chr.2: 178453478-178453478 on Build GRCh38
rs749343941 Chr.2: 178453419-178453419 on Build GRCh38
rs375789304 Chr.2: 178453395-178453395 on Build GRCh38
rs771700961 Chr.2: 178453447-178453447 on Build GRCh38
rs762258765 Chr.2: 178453249-178453249 on Build GRCh38
rs778398742 Chr.2: 178453505-178453505 on Build GRCh38
rs974192439 Chr.2: 178453323-178453323 on Build GRCh38
rs112758005 Chr.2: 178453373-178453373 on Build GRCh38
rs375687411 Chr.2: 178453544-178453544 on Build GRCh38
rs944287431 Chr.2: 178453396-178453396 on Build GRCh38
rs746776380 Chr.2: 178453549-178453549 on Build GRCh38
rs776538230 Chr.2: 178453572-178453572 on Build GRCh38
rs992133393 Chr.2: 178453194-178453194 on Build GRCh38
rs1042817579 Chr.2: 178453535-178453535 on Build GRCh38
rs769127391 Chr.2: 178453593-178453593 on Build GRCh38
rs369215803 Chr.2: 178453348-178453348 on Build GRCh38
rs746165029 Chr.2: 178453445-178453445 on Build GRCh38
rs917577049 Chr.2: 178453300-178453300 on Build GRCh38
rs761878522 Chr.2: 178453584-178453584 on Build GRCh38
rs187816687 Chr.2: 178453472-178453472 on Build GRCh38
rs777960074 Chr.2: 178453406-178453406 on Build GRCh38
rs727504946 Chr.2: 178453604-178453604 on Build GRCh38
rs753143124 Chr.2: 178453498-178453498 on Build GRCh38
rs969452431 Chr.2: 178453227-178453227 on Build GRCh38
rs746022606 Chr.2: 178453534-178453534 on Build GRCh38
rs77758075 Chr.2: 178453333-178453333 on Build GRCh38
rs538027448 Chr.2: 178453567-178453567 on Build GRCh38
rs756585755 Chr.2: 178453502-178453502 on Build GRCh38
rs748606745 Chr.2: 178453388-178453388 on Build GRCh38
rs776591320 Chr.2: 178453477-178453477 on Build GRCh38
rs757276963 Chr.2: 178453521-178453521 on Build GRCh38
rs397516555 Chr.2: 178453462-178453462 on Build GRCh38
rs778986514 Chr.2: 178453439-178453439 on Build GRCh38
rs779058198 Chr.2: 178453533-178453533 on Build GRCh38
rs959396817 Chr.2: 178453217-178453217 on Build GRCh38
rs754450957 Chr.2: 178453513-178453513 on Build GRCh38
rs750041216 Chr.2: 178453493-178453493 on Build GRCh38
rs200502817 Chr.2: 178453495-178453495 on Build GRCh38
rs144704250 Chr.2: 178453382-178453382 on Build GRCh38
rs768488521 Chr.2: 178453554-178453554 on Build GRCh38
rs959949943 Chr.2: 178453587-178453587 on Build GRCh38
rs1035584723 Chr.2: 178453180-178453180 on Build GRCh38
rs775279693 Chr.2: 178453456-178453456 on Build GRCh38

View Full Product Details