Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181544526-181545064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERKL
Entrez Gene ID
375298
Gene Name
ceramide kinase like
Gene Aliases
RP26
UniGene
Hs.732358
Gene Chromosome Location
Chr.2: 181536672-181657107 on Build GRCh38
Transcripts
NM_001030311.2 NM_001030312.2 NM_001030313.2 NM_001160277.1 NM_201548.4 NR_027689.1 NR_027690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466051 c.1414C>G p.L472V Chr.2: 181544729-181544729 on Build GRCh38
1148859 c.1414C>A p.L472M Chr.2: 181544729-181544729 on Build GRCh38
3574785 c.1279G>A p.G427R Chr.2: 181544786-181544786 on Build GRCh38
4466052 c.1336C>G p.L446V Chr.2: 181544729-181544729 on Build GRCh38
1326608 c.1343G>C p.R448T Chr.2: 181544722-181544722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376325325 Chr.2: 181544873-181544873 on Build GRCh38
rs781662259 Chr.2: 181544782-181544782 on Build GRCh38
rs75425590 Chr.2: 181544643-181544643 on Build GRCh38
rs879189421 Chr.2: 181544610-181544610 on Build GRCh38
rs34936253 Chr.2: 181544854-181544855 on Build GRCh38
rs1039793113 Chr.2: 181544651-181544651 on Build GRCh38
rs915370763 Chr.2: 181544634-181544634 on Build GRCh38
rs748713352 Chr.2: 181544565-181544565 on Build GRCh38
rs370710013 Chr.2: 181545038-181545038 on Build GRCh38
rs994598923 Chr.2: 181544947-181544947 on Build GRCh38
rs747980225 Chr.2: 181544693-181544693 on Build GRCh38
rs750553300 Chr.2: 181544771-181544771 on Build GRCh38
rs139300604 Chr.2: 181544750-181544750 on Build GRCh38
rs973448488 Chr.2: 181544614-181544614 on Build GRCh38
rs978195392 Chr.2: 181544983-181544983 on Build GRCh38
rs748979374 Chr.2: 181544717-181544717 on Build GRCh38
rs1018258434 Chr.2: 181544578-181544578 on Build GRCh38
rs987766406 Chr.2: 181544576-181544576 on Build GRCh38
rs1002303511 Chr.2: 181544886-181544886 on Build GRCh38
rs965331806 Chr.2: 181544931-181544931 on Build GRCh38
rs780944999 Chr.2: 181544667-181544667 on Build GRCh38
rs748855871 Chr.2: 181544839-181544839 on Build GRCh38
rs993228386 Chr.2: 181544823-181544823 on Build GRCh38
rs148507801 Chr.2: 181544761-181544761 on Build GRCh38
rs34150300 Chr.2: 181544774-181544774 on Build GRCh38
rs539773990 Chr.2: 181544784-181544784 on Build GRCh38
rs770284500 Chr.2: 181544718-181544718 on Build GRCh38
rs181846399 Chr.2: 181544623-181544623 on Build GRCh38
rs750834554 Chr.2: 181544646-181544646 on Build GRCh38
rs186404709 Chr.2: 181544867-181544867 on Build GRCh38
rs1033411512 Chr.2: 181544598-181544598 on Build GRCh38
rs565668147 Chr.2: 181544624-181544624 on Build GRCh38
rs964939538 Chr.2: 181544591-181544591 on Build GRCh38
rs79592949 Chr.2: 181544821-181544821 on Build GRCh38
rs1008970617 Chr.2: 181544882-181544882 on Build GRCh38
rs747115548 Chr.2: 181544954-181544954 on Build GRCh38
rs1001904241 Chr.2: 181544589-181544589 on Build GRCh38
rs755802947 Chr.2: 181544810-181544810 on Build GRCh38
rs773946833 Chr.2: 181544737-181544737 on Build GRCh38
rs890159121 Chr.2: 181544877-181544877 on Build GRCh38
rs1017892329 Chr.2: 181544926-181544926 on Build GRCh38
rs1025061334 Chr.2: 181544550-181544550 on Build GRCh38
rs747922398 Chr.2: 181544803-181544803 on Build GRCh38
rs754931712 Chr.2: 181544664-181544664 on Build GRCh38
rs928004913 Chr.2: 181544702-181544702 on Build GRCh38
rs376764539 Chr.2: 181544655-181544655 on Build GRCh38
rs769486666 Chr.2: 181544696-181544696 on Build GRCh38
rs1026396769 Chr.2: 181544953-181544953 on Build GRCh38
rs770642669 Chr.2: 181544843-181544843 on Build GRCh38
rs766319295 Chr.2: 181544778-181544778 on Build GRCh38
rs760956178 Chr.2: 181544858-181544858 on Build GRCh38
rs1026002808 Chr.2: 181544876-181544876 on Build GRCh38
rs781170341 Chr.2: 181544788-181544788 on Build GRCh38
rs987371895 Chr.2: 181545001-181545001 on Build GRCh38
rs928031044 Chr.2: 181545035-181545035 on Build GRCh38
rs772801688 Chr.2: 181544709-181544709 on Build GRCh38
rs771011979 Chr.2: 181544978-181544978 on Build GRCh38
rs765484312 Chr.2: 181544763-181544763 on Build GRCh38
rs556405190 Chr.2: 181544785-181544785 on Build GRCh38
rs963115557 Chr.2: 181544612-181544612 on Build GRCh38
rs761217466 Chr.2: 181544741-181544741 on Build GRCh38
rs970911911 Chr.2: 181544552-181544552 on Build GRCh38
rs553792277 Chr.2: 181544749-181544749 on Build GRCh38
rs936783486 Chr.2: 181544695-181544695 on Build GRCh38
rs576142350 Chr.2: 181544932-181544932 on Build GRCh38

View Full Product Details