Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181547533-181548031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERKL
Entrez Gene ID
375298
Gene Name
ceramide kinase like
Gene Aliases
RP26
UniGene
Hs.732358
Gene Chromosome Location
Chr.2: 181536672-181657107 on Build GRCh38
Transcripts
NM_001030311.2 NM_001030312.2 NM_001030313.2 NM_001160277.1 NM_201548.4 NR_027689.1 NR_027690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3574786 c.1346+2T>C p.? Chr.2: 181547616-181547616 on Build GRCh38
6627319 c.1301C>A p.P434H Chr.2: 181547663-181547663 on Build GRCh38
5551170 c.1164T>C p.C388C Chr.2: 181547722-181547722 on Build GRCh38
3574788 c.1235C>T p.S412F Chr.2: 181547824-181547824 on Build GRCh38
3574787 c.1268+2T>C p.? Chr.2: 181547616-181547616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765084311 Chr.2: 181547887-181547887 on Build GRCh38
rs56263562 Chr.2: 181547983-181547983 on Build GRCh38
rs767594454 Chr.2: 181547621-181547621 on Build GRCh38
rs62189984 Chr.2: 181547898-181547898 on Build GRCh38
rs770561248 Chr.2: 181547798-181547798 on Build GRCh38
rs779716333 Chr.2: 181547687-181547687 on Build GRCh38
rs760510226 Chr.2: 181547746-181547746 on Build GRCh38
rs778400039 Chr.2: 181547807-181547807 on Build GRCh38
rs201778792 Chr.2: 181547814-181547814 on Build GRCh38
rs370307513 Chr.2: 181547893-181547893 on Build GRCh38
rs145489428 Chr.2: 181547649-181547649 on Build GRCh38
rs751631672 Chr.2: 181547617-181547617 on Build GRCh38
rs371304137 Chr.2: 181547995-181547995 on Build GRCh38
rs771650977 Chr.2: 181547683-181547683 on Build GRCh38
rs756111982 Chr.2: 181547634-181547634 on Build GRCh38
rs770167336 Chr.2: 181547609-181547612 on Build GRCh38
rs745413620 Chr.2: 181547578-181547578 on Build GRCh38
rs952733964 Chr.2: 181547779-181547779 on Build GRCh38
rs377625662 Chr.2: 181547631-181547631 on Build GRCh38
rs759709536 Chr.2: 181547839-181547839 on Build GRCh38
rs949291156 Chr.2: 181547688-181547688 on Build GRCh38
rs770381086 Chr.2: 181547711-181547711 on Build GRCh38
rs758146598 Chr.2: 181547786-181547786 on Build GRCh38
rs139036113 Chr.2: 181547906-181547907 on Build GRCh38
rs747024040 Chr.2: 181547667-181547667 on Build GRCh38
rs766663317 Chr.2: 181547614-181547614 on Build GRCh38
rs535881034 Chr.2: 181547574-181547574 on Build GRCh38
rs370942261 Chr.2: 181547888-181547888 on Build GRCh38
rs757135390 Chr.2: 181547764-181547764 on Build GRCh38
rs558326688 Chr.2: 181547755-181547755 on Build GRCh38
rs776211842 Chr.2: 181547836-181547836 on Build GRCh38
rs753753048 Chr.2: 181547881-181547881 on Build GRCh38
rs773648306 Chr.2: 181547607-181547607 on Build GRCh38
rs753718511 Chr.2: 181547653-181547653 on Build GRCh38
rs773416880 Chr.2: 181547712-181547712 on Build GRCh38
rs773410577 Chr.2: 181547810-181547811 on Build GRCh38
rs771214260 Chr.2: 181547815-181547815 on Build GRCh38
rs984424507 Chr.2: 181547999-181547999 on Build GRCh38
rs917223036 Chr.2: 181547947-181547947 on Build GRCh38
rs114503048 Chr.2: 181547889-181547889 on Build GRCh38
rs777336671 Chr.2: 181547586-181547586 on Build GRCh38
rs772997424 Chr.2: 181547628-181547628 on Build GRCh38
rs949340312 Chr.2: 181547953-181547953 on Build GRCh38
rs771126203 Chr.2: 181547736-181547736 on Build GRCh38
rs757788333 Chr.2: 181547897-181547897 on Build GRCh38
rs778776805 Chr.2: 181547660-181547660 on Build GRCh38
rs755105366 Chr.2: 181547618-181547618 on Build GRCh38
rs769283517 Chr.2: 181547585-181547585 on Build GRCh38
rs184936801 Chr.2: 181547890-181547890 on Build GRCh38
rs748555018 Chr.2: 181547709-181547709 on Build GRCh38
rs763062899 Chr.2: 181547609-181547609 on Build GRCh38
rs869084061 Chr.2: 181547999-181548000 on Build GRCh38
rs144793035 Chr.2: 181547694-181547694 on Build GRCh38
rs998957845 Chr.2: 181547894-181547901 on Build GRCh38
rs866804422 Chr.2: 181547750-181547751 on Build GRCh38
rs749565720 Chr.2: 181547893-181547894 on Build GRCh38
rs140237265 Chr.2: 181547789-181547789 on Build GRCh38
rs989219893 Chr.2: 181547912-181547912 on Build GRCh38
rs761847069 Chr.2: 181547885-181547885 on Build GRCh38
rs148206460 Chr.2: 181547777-181547777 on Build GRCh38
rs760878115 Chr.2: 181547872-181547872 on Build GRCh38
rs926487034 Chr.2: 181548002-181548002 on Build GRCh38
rs772333970 Chr.2: 181547843-181547843 on Build GRCh38
rs778961980 Chr.2: 181547602-181547602 on Build GRCh38
rs748587518 Chr.2: 181547594-181547594 on Build GRCh38
rs866139873 Chr.2: 181547797-181547797 on Build GRCh38
rs34890422 Chr.2: 181547996-181547997 on Build GRCh38
rs377332590 Chr.2: 181547898-181547899 on Build GRCh38
rs373597465 Chr.2: 181547819-181547819 on Build GRCh38
rs533705291 Chr.2: 181547590-181547590 on Build GRCh38
rs377703431 Chr.2: 181547994-181547994 on Build GRCh38
rs763927549 Chr.2: 181547875-181547875 on Build GRCh38
rs759754421 Chr.2: 181547743-181547743 on Build GRCh38
rs923320747 Chr.2: 181547867-181547867 on Build GRCh38
rs775405083 Chr.2: 181547853-181547853 on Build GRCh38
rs762395418 Chr.2: 181547588-181547588 on Build GRCh38
rs67145738 Chr.2: 181547997-181547998 on Build GRCh38
rs77584054 Chr.2: 181547996-181547996 on Build GRCh38
rs372755116 Chr.2: 181547682-181547682 on Build GRCh38
rs369507626 Chr.2: 181547672-181547672 on Build GRCh38
rs575397538 Chr.2: 181547762-181547762 on Build GRCh38
rs28450214 Chr.2: 181547906-181547906 on Build GRCh38
rs530000435 Chr.2: 181547806-181547806 on Build GRCh38
rs116599132 Chr.2: 181547892-181547892 on Build GRCh38
rs934643895 Chr.2: 181547606-181547606 on Build GRCh38
rs905011448 Chr.2: 181547722-181547722 on Build GRCh38
rs58144427 Chr.2: 181547915-181547916 on Build GRCh38
rs763439339 Chr.2: 181547713-181547713 on Build GRCh38
rs774496068 Chr.2: 181547741-181547741 on Build GRCh38
rs889333607 Chr.2: 181547622-181547622 on Build GRCh38
rs959802279 Chr.2: 181547742-181547742 on Build GRCh38
rs959750557 Chr.2: 181547946-181547948 on Build GRCh38
rs372111150 Chr.2: 181547891-181547891 on Build GRCh38
rs1040916917 Chr.2: 181547834-181547834 on Build GRCh38
rs567649380 Chr.2: 181547962-181547962 on Build GRCh38
rs143023765 Chr.2: 181547744-181547744 on Build GRCh38
rs376997713 Chr.2: 181547774-181547774 on Build GRCh38

View Full Product Details