Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181548467-181548983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERKL
Entrez Gene ID
375298
Gene Name
ceramide kinase like
Gene Aliases
RP26
UniGene
Hs.732358
Gene Chromosome Location
Chr.2: 181536672-181657107 on Build GRCh38
Transcripts
NM_001030311.2 NM_001030312.2 NM_001030313.2 NM_001160277.1 NM_201548.4 NR_027689.1 NR_027690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1591468 c.1090C>T p.R364* Chr.2: 181548741-181548741 on Build GRCh38
1306199 c.912C>T p.V304V Chr.2: 181548841-181548841 on Build GRCh38
6577756 c.1073+5G>A p.? Chr.2: 181548675-181548675 on Build GRCh38
6627323 c.1099T>C p.S367P Chr.2: 181548732-181548732 on Build GRCh38
5736345 c.1123G>A p.A375T Chr.2: 181548708-181548708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13003064 Chr.2: 181548875-181548875 on Build GRCh38
rs755223434 Chr.2: 181548738-181548738 on Build GRCh38
rs952026209 Chr.2: 181548934-181548934 on Build GRCh38
rs200778934 Chr.2: 181548870-181548870 on Build GRCh38
rs774034295 Chr.2: 181548643-181548643 on Build GRCh38
rs375613779 Chr.2: 181548771-181548771 on Build GRCh38
rs759320359 Chr.2: 181548773-181548773 on Build GRCh38
rs370011429 Chr.2: 181548534-181548534 on Build GRCh38
rs777974407 Chr.2: 181548847-181548847 on Build GRCh38
rs780215788 Chr.2: 181548508-181548508 on Build GRCh38
rs530250724 Chr.2: 181548869-181548869 on Build GRCh38
rs762736738 Chr.2: 181548844-181548844 on Build GRCh38
rs748394238 Chr.2: 181548741-181548741 on Build GRCh38
rs141763526 Chr.2: 181548762-181548762 on Build GRCh38
rs562467333 Chr.2: 181548665-181548665 on Build GRCh38
rs142530341 Chr.2: 181548804-181548804 on Build GRCh38
rs72883670 Chr.2: 181548511-181548511 on Build GRCh38
rs746784970 Chr.2: 181548628-181548628 on Build GRCh38
rs756214910 Chr.2: 181548498-181548498 on Build GRCh38
rs765108142 Chr.2: 181548712-181548713 on Build GRCh38
rs778775955 Chr.2: 181548854-181548854 on Build GRCh38
rs745341953 Chr.2: 181548800-181548800 on Build GRCh38
rs761789633 Chr.2: 181548613-181548613 on Build GRCh38
rs933690522 Chr.2: 181548728-181548728 on Build GRCh38
rs767642187 Chr.2: 181548524-181548536 on Build GRCh38
rs140582540 Chr.2: 181548550-181548550 on Build GRCh38
rs760326517 Chr.2: 181548785-181548785 on Build GRCh38
rs532672989 Chr.2: 181548891-181548891 on Build GRCh38
rs12997453 Chr.2: 181548532-181548532 on Build GRCh38
rs769924910 Chr.2: 181548742-181548742 on Build GRCh38
rs748304433 Chr.2: 181548619-181548619 on Build GRCh38
rs972596991 Chr.2: 181548946-181548946 on Build GRCh38
rs749363766 Chr.2: 181548848-181548848 on Build GRCh38
rs1029609033 Chr.2: 181548620-181548620 on Build GRCh38
rs375617532 Chr.2: 181548496-181548496 on Build GRCh38
rs752257366 Chr.2: 181548545-181548545 on Build GRCh38
rs896807298 Chr.2: 181548880-181548880 on Build GRCh38
rs761820123 Chr.2: 181548510-181548510 on Build GRCh38
rs781325278 Chr.2: 181548740-181548740 on Build GRCh38
rs180705205 Chr.2: 181548529-181548529 on Build GRCh38
rs896858206 Chr.2: 181548593-181548593 on Build GRCh38
rs372575043 Chr.2: 181548763-181548763 on Build GRCh38
rs727503856 Chr.2: 181548719-181548719 on Build GRCh38
rs1016846315 Chr.2: 181548937-181548937 on Build GRCh38
rs186100602 Chr.2: 181548669-181548669 on Build GRCh38
rs776727320 Chr.2: 181548591-181548592 on Build GRCh38
rs866794217 Chr.2: 181548584-181548584 on Build GRCh38
rs544311424 Chr.2: 181548828-181548828 on Build GRCh38
rs762897430 Chr.2: 181548519-181548519 on Build GRCh38
rs562686255 Chr.2: 181548601-181548601 on Build GRCh38
rs139270710 Chr.2: 181548841-181548841 on Build GRCh38
rs369454299 Chr.2: 181548648-181548648 on Build GRCh38
rs568712409 Chr.2: 181548936-181548936 on Build GRCh38
rs868737394 Chr.2: 181548758-181548758 on Build GRCh38
rs756424312 Chr.2: 181548846-181548846 on Build GRCh38
rs776113557 Chr.2: 181548887-181548887 on Build GRCh38
rs751908951 Chr.2: 181548734-181548734 on Build GRCh38
rs202124278 Chr.2: 181548873-181548873 on Build GRCh38
rs941111205 Chr.2: 181548872-181548875 on Build GRCh38
rs773135809 Chr.2: 181548512-181548512 on Build GRCh38
rs1022558791 Chr.2: 181548706-181548706 on Build GRCh38
rs924599354 Chr.2: 181548731-181548731 on Build GRCh38
rs1034329248 Chr.2: 181548928-181548928 on Build GRCh38
rs200179024 Chr.2: 181548610-181548610 on Build GRCh38
rs757735194 Chr.2: 181548546-181548546 on Build GRCh38
rs755238456 Chr.2: 181548832-181548832 on Build GRCh38
rs372622726 Chr.2: 181548558-181548558 on Build GRCh38
rs769479669 Chr.2: 181548807-181548807 on Build GRCh38
rs761288116 Chr.2: 181548889-181548889 on Build GRCh38
rs769838976 Chr.2: 181548626-181548626 on Build GRCh38
rs771744533 Chr.2: 181548775-181548775 on Build GRCh38
rs758116997 Chr.2: 181548791-181548792 on Build GRCh38
rs770716465 Chr.2: 181548638-181548638 on Build GRCh38
rs747214130 Chr.2: 181548570-181548570 on Build GRCh38
rs201231831 Chr.2: 181548720-181548720 on Build GRCh38
rs752202160 Chr.2: 181548658-181548658 on Build GRCh38
rs778028617 Chr.2: 181548752-181548752 on Build GRCh38
rs781692915 Chr.2: 181548837-181548837 on Build GRCh38
rs144917662 Chr.2: 181548689-181548689 on Build GRCh38
rs780287984 Chr.2: 181548566-181548566 on Build GRCh38
rs750151209 Chr.2: 181548785-181548786 on Build GRCh38
rs746220721 Chr.2: 181548551-181548551 on Build GRCh38
rs150587104 Chr.2: 181548803-181548803 on Build GRCh38
rs148062068 Chr.2: 181548778-181548778 on Build GRCh38
rs374294052 Chr.2: 181548895-181548895 on Build GRCh38
rs376617675 Chr.2: 181548712-181548712 on Build GRCh38
rs183252158 Chr.2: 181548853-181548853 on Build GRCh38
rs768716099 Chr.2: 181548502-181548502 on Build GRCh38
rs751155027 Chr.2: 181548530-181548530 on Build GRCh38
rs971842447 Chr.2: 181548565-181548565 on Build GRCh38
rs927261937 Chr.2: 181548605-181548605 on Build GRCh38
rs201084053 Chr.2: 181548650-181548650 on Build GRCh38
rs747210372 Chr.2: 181548501-181548501 on Build GRCh38
rs768347335 Chr.2: 181548881-181548881 on Build GRCh38
rs369379908 Chr.2: 181548726-181548726 on Build GRCh38
rs202025850 Chr.2: 181548661-181548661 on Build GRCh38
rs762985021 Chr.2: 181548635-181548635 on Build GRCh38
rs774652583 Chr.2: 181548892-181548892 on Build GRCh38
rs371048058 Chr.2: 181548820-181548820 on Build GRCh38
rs76427948 Chr.2: 181548942-181548942 on Build GRCh38
rs760238967 Chr.2: 181548664-181548664 on Build GRCh38
rs1049431984 Chr.2: 181548730-181548730 on Build GRCh38
rs190967171 Chr.2: 181548819-181548819 on Build GRCh38

View Full Product Details