Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 182719843-182720342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
UniGene
Hs.516632
Gene Chromosome Location
Chr.2: 182716041-182780025 on Build GRCh38
Transcripts
NM_001271581.1 NM_018981.2 NR_073365.1 NR_073366.1 NR_073367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832124 c.316G>A p.D106N Chr.2: 182720118-182720118 on Build GRCh38
4410795 c.247G>A p.A83T Chr.2: 182720049-182720049 on Build GRCh38
6156398 c.277C>T p.R93W Chr.2: 182720079-182720079 on Build GRCh38
1494696 c.233T>A p.L78* Chr.2: 182720035-182720035 on Build GRCh38
3391333 c.210C>A p.N70K Chr.2: 182720012-182720012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748413503 Chr.2: 182719985-182719985 on Build GRCh38
rs201607521 Chr.2: 182720147-182720147 on Build GRCh38
rs763803228 Chr.2: 182720157-182720157 on Build GRCh38
rs779165009 Chr.2: 182720193-182720193 on Build GRCh38
rs769071349 Chr.2: 182719966-182719966 on Build GRCh38
rs148224670 Chr.2: 182719884-182719884 on Build GRCh38
rs1002315771 Chr.2: 182720087-182720087 on Build GRCh38
rs952881921 Chr.2: 182720124-182720124 on Build GRCh38
rs531513055 Chr.2: 182720182-182720182 on Build GRCh38
rs775499650 Chr.2: 182720212-182720212 on Build GRCh38
rs969357396 Chr.2: 182720080-182720080 on Build GRCh38
rs377549612 Chr.2: 182720110-182720110 on Build GRCh38
rs757470477 Chr.2: 182720189-182720189 on Build GRCh38
rs774550749 Chr.2: 182720105-182720105 on Build GRCh38
rs781569732 Chr.2: 182720082-182720082 on Build GRCh38
rs934374271 Chr.2: 182720280-182720280 on Build GRCh38
rs894945620 Chr.2: 182720264-182720264 on Build GRCh38
rs201955943 Chr.2: 182720127-182720127 on Build GRCh38
rs755503148 Chr.2: 182720179-182720179 on Build GRCh38
rs745654560 Chr.2: 182720214-182720214 on Build GRCh38
rs895012054 Chr.2: 182720008-182720008 on Build GRCh38
rs995092496 Chr.2: 182720267-182720267 on Build GRCh38
rs774221574 Chr.2: 182720020-182720020 on Build GRCh38
rs200695145 Chr.2: 182720013-182720013 on Build GRCh38
rs767468170 Chr.2: 182720023-182720023 on Build GRCh38
rs374079846 Chr.2: 182720215-182720215 on Build GRCh38
rs749520487 Chr.2: 182720094-182720094 on Build GRCh38
rs199619368 Chr.2: 182720107-182720107 on Build GRCh38
rs906263835 Chr.2: 182719944-182719944 on Build GRCh38
rs1042775372 Chr.2: 182719940-182719940 on Build GRCh38
rs753399511 Chr.2: 182720050-182720050 on Build GRCh38
rs755450702 Chr.2: 182720079-182720079 on Build GRCh38
rs763602266 Chr.2: 182720044-182720044 on Build GRCh38
rs770971543 Chr.2: 182720099-182720099 on Build GRCh38
rs946303106 Chr.2: 182720251-182720251 on Build GRCh38
rs759532288 Chr.2: 182720021-182720021 on Build GRCh38
rs965299313 Chr.2: 182720162-182720162 on Build GRCh38
rs370314672 Chr.2: 182720174-182720174 on Build GRCh38
rs6729801 Chr.2: 182720028-182720028 on Build GRCh38
rs904242919 Chr.2: 182719963-182719963 on Build GRCh38
rs372983378 Chr.2: 182720176-182720176 on Build GRCh38
rs548440427 Chr.2: 182720209-182720209 on Build GRCh38
rs756375421 Chr.2: 182720091-182720091 on Build GRCh38
rs575073487 Chr.2: 182719969-182719969 on Build GRCh38
rs1042137212 Chr.2: 182719934-182719934 on Build GRCh38
rs770906503 Chr.2: 182720015-182720015 on Build GRCh38
rs377016343 Chr.2: 182720185-182720185 on Build GRCh38
rs1042061480 Chr.2: 182720252-182720252 on Build GRCh38
rs945035081 Chr.2: 182719885-182719885 on Build GRCh38
rs983437541 Chr.2: 182720047-182720047 on Build GRCh38
rs936172913 Chr.2: 182720228-182720228 on Build GRCh38
rs776207150 Chr.2: 182720158-182720158 on Build GRCh38
rs1031549494 Chr.2: 182719998-182719998 on Build GRCh38
rs1003731559 Chr.2: 182719961-182719961 on Build GRCh38
rs1055117726 Chr.2: 182719878-182719878 on Build GRCh38
rs769964845 Chr.2: 182719990-182719990 on Build GRCh38
rs913502060 Chr.2: 182720245-182720245 on Build GRCh38
rs772844876 Chr.2: 182719949-182719949 on Build GRCh38
rs990386860 Chr.2: 182720240-182720240 on Build GRCh38
rs150009047 Chr.2: 182720027-182720027 on Build GRCh38
rs778185472 Chr.2: 182720184-182720184 on Build GRCh38
rs932332344 Chr.2: 182720231-182720231 on Build GRCh38
rs769769678 Chr.2: 182720256-182720256 on Build GRCh38
rs370741044 Chr.2: 182720199-182720199 on Build GRCh38
rs761506704 Chr.2: 182720169-182720169 on Build GRCh38
rs376345220 Chr.2: 182719996-182719996 on Build GRCh38
rs767954814 Chr.2: 182720181-182720181 on Build GRCh38
rs141135661 Chr.2: 182720118-182720118 on Build GRCh38
rs932441972 Chr.2: 182719893-182719893 on Build GRCh38
rs748462996 Chr.2: 182720089-182720089 on Build GRCh38
rs764792929 Chr.2: 182720073-182720073 on Build GRCh38
rs374378803 Chr.2: 182719919-182719919 on Build GRCh38
rs1052652805 Chr.2: 182720295-182720295 on Build GRCh38
rs561958567 Chr.2: 182720263-182720263 on Build GRCh38
rs982637666 Chr.2: 182720129-182720129 on Build GRCh38
rs762853337 Chr.2: 182720024-182720024 on Build GRCh38
rs756788319 Chr.2: 182720051-182720051 on Build GRCh38
rs554825684 Chr.2: 182719937-182719937 on Build GRCh38
rs906231313 Chr.2: 182720253-182720253 on Build GRCh38
rs879822227 Chr.2: 182720062-182720062 on Build GRCh38

View Full Product Details