Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 185755937-185756444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP2
Entrez Gene ID
401024
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
-
UniGene
Hs.98025
Gene Chromosome Location
Chr.2: 185738895-185833289 on Build GRCh38
Transcripts
NM_173651.3

SNP Detail

SNP ID Chromosome Location
rs938180528 Chr.2: 185756092-185756092 on Build GRCh38
rs745571898 Chr.2: 185756240-185756249 on Build GRCh38
rs114498031 Chr.2: 185756298-185756298 on Build GRCh38
rs140245371 Chr.2: 185755967-185755967 on Build GRCh38
rs144899704 Chr.2: 185756283-185756283 on Build GRCh38
rs150295363 Chr.2: 185756039-185756039 on Build GRCh38
rs536167768 Chr.2: 185756180-185756180 on Build GRCh38
rs777035091 Chr.2: 185756285-185756285 on Build GRCh38
rs1009862682 Chr.2: 185756413-185756413 on Build GRCh38
rs1016981913 Chr.2: 185756142-185756142 on Build GRCh38
rs541480643 Chr.2: 185756327-185756327 on Build GRCh38
rs564579899 Chr.2: 185755989-185755989 on Build GRCh38
rs565639916 Chr.2: 185756129-185756129 on Build GRCh38
rs189753181 Chr.2: 185756230-185756230 on Build GRCh38
rs750226627 Chr.2: 185756002-185756002 on Build GRCh38
rs968836224 Chr.2: 185756041-185756041 on Build GRCh38
rs770495450 Chr.2: 185756007-185756007 on Build GRCh38
rs935739332 Chr.2: 185756350-185756351 on Build GRCh38
rs901064773 Chr.2: 185756398-185756398 on Build GRCh38
rs543681247 Chr.2: 185756331-185756331 on Build GRCh38
rs755190836 Chr.2: 185756058-185756058 on Build GRCh38
rs949379182 Chr.2: 185756234-185756234 on Build GRCh38
rs962786763 Chr.2: 185756264-185756264 on Build GRCh38
rs1028484911 Chr.2: 185756275-185756275 on Build GRCh38
rs1046079208 Chr.2: 185756059-185756059 on Build GRCh38
rs925696189 Chr.2: 185755977-185755977 on Build GRCh38
rs1004630375 Chr.2: 185756137-185756137 on Build GRCh38
rs1020411664 Chr.2: 185756414-185756414 on Build GRCh38
rs1048466844 Chr.2: 185756319-185756319 on Build GRCh38
rs576344341 Chr.2: 185756246-185756246 on Build GRCh38
rs977488712 Chr.2: 185756339-185756339 on Build GRCh38
rs10084180 Chr.2: 185756062-185756062 on Build GRCh38
rs763533573 Chr.2: 185756167-185756167 on Build GRCh38
rs184376848 Chr.2: 185755974-185755974 on Build GRCh38
rs780672854 Chr.2: 185756235-185756235 on Build GRCh38
rs991094161 Chr.2: 185756134-185756134 on Build GRCh38
rs181232065 Chr.2: 185756269-185756269 on Build GRCh38
rs142223453 Chr.2: 185756366-185756366 on Build GRCh38
rs116667549 Chr.2: 185756340-185756340 on Build GRCh38
rs753436296 Chr.2: 185756185-185756185 on Build GRCh38
rs931427315 Chr.2: 185756308-185756308 on Build GRCh38
rs756690417 Chr.2: 185756202-185756202 on Build GRCh38
rs752767728 Chr.2: 185756082-185756082 on Build GRCh38
rs570994675 Chr.2: 185756023-185756023 on Build GRCh38
rs771601946 Chr.2: 185756376-185756376 on Build GRCh38
rs755721870 Chr.2: 185756151-185756151 on Build GRCh38
rs918151296 Chr.2: 185756224-185756224 on Build GRCh38
rs761798539 Chr.2: 185755984-185755984 on Build GRCh38

View Full Product Details