Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196656553-196657054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC150
Entrez Gene ID
284992
Gene Name
coiled-coil domain containing 150
Gene Aliases
-
UniGene
Hs.132519
Gene Chromosome Location
Chr.2: 196639632-196733668 on Build GRCh38
Transcripts
NM_001080539.1 XM_006712438.3 XM_011510985.2 XM_011510988.2 XM_017003861.1 XM_017003862.1 XM_017003863.1 XM_017003864.1 XM_017003865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1161332 c.347T>G p.M116R Chr.2: 196656803-196656803 on Build GRCh38
3575970 c.210C>T p.D70D Chr.2: 196656666-196656666 on Build GRCh38
442057 c.276T>G p.I92M Chr.2: 196656732-196656732 on Build GRCh38
6621715 c.367A>G p.T123A Chr.2: 196656823-196656823 on Build GRCh38
1723500 c.339C>T p.S113S Chr.2: 196656795-196656795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554218636 Chr.2: 196656744-196656744 on Build GRCh38
rs767062411 Chr.2: 196656910-196656910 on Build GRCh38
rs959422573 Chr.2: 196656735-196656735 on Build GRCh38
rs747654565 Chr.2: 196656973-196656973 on Build GRCh38
rs182157014 Chr.2: 196656912-196656912 on Build GRCh38
rs909877313 Chr.2: 196656954-196656954 on Build GRCh38
rs965531104 Chr.2: 196656926-196656926 on Build GRCh38
rs779075739 Chr.2: 196656731-196656731 on Build GRCh38
rs757096178 Chr.2: 196656705-196656705 on Build GRCh38
rs34133636 Chr.2: 196657026-196657026 on Build GRCh38
rs1020626783 Chr.2: 196656925-196656925 on Build GRCh38
rs750339292 Chr.2: 196656795-196656795 on Build GRCh38
rs766221250 Chr.2: 196656814-196656814 on Build GRCh38
rs368606812 Chr.2: 196656827-196656827 on Build GRCh38
rs143904259 Chr.2: 196656745-196656746 on Build GRCh38
rs758716984 Chr.2: 196656947-196656947 on Build GRCh38
rs781731493 Chr.2: 196657016-196657016 on Build GRCh38
rs900750379 Chr.2: 196656851-196656851 on Build GRCh38
rs781125429 Chr.2: 196656717-196656718 on Build GRCh38
rs763837364 Chr.2: 196656702-196656702 on Build GRCh38
rs192386368 Chr.2: 196656948-196656948 on Build GRCh38
rs186763920 Chr.2: 196656915-196656915 on Build GRCh38
rs764762442 Chr.2: 196656684-196656684 on Build GRCh38
rs771332938 Chr.2: 196656764-196656764 on Build GRCh38
rs776986542 Chr.2: 196656854-196656854 on Build GRCh38
rs1002111923 Chr.2: 196656846-196656846 on Build GRCh38
rs890864456 Chr.2: 196656634-196656634 on Build GRCh38
rs758987051 Chr.2: 196656683-196656683 on Build GRCh38
rs760354989 Chr.2: 196656913-196656913 on Build GRCh38
rs746311530 Chr.2: 196656963-196656963 on Build GRCh38
rs749459156 Chr.2: 196656583-196656583 on Build GRCh38
rs368906039 Chr.2: 196656972-196656972 on Build GRCh38
rs772370932 Chr.2: 196656746-196656746 on Build GRCh38
rs200072904 Chr.2: 196657011-196657011 on Build GRCh38
rs760537528 Chr.2: 196656979-196656979 on Build GRCh38
rs769020235 Chr.2: 196656594-196656594 on Build GRCh38
rs753958693 Chr.2: 196656815-196656815 on Build GRCh38
rs768241654 Chr.2: 196656868-196656868 on Build GRCh38
rs774755610 Chr.2: 196656610-196656610 on Build GRCh38
rs112754611 Chr.2: 196656728-196656728 on Build GRCh38
rs755025333 Chr.2: 196656816-196656816 on Build GRCh38
rs762160681 Chr.2: 196656647-196656647 on Build GRCh38
rs374773530 Chr.2: 196656758-196656758 on Build GRCh38
rs776550640 Chr.2: 196657002-196657002 on Build GRCh38
rs376154930 Chr.2: 196656949-196656949 on Build GRCh38
rs781772582 Chr.2: 196656958-196656958 on Build GRCh38
rs936810732 Chr.2: 196656824-196656824 on Build GRCh38
rs757646842 Chr.2: 196657033-196657033 on Build GRCh38
rs756155108 Chr.2: 196656717-196656717 on Build GRCh38
rs769826586 Chr.2: 196656741-196656742 on Build GRCh38
rs370689689 Chr.2: 196656666-196656666 on Build GRCh38
rs1013398446 Chr.2: 196656794-196656794 on Build GRCh38
rs766839293 Chr.2: 196656945-196656945 on Build GRCh38
rs755078518 Chr.2: 196656918-196656918 on Build GRCh38
rs774151984 Chr.2: 196656882-196656882 on Build GRCh38
rs765158094 Chr.2: 196657007-196657007 on Build GRCh38
rs768296702 Chr.2: 196656970-196656970 on Build GRCh38
rs773742697 Chr.2: 196656679-196656679 on Build GRCh38
rs1015344453 Chr.2: 196656665-196656665 on Build GRCh38
rs767453236 Chr.2: 196656739-196656739 on Build GRCh38
rs368558740 Chr.2: 196656937-196656937 on Build GRCh38
rs200403800 Chr.2: 196656763-196656763 on Build GRCh38
rs200786144 Chr.2: 196656825-196656825 on Build GRCh38
rs765231798 Chr.2: 196656923-196656923 on Build GRCh38
rs779126800 Chr.2: 196656822-196656822 on Build GRCh38
rs748829723 Chr.2: 196656857-196656857 on Build GRCh38
rs761501426 Chr.2: 196656890-196656890 on Build GRCh38
rs267599147 Chr.2: 196656810-196656810 on Build GRCh38
rs767278662 Chr.2: 196656788-196656788 on Build GRCh38
rs1028114321 Chr.2: 196656595-196656595 on Build GRCh38
rs753722817 Chr.2: 196656916-196656916 on Build GRCh38
rs764698955 Chr.2: 196656773-196656773 on Build GRCh38
rs749313539 Chr.2: 196656726-196656726 on Build GRCh38
rs771795597 Chr.2: 196656974-196656974 on Build GRCh38
rs750003309 Chr.2: 196656804-196656804 on Build GRCh38
rs774098684 Chr.2: 196656786-196656786 on Build GRCh38
rs773621249 Chr.2: 196656754-196656754 on Build GRCh38
rs762923560 Chr.2: 196657025-196657025 on Build GRCh38
rs780122546 Chr.2: 196656724-196656724 on Build GRCh38
rs370426343 Chr.2: 196656713-196656713 on Build GRCh38
rs377366104 Chr.2: 196656711-196656711 on Build GRCh38
rs751444452 Chr.2: 196656704-196656704 on Build GRCh38
rs747333043 Chr.2: 196656840-196656840 on Build GRCh38
rs763888392 Chr.2: 196656784-196656784 on Build GRCh38
rs770588840 Chr.2: 196656985-196656985 on Build GRCh38
rs536411223 Chr.2: 196656844-196656844 on Build GRCh38
rs755378315 Chr.2: 196656990-196656990 on Build GRCh38
rs758763208 Chr.2: 196656826-196656826 on Build GRCh38
rs950093867 Chr.2: 196656995-196656995 on Build GRCh38
rs202212006 Chr.2: 196656691-196656691 on Build GRCh38
rs372655242 Chr.2: 196657003-196657003 on Build GRCh38
rs565319357 Chr.2: 196656580-196656580 on Build GRCh38
rs370440820 Chr.2: 196656782-196656782 on Build GRCh38
rs761732630 Chr.2: 196656787-196656787 on Build GRCh38
rs781015324 Chr.2: 196656709-196656709 on Build GRCh38
rs949413562 Chr.2: 196656789-196656789 on Build GRCh38
rs761524345 Chr.2: 196656591-196656591 on Build GRCh38
rs748026199 Chr.2: 196656734-196656735 on Build GRCh38

View Full Product Details