Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196658608-196659127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC150
Entrez Gene ID
284992
Gene Name
coiled-coil domain containing 150
Gene Aliases
-
UniGene
Hs.132519
Gene Chromosome Location
Chr.2: 196639632-196733668 on Build GRCh38
Transcripts
NM_001080539.1 XM_006712438.3 XM_011510985.2 XM_011510988.2 XM_017003861.1 XM_017003862.1 XM_017003863.1 XM_017003864.1 XM_017003865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6052169 c.601G>T p.E201* Chr.2: 196658816-196658816 on Build GRCh38
3838209 c.598G>A p.E200K Chr.2: 196658813-196658813 on Build GRCh38
5814333 c.579G>A p.K193K Chr.2: 196658794-196658794 on Build GRCh38
6621714 c.600G>T p.E200D Chr.2: 196658815-196658815 on Build GRCh38
5078890 c.620G>A p.R207H Chr.2: 196658835-196658835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773514728 Chr.2: 196658752-196658752 on Build GRCh38
rs748257358 Chr.2: 196658862-196658862 on Build GRCh38
rs1029492230 Chr.2: 196658672-196658672 on Build GRCh38
rs370222110 Chr.2: 196658784-196658784 on Build GRCh38
rs944749077 Chr.2: 196658933-196658933 on Build GRCh38
rs956411388 Chr.2: 196658759-196658759 on Build GRCh38
rs760922285 Chr.2: 196658880-196658880 on Build GRCh38
rs34638305 Chr.2: 196658826-196658826 on Build GRCh38
rs552240470 Chr.2: 196659101-196659101 on Build GRCh38
rs1049707161 Chr.2: 196658966-196658966 on Build GRCh38
rs749107179 Chr.2: 196658855-196658855 on Build GRCh38
rs773573597 Chr.2: 196658874-196658874 on Build GRCh38
rs546745957 Chr.2: 196659021-196659021 on Build GRCh38
rs774363461 Chr.2: 196658748-196658748 on Build GRCh38
rs780854947 Chr.2: 196658828-196658828 on Build GRCh38
rs779774670 Chr.2: 196658842-196658842 on Build GRCh38
rs566260668 Chr.2: 196659040-196659040 on Build GRCh38
rs534206869 Chr.2: 196658817-196658817 on Build GRCh38
rs764414407 Chr.2: 196659006-196659006 on Build GRCh38
rs1049189630 Chr.2: 196658645-196658645 on Build GRCh38
rs113426918 Chr.2: 196658859-196658859 on Build GRCh38
rs751080342 Chr.2: 196658818-196658818 on Build GRCh38
rs768514213 Chr.2: 196658858-196658858 on Build GRCh38
rs77680272 Chr.2: 196658659-196658659 on Build GRCh38
rs766608625 Chr.2: 196658890-196658890 on Build GRCh38
rs141389222 Chr.2: 196658959-196658959 on Build GRCh38
rs765768562 Chr.2: 196658801-196658801 on Build GRCh38
rs918242899 Chr.2: 196658760-196658760 on Build GRCh38
rs775509388 Chr.2: 196658794-196658794 on Build GRCh38
rs1041736426 Chr.2: 196658972-196658972 on Build GRCh38
rs536731289 Chr.2: 196658802-196658802 on Build GRCh38
rs1023623360 Chr.2: 196658731-196658731 on Build GRCh38
rs997550660 Chr.2: 196658667-196658667 on Build GRCh38
rs907448013 Chr.2: 196659020-196659020 on Build GRCh38
rs533978833 Chr.2: 196658739-196658739 on Build GRCh38
rs766782719 Chr.2: 196658781-196658781 on Build GRCh38
rs772235262 Chr.2: 196658872-196658872 on Build GRCh38
rs550312405 Chr.2: 196658942-196658942 on Build GRCh38
rs761071958 Chr.2: 196658835-196658835 on Build GRCh38
rs1040317729 Chr.2: 196658934-196658934 on Build GRCh38
rs557380307 Chr.2: 196658656-196658656 on Build GRCh38
rs981149759 Chr.2: 196658814-196658814 on Build GRCh38
rs77870358 Chr.2: 196659098-196659098 on Build GRCh38
rs112342041 Chr.2: 196658998-196658998 on Build GRCh38
rs1030337343 Chr.2: 196659058-196659058 on Build GRCh38
rs1044689541 Chr.2: 196659003-196659003 on Build GRCh38
rs967467053 Chr.2: 196658661-196658661 on Build GRCh38
rs760963943 Chr.2: 196658849-196658849 on Build GRCh38
rs893403813 Chr.2: 196658945-196658945 on Build GRCh38
rs776925508 Chr.2: 196658892-196658892 on Build GRCh38
rs1034647024 Chr.2: 196659068-196659068 on Build GRCh38
rs377252880 Chr.2: 196658749-196658749 on Build GRCh38
rs564146643 Chr.2: 196658948-196658948 on Build GRCh38
rs745461501 Chr.2: 196658834-196658834 on Build GRCh38
rs548682205 Chr.2: 196658708-196658708 on Build GRCh38
rs755494184 Chr.2: 196658800-196658800 on Build GRCh38
rs887978758 Chr.2: 196658971-196658971 on Build GRCh38
rs559121992 Chr.2: 196658733-196658733 on Build GRCh38
rs561665983 Chr.2: 196658841-196658841 on Build GRCh38
rs761147868 Chr.2: 196658771-196658771 on Build GRCh38

View Full Product Details