Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196987582-196988102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD44
Entrez Gene ID
91526
Gene Name
ankyrin repeat domain 44
Gene Aliases
PP6-ARS-B
UniGene
Hs.432706
Gene Chromosome Location
Chr.2: 196962617-197310797 on Build GRCh38
Transcripts
NM_001195144.1 XM_005246945.1 XM_005246946.1 XM_005246947.2 XM_005246948.2 XM_006712838.3 XM_011512148.2 XM_017005252.1

SNP Detail

SNP ID Chromosome Location
rs1036736119 Chr.2: 196987851-196987851 on Build GRCh38
rs970826620 Chr.2: 196987974-196987974 on Build GRCh38
rs780679821 Chr.2: 196988038-196988038 on Build GRCh38
rs979814644 Chr.2: 196987763-196987768 on Build GRCh38
rs113061711 Chr.2: 196987662-196987662 on Build GRCh38
rs371418842 Chr.2: 196987712-196987712 on Build GRCh38
rs113410782 Chr.2: 196987978-196987978 on Build GRCh38
rs936974830 Chr.2: 196987745-196987745 on Build GRCh38
rs924186945 Chr.2: 196987719-196987719 on Build GRCh38
rs111681104 Chr.2: 196988072-196988072 on Build GRCh38
rs990854401 Chr.2: 196987677-196987677 on Build GRCh38
rs1024997220 Chr.2: 196987963-196987963 on Build GRCh38
rs769098088 Chr.2: 196987781-196987781 on Build GRCh38
rs958284249 Chr.2: 196988041-196988041 on Build GRCh38
rs147195565 Chr.2: 196987980-196987981 on Build GRCh38
rs918014864 Chr.2: 196987665-196987665 on Build GRCh38
rs960950027 Chr.2: 196987995-196987995 on Build GRCh38
rs989727604 Chr.2: 196988064-196988064 on Build GRCh38
rs979565639 Chr.2: 196988081-196988081 on Build GRCh38
rs543924542 Chr.2: 196987825-196987825 on Build GRCh38
rs191427127 Chr.2: 196987947-196987947 on Build GRCh38
rs566037307 Chr.2: 196987704-196987704 on Build GRCh38
rs970689246 Chr.2: 196987708-196987708 on Build GRCh38
rs1005076918 Chr.2: 196987979-196987980 on Build GRCh38
rs897054588 Chr.2: 196987856-196987856 on Build GRCh38
rs995400534 Chr.2: 196987892-196987892 on Build GRCh38
rs916627063 Chr.2: 196987744-196987744 on Build GRCh38
rs556861786 Chr.2: 196987912-196987912 on Build GRCh38
rs554560155 Chr.2: 196987740-196987740 on Build GRCh38
rs398080958 Chr.2: 196987848-196987849 on Build GRCh38
rs1030969873 Chr.2: 196988021-196988021 on Build GRCh38
rs1012337719 Chr.2: 196988042-196988042 on Build GRCh38
rs1023819814 Chr.2: 196988067-196988067 on Build GRCh38
rs923983945 Chr.2: 196987764-196987764 on Build GRCh38
rs139549096 Chr.2: 196987765-196987765 on Build GRCh38
rs142947195 Chr.2: 196987958-196987958 on Build GRCh38
rs75088057 Chr.2: 196988070-196988070 on Build GRCh38
rs911721011 Chr.2: 196987853-196987856 on Build GRCh38
rs894955286 Chr.2: 196987952-196987952 on Build GRCh38
rs58942301 Chr.2: 196987840-196987841 on Build GRCh38
rs1054455224 Chr.2: 196987769-196987769 on Build GRCh38
rs932161508 Chr.2: 196987793-196987793 on Build GRCh38
rs538957192 Chr.2: 196987615-196987615 on Build GRCh38
rs1027319707 Chr.2: 196987950-196987950 on Build GRCh38
rs1035267679 Chr.2: 196987979-196987980 on Build GRCh38
rs1007878233 Chr.2: 196987844-196987844 on Build GRCh38
rs959475483 Chr.2: 196987649-196987649 on Build GRCh38
rs1049475541 Chr.2: 196987824-196987824 on Build GRCh38
rs776990284 Chr.2: 196987954-196987954 on Build GRCh38
rs397721390 Chr.2: 196987849-196987850 on Build GRCh38
rs944561383 Chr.2: 196987865-196987865 on Build GRCh38
rs559638024 Chr.2: 196988053-196988053 on Build GRCh38

View Full Product Details